This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00138061_cn
See other KCNE3 CNV Assays ›
Gene Symbol
KCNE3
Assay Reference Genome
Location

Chr.11:74457610 on build GRCh38
Cytoband
11q13.4
Assay ID Hs00138061_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

10008

Gene Name:

potassium voltage-gated channel subfamily E regulatory subunit 3

Gene Aliases:

HOKPP, HYPP, MiRP2

Location:

Chr.11:74454841-74467729 on Build GRCh38

Assay Gene Location:

Overlaps Intron 5 - Exon 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
KCNE3 NM_005472.4 NP_005463.1
XM_011544713.2 XP_011543015.1
XM_017017047.1 XP_016872536.1
XM_017017048.1 XP_016872537.1
XM_017017049.1 XP_016872538.1
XM_017017050.1 XP_016872539.1
XM_017017051.1 XP_016872540.1
XM_017017052.1 XP_016872541.1
AF076531.1 AAD28089.1
AF302494.1 AAG16255.1
AK311876.1
BC110612.2 AAI10613.1
BC113743.1 AAI13744.1
CD709836.1
CR457209.1 CAG33490.1
CR936625.1 CAI56768.1
DQ192291.1 ABB00304.1
DQ192292.1 ABB00305.1
DR006135.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1041154 Chr.11:74403756 - 74466864 on Build GRCh38 Loss KCNE3
esv34979 Chr.11:74167643 - 74822407 on Build GRCh38 Gain RNF169 POLD3 CHRDL2 LIPT2 LOC101928580 MIR4696 LOC100287896 C2CD3 MIR548AL P4HA3 PGM2L1 PPME1 KCNE3
esv3626840 Chr.11:74371082 - 74464389 on Build GRCh38 Gain PGM2L1 KCNE3 MIR548AL
esv2759837 Chr.11:74131564 - 74846552 on Build GRCh38 Gain RNF169 POLD3 CHRDL2 LIPT2 LOC101928580 MIR4696 LOC100287896 C2CD3 MIR548AL XRRA1 P4HA3 PGM2L1 PPME1 KCNE3
nsv555443 Chr.11:74398121 - 74469444 on Build GRCh38 Loss PGM2L1 KCNE3 MIR548AL
nsv832209 Chr.11:74444838 - 74618074 on Build GRCh38 Gain POLD3 LIPT2 KCNE3 LOC100287896
dgv209e214 Chr.11:74225600 - 74828943 on Build GRCh38 Gain P4HA3 RNF169 PGM2L1 PPME1 POLD3 CHRDL2 LIPT2 KCNE3 LOC101928580 MIR4696 LOC100287896 MIR548AL
nsv393 Chr.11:74424817 - 74470372 on Build GRCh38 Deletion KCNE3
nsv825994 Chr.11:74225622 - 74823410 on Build GRCh38 Gain P4HA3 RNF169 PGM2L1 PPME1 POLD3 CHRDL2 LIPT2 KCNE3 LOC101928580 MIR4696 LOC100287896 MIR548AL

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

voltage-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

regulation of membrane repolarization
potassium ion transmembrane transport
positive regulation of voltage-gated calcium channel activity
negative regulation of delayed rectifier potassium channel activity
voltage-gated potassium channel activity
protein binding
potassium channel regulator activity
ion channel binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline