This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00215510_cn
See other SLC2A4RG CNV Assays ›
Gene Symbol
SLC2A4RG
Assay Reference Genome
Location

Chr.20:63743779 on build GRCh38
Cytoband
20q13.33
Assay ID Hs00215510_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

56731

Gene Name:

SLC2A4 regulator

Gene Aliases:

GEF, HDBP-1, HDBP1, Si-1-2, Si-1-2-19

Location:

Chr.20:63739858-63744050 on Build GRCh38

Assay Gene Location:

Within Exon 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SLC2A4RG NM_020062.3 8 2008 NP_064446.2
AV659783.2 1 344
BM983611.1 1 289

Target Gene Details

Entrez Gene ID:

140685

Gene Name:

zinc finger and BTB domain containing 46

Gene Aliases:

BTBD4, BZEL, RINZF, ZNF340, dJ583P15.7, dJ583P15.8

Location:

Chr.20:63743668-63832332 on Build GRCh38

Assay Gene Location:

Within Exon 12
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
ZBTB46 NM_025224.3 5 5122 NP_079500.2
XM_005260195.4 5 5202 XP_005260252.1
XM_005260196.3 5 5018 XP_005260253.1
XM_005260197.4 5 5338 XP_005260254.1
XM_005260198.4 5 5001 XP_005260255.1
XM_006723700.3 5 5034 XP_006723763.1
XM_011528548.2 5 5302 XP_011526850.1
AK023564.1 1 2106
AK126720.1 4 4139

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2722959 Chr.20:63715664 - 63806918 on Build GRCh38 Deletion SLC2A4RG ZGPAT LIME1 ZBTB46
nsv953308 Chr.20:63650148 - 63831847 on Build GRCh38 Deletion ZBTB46-AS1 SLC2A4RG ZGPAT RTEL1-TNFRSF6B LIME1 TNFRSF6B STMN3 ZBTB46 ARFRP1 RTEL1
nsv828803 Chr.20:63743853 - 63744438 on Build GRCh38 Gain SLC2A4RG ZBTB46
nsv828802 Chr.20:63743820 - 63744589 on Build GRCh38 Loss SLC2A4RG ZBTB46
nsv1160690 Chr.20:63639552 - 63744287 on Build GRCh38 Deletion SLC2A4RG ZGPAT RTEL1-TNFRSF6B LIME1 TNFRSF6B STMN3 ZBTB46 ARFRP1 RTEL1
nsv428381 Chr.20:63583375 - 64334167 on Build GRCh38 Gain+Loss MIR941-3 SLC2A4RG ABHD16B LINC00266-1 STMN3 ZBTB46 ARFRP1 ZBTB46-AS1 PRPF6 GMEB2 LOC100505771 TNFRSF6B UCKL1 LINC00176 MIR1914 MIR6813 UCKL1-AS1 MIR941-1 PCMTD2 LKAAEAR1 DNAJC5 LIME1 TPD52L2 NPBWR2 ZNF512B MIR941-5 OPRL1 SOX18 MIR647 MYT1 ZGPAT RTEL1-TNFRSF6B C20orf181 MIR941-4 TCEA2 RGS19 SAMD10 MIR941-2 RTEL1
esv7227 Chr.20:63715550 - 63807161 on Build GRCh38 Loss SLC2A4RG ZGPAT LIME1 ZBTB46
nsv1059142 Chr.20:63731978 - 63798885 on Build GRCh38 Gain SLC2A4RG ZGPAT LIME1 ZBTB46
nsv470563 Chr.20:63486715 - 63793308 on Build GRCh38 Loss PPDPF SLC2A4RG FNDC11 LIME1 PTK6 SRMS STMN3 ZBTB46 ARFRP1 HELZ2 ZGPAT RTEL1-TNFRSF6B GMEB2 LOC100505771 TNFRSF6B EEF1A2 RTEL1
dgv7684n54 Chr.20:63738644 - 63789659 on Build GRCh38 Loss SLC2A4RG LIME1 ZBTB46
esv2758806 Chr.20:63470996 - 63745014 on Build GRCh38 Gain+Loss PPDPF SLC2A4RG FNDC11 LIME1 PTK6 SRMS KCNQ2 STMN3 ZBTB46 ARFRP1 HELZ2 ZGPAT RTEL1-TNFRSF6B GMEB2 LOC100505771 TNFRSF6B EEF1A2 RTEL1

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

transcription cofactor non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of granulocyte differentiation
negative regulation of macrophage differentiation
negative regulation of monocyte differentiation
negative regulation of dendritic cell differentiation
positive regulation of dendritic cell differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
nucleic acid binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline