This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00252181_cn
See other MIB2 CNV Assays ›
Gene Symbol
MIB2
Assay Reference Genome
Location

Chr.1:1616533 on build GRCh38
Cytoband
1p36.33
Assay ID Hs00252181_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

142678

Gene Name:

mindbomb E3 ubiquitin protein ligase 2

Gene Aliases:

ZZANK1, ZZZ5

Location:

Chr.1:1615188-1630610 on Build GRCh38

Assay Gene Location:

Overlaps Intron 1 - Exon 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MIB2 NM_001170686.1 NP_001164157.1
NM_001170687.1 NP_001164158.1
NM_001170688.1 NP_001164159.1
NM_001170689.1 NP_001164160.1
NM_080875.2 NP_543151.2
NR_033183.1
XM_006710372.1 XP_006710435.1
XM_011540731.2 XP_011539033.1
XM_011540736.2 XP_011539038.1
XM_011540737.2 XP_011539039.1
XM_011540741.2 XP_011539043.1
XM_011540742.2 XP_011539044.1
XM_017000349.1 XP_016855838.1
XM_017000350.1 XP_016855839.1
XM_017000351.1 XP_016855840.1
XM_017000352.1 XP_016855841.1
XM_017000353.1 XP_016855842.1
XM_017000354.1 XP_016855843.1
XM_017000355.1 XP_016855844.1
XM_017000356.1 XP_016855845.1
XM_017000357.1 XP_016855846.1
XM_017000358.1 XP_016855847.1
XM_017000359.1 XP_016855848.1
XM_017000360.1 XP_016855849.1
XM_017000361.1 XP_016855850.1
XM_017000362.1 XP_016855851.1
XM_017000363.1 XP_016855852.1
XM_017000364.1 XP_016855853.1
XM_017000365.1 XP_016855854.1
XM_017000366.1 XP_016855855.1
AB074480.1 BAB92950.1
AB076691.1 BAC00992.1
AB076692.1 BAC00993.1
AB076693.1 BAC00994.1
AB097000.1 BAC77353.1
AK098785.1 BAC05413.1
AK122751.1
AK122863.1
AK123400.1
AK128167.1
AK128553.1
AK293642.1 1 225
AK302757.1
AK309362.1 1 223
BC016490.1 AAH16490.1
BC037542.1 AAH37542.1
DA391220.1
DC402226.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion GNB1 MIB2 SLC35E2B FNDC10 CDK11B CDK11A MMP23B MMP23A SLC35E2 NADK
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss SSU72 MIB2 SLC35E2B FNDC10 CDK11B CDK11A MMP23B MMP23A SLC35E2
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss LINC00115 MIR200A TMEM88B NOC2L FAM87B LOC148413 SAMD11 UBE2J2 PUSL1 LOC102724312 PERM1 TMEM240 SCNN1D TNFRSF4 KLHL17 AGRN TNFRSF18 SLC35E2 HES4 LOC100287934 MIR6726 ATAD3B MIR429 FNDC10 CDK11B RNF223 CDK11A LINC01128 MXRA8 TAS1R3 TTLL10-AS1 MIB2 LOC107985728 AURKAIP1 MMP23A NADK DVL1 PLEKHN1 CPTP ATAD3C ANKRD65 LOC100288175 MMP23B CCNL2 SSU72 LOC100288069 SLC35E2B ACAP3 SDF4 VWA1 FAM132A FAM41C LOC284600 CPSF3L LOC105378948 MIR6808 TTLL10 MIR6727 B3GALT6 MIR200B LOC107985729 LINC01342 C1orf159 ISG15 MRPL20 LOC100130417 ATAD3A
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss MIR200A LOC100129534 TMEM88B OR4F29 NOC2L UBE2J2 PERM1 OR4F5 SCNN1D LOC100134822 AGRN TNFRSF18 SLC35E2 LOC105378591 CALML6 LOC100287934 MIR6726 LOC105378592 MIR429 FNDC10 CDK11B TAS1R3 MORN1 LOC105378589 MIB2 SKI MIR6859-2 DVL1 PLEKHN1 FAAP20 LOC100133331 ATAD3C ANKRD65 PRKCZ CCNL2 MTND1P23 LOC100288069 SLC35E2B FAM138A WASH7P LOC100506504 VWA1 LOC284600 CPSF3L LOC105378948 B3GALT6 MIR200B LINC01342 C1orf159 ISG15 LOC100130417 ATAD3A DDX11L1 LINC00115 CFAP74 FAM87B LOC148413 SAMD11 PUSL1 LOC102724312 OR4F16 MIR1302-2 TMEM240 TNFRSF4 KLHL17 HES4 ATAD3B RNF223 CDK11A MTND2P28 LINC01128 MXRA8 TTLL10-AS1 LOC107985728 GABRD AURKAIP1 MMP23A NADK CPTP LOC102725121 LOC100288175 MMP23B SSU72 GNB1 LOC101928626 ACAP3 SDF4 LOC100132287 FAM132A FAM41C MIR6859-1 LOC729737 LOC105378947 MIR6808 TTLL10 MIR6727 MIR6723 LOC107984841 LOC107985729 LOC105378949 TMEM52 MRPL20
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication SSU72 GNB1 MIB2 SLC35E2B FNDC10 CDK11B CDK11A MMP23B MMP23A SLC35E2 NADK
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion DVL1 MIR200A CPTP TMEM88B ATAD3C LOC148413 UBE2J2 ANKRD65 MMP23B CCNL2 SSU72 PUSL1 LOC102724312 TMEM240 ACAP3 SCNN1D SDF4 VWA1 TNFRSF4 FAM132A CPSF3L TNFRSF18 MIR6726 MIR6808 TTLL10 MIR6727 ATAD3B MIR429 B3GALT6 FNDC10 CDK11B MIR200B MXRA8 TAS1R3 TTLL10-AS1 LOC107985729 LINC01342 C1orf159 MIB2 MRPL20 AURKAIP1 ATAD3A
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss SSU72 MIB2 SLC35E2B FNDC10 CDK11B CDK11A MMP23B MMP23A SLC35E2 NADK
nsv545017 Chr.1:1615322 - 1663759 on Build GRCh38 Loss MIB2 SLC35E2B CDK11B MMP23B
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss CFAP74 TMEM88B ATAD3C LOC148413 ANKRD65 PRKCZ MMP23B CCNL2 SSU72 LOC102724312 GNB1 SLC35E2B TMEM240 VWA1 SLC35E2 LOC105378591 CALML6 ATAD3B FNDC10 CDK11B CDK11A LOC107985729 LOC105378589 MIB2 LOC105378949 TMEM52 GABRD MRPL20 ATAD3A MMP23A NADK

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
Notch signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
ubiquitin-protein transferase activity
signal transducer activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline