This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00278547_cn
See other RPF1 CNV Assays ›
Gene Symbol
RPF1
Assay Reference Genome
Location

Chr.1:84479465 on build GRCh38
Cytoband
1p22.3
Assay ID Hs00278547_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

80135

Gene Name:

ribosome production factor 1 homolog

Gene Aliases:

BXDC5

Location:

Chr.1:84479237-84498350 on Build GRCh38

Assay Gene Location:

Overlaps Exon 1 - Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
RPF1 NM_025065.6 NP_079341.2
AF322053.1 AAL37236.1
AK022537.1 BAB14086.1
BC016051.1 AAH16051.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv830437 Chr.1:84371841 - 84535586 on Build GRCh38 Gain+Loss RPF1 UOX SPATA1 DNASE2B GNG5

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

ribosomal large subunit assembly
maturation of 5.8S rRNA
maturation of LSU-rRNA
RNA binding
protein binding
rRNA primary transcript binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0