This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00326552_cn
See other VSTM1 CNV Assays ›
Gene Symbol
VSTM1
Assay Reference Genome
Location

Chr.19:54041939 on build GRCh38
Cytoband
19q13.42
Assay ID Hs00326552_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

284415

Gene Name:

V-set and transmembrane domain containing 1

Gene Aliases:

SIRL-1, SIRL1, UNQ3033

Location:

Chr.19:54040825-54063966 on Build GRCh38

Assay Gene Location:

Overlaps Intron 10 - Intron 11
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
VSTM1 NM_001288791.1 NP_001275720.1
NM_001288792.1 NP_001275721.1
NM_001288793.1 NP_001275722.1
NM_198481.3 NP_940883.2
NR_110142.1
XM_011526845.1 XP_011525147.1
XM_011526846.1 XP_011525148.1
XM_011526847.1 XP_011525149.1
XM_011526848.1 XP_011525150.1
XM_011526849.1 XP_011525151.1
XM_017026666.1 XP_016882155.1
AY358542.1 AAQ88906.1
BC100943.2 AAI00944.1
BG189223.1
BQ446114.1
DQ479397.1
FJ584316.1
FJ584317.1
FJ584318.1
FJ584319.1
FJ584320.1
FJ882051.1
FN398145.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv458782 Chr.19:53961144 - 54093354 on Build GRCh38 Gain MIR935 TARM1 CACNG6 VSTM1 CACNG8
nsv510773 Chr.19:54017180 - 54132236 on Build GRCh38 Deletion OSCAR TARM1 NDUFA3 PRPF31 TFPT VSTM1
nsv1063127 Chr.19:53926876 - 54068977 on Build GRCh38 Gain MIR935 CACNG7 CACNG6 VSTM1 CACNG8
nsv1057516 Chr.19:53986233 - 54046901 on Build GRCh38 Gain CACNG6 VSTM1 CACNG8
esv2718812 Chr.19:53184636 - 54322450 on Build GRCh38 Deletion MIR523 MIR527 MIR1283-1 MIR518B MIR526A1 MIR4752 NLRP12 MIR520D LOC105372457 NDUFA3 MYADM TPM3P9 ZNF677 MIR520C MIR517C MIR522 LILRB2 TSEN34 MIR519D MBOAT7 MIR518C MIR526A2 MIR518D MIR512-1 MIR517A MIR373 MIR519B MIR520B MIR518E RPS9 MIR516B2 LILRB5 MIR516A2 MIR371B MIR518F MIR516B1 MIR520A MIR521-2 MIR518A2 MIR1283-2 ZNF845 LOC284379 BIRC8 MIR516A1 CACNG6 LILRA6 MIR526B CNOT3 ZNF813 TMC4 TARM1 MIR1323 MIR517B MIR520F FAM90A27P MIR519C VN1R2 MIR512-2 MIR518A1 ZNF761 MIR371A MIR520H LILRA5 VN1R4 DPRX MIR935 PRKCG ZNF525 MIR521-1 LENG1 MIR520G PRPF31 MIR520E VSTM1 MIR525 OSCAR MIR519A2 CACNG7 ZNF818P MIR372 MIR515-2 MIR519A1 ZNF665 ZNF765 MIR515-1 TFPT LILRB3 ZNF331 MIR519E MIR524 CACNG8 MIR498
nsv833877 Chr.19:53951529 - 54117323 on Build GRCh38 Loss MIR935 OSCAR TARM1 NDUFA3 PRPF31 CACNG6 TFPT VSTM1 CACNG8

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

immune system process
cytokine activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline