This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00417971_cn
See other FAM118B CNV Assays ›
Gene Symbol
FAM118B
Assay Reference Genome
Location

Chr.11:126256802 on build GRCh38
Cytoband
11q24.2
Assay ID Hs00417971_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

79607

Gene Name:

family with sequence similarity 118 member B

Gene Aliases:

-

Location:

Chr.11:126211414-126264540 on Build GRCh38

Assay Gene Location:

Within Exon 7
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FAM118B NM_024556.3 7 1115 NP_078832.1
XM_005271665.3 7 1425 XP_005271722.1
XM_011542977.2 6 1046 XP_011541279.1
XM_011542978.2 7 1471 XP_011541280.1
XM_017018285.1 6 1046 XP_016873774.1
XM_017018286.1 7 1471 XP_016873775.1
AK024756.1 7 1039 BAB14987.1
AK225081.1 7 1039
AK315615.1 8 1204
BC001340.1 6 1020 AAH01340.1
BC001647.2 7 1034 AAH01647.1
CR457334.1 5 932 CAG33615.1

Target Gene Details

Entrez Gene ID:

6734

Gene Name:

SRP receptor alpha subunit

Gene Aliases:

DP, SRPR, Sralpha

Location:

Chr.11:126255151-126268982 on Build GRCh38

Assay Gene Location:

Within Exon 15
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SRPRA XM_017018179.1 14 2076 XP_016873668.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv556529 Chr.11:126204297 - 126264724 on Build GRCh38 Loss RPUSD4 SRPRA FAM118B

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

G-protein protein-binding activity modulator

Gene Ontology Categories:

Function(s) Process(es)

Cajal body organization
cotranslational protein targeting to membrane
SRP-dependent cotranslational protein targeting to membrane
IRE1-mediated unfolded protein response
protein binding
GTPase activity
signal recognition particle binding
GTP binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline