This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00465491_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1254869 on build GRCh38
Cytoband
1p36.33
Assay ID Hs00465491_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Exon 10
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 7 1303 NP_477515.2
NM_194315.1 8 1351 NP_919296.1
NM_194457.1 6 1172 NP_919439.1
NM_194458.1 7 1432 NP_919440.1
XM_005244718.3 7 1581 XP_005244775.1
XM_005244719.4 7 1469 XP_005244776.1
XM_006710333.3 7 1748 XP_006710396.1
XM_011540613.2 6 1176 XP_011538915.1
XM_011540614.2 7 1534 XP_011538916.1
XM_011540616.2 8 1543 XP_011538918.1
XM_011540617.2 7 1310 XP_011538919.1
XM_017000238.1 8 1414 XP_016855727.1
XM_017000239.1 6 1632 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 7 1213 BAB71086.1
AK075017.1 7 1282 BAC11355.1
AK098803.1 2 669
AK291935.1
BC094777.1 7 1302 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication ACAP3 CPSF3L MIR200B LINC01342 PUSL1 C1orf159 TAS1R3 SCNN1D TNFRSF4 UBE2J2 MIR6727 TTLL10 TNFRSF18 AGRN CPTP B3GALT6 MIR429 TTLL10-AS1 MIR200A LOC105378948 FAM132A RNF223 LOC100288175 MIR6726 SDF4
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss MMP23A ACAP3 SAMD11 FNDC10 LOC100130417 CPSF3L LOC102724312 TMEM240 VWA1 LINC01342 CDK11A PUSL1 C1orf159 SLC35E2B TAS1R3 ANKRD65 SCNN1D ATAD3C TNFRSF18 LOC107985728 CPTP PLEKHN1 B3GALT6 MIR429 TTLL10-AS1 RNF223 SSU72 LOC100288175 FAM41C NOC2L LOC107985729 LINC00115 MIR6726 DVL1 SDF4 LOC100287934 CDK11B ATAD3A KLHL17 MIB2 MIR200B LOC284600 TMEM88B ISG15 TNFRSF4 UBE2J2 NADK FAM87B MIR6727 TTLL10 LINC01128 HES4 ATAD3B AGRN MIR200A SLC35E2 LOC105378948 AURKAIP1 LOC148413 FAM132A MRPL20 LOC100288069 CCNL2 MMP23B MXRA8 PERM1 MIR6808
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss SAMD11 LOC100130417 LOC102724312 TMEM240 LINC01342 CDK11A PUSL1 C1orf159 SLC35E2B ANKRD65 SCNN1D LOC729737 GNB1 MTND1P23 MIR6859-2 PRKCZ TNFRSF18 LOC107985728 CPTP B3GALT6 MIR429 TTLL10-AS1 LOC105378592 SSU72 FAM138A DDX11L1 LOC101928626 FAM41C LOC100133331 MIR6726 LOC100287934 CDK11B ATAD3A KLHL17 MIB2 OR4F29 LOC102725121 CFAP74 LOC284600 TMEM88B LOC100506504 ISG15 LOC105378589 SKI TNFRSF4 MIR1302-2 FAM87B MIR6727 TTLL10 LINC01128 ATAD3B AGRN LOC105378591 MIR200A LOC105378948 LOC148413 TMEM52 WASH7P PERM1 MORN1 CALML6 MMP23A ACAP3 FNDC10 CPSF3L VWA1 MIR6859-1 TAS1R3 LOC100132287 LOC105378949 LOC107984841 ATAD3C PLEKHN1 RNF223 LOC100288175 NOC2L LOC107985729 LINC00115 DVL1 SDF4 LOC100134822 OR4F5 MIR200B FAAP20 UBE2J2 OR4F16 LOC105378947 NADK HES4 SLC35E2 GABRD AURKAIP1 FAM132A MRPL20 LOC100129534 MIR6723 LOC100288069 CCNL2 MMP23B MXRA8 MTND2P28 MIR6808
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss FAM132A ACAP3 SCNN1D UBE2J2 B3GALT6 MIR6726 SDF4
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss KLHL17 SAMD11 LOC100130417 MIR200B LINC01342 LOC284600 ISG15 C1orf159 TNFRSF4 UBE2J2 FAM87B TTLL10 LINC01128 TNFRSF18 HES4 AGRN LOC107985728 PLEKHN1 B3GALT6 MIR429 TTLL10-AS1 MIR200A LOC105378948 FAM132A RNF223 LOC100288175 FAM41C NOC2L LINC00115 SDF4 PERM1
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion ACAP3 MIB2 FNDC10 CPSF3L LOC102724312 MIR200B TMEM240 VWA1 LINC01342 TMEM88B PUSL1 C1orf159 TAS1R3 ANKRD65 SCNN1D TNFRSF4 UBE2J2 ATAD3C MIR6727 TTLL10 TNFRSF18 ATAD3B CPTP B3GALT6 MIR429 TTLL10-AS1 MIR200A AURKAIP1 LOC148413 FAM132A MRPL20 SSU72 CCNL2 LOC107985729 MMP23B MXRA8 MIR6726 DVL1 SDF4 CDK11B MIR6808 ATAD3A
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain KLHL17 ACAP3 SAMD11 LOC100130417 CPSF3L MIR200B LINC01342 LOC284600 TMEM88B ISG15 PUSL1 C1orf159 TAS1R3 ANKRD65 SCNN1D TNFRSF4 UBE2J2 MIR6727 TTLL10 TNFRSF18 HES4 AGRN LOC107985728 CPTP PLEKHN1 B3GALT6 MIR429 TTLL10-AS1 MIR200A LOC105378948 AURKAIP1 LOC148413 FAM132A MRPL20 RNF223 LOC100288175 NOC2L CCNL2 MXRA8 MIR6726 DVL1 SDF4 PERM1 MIR6808
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain TTLL10 TNFRSF18 MIR200B B3GALT6 MIR429 TTLL10-AS1 LINC01342 MIR200A LOC105378948 FAM132A RNF223 C1orf159 SCNN1D TNFRSF4 UBE2J2 SDF4
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss ACAP3 TTLL10 TNFRSF18 MIR200B B3GALT6 MIR429 TTLL10-AS1 LINC01342 MIR200A FAM132A RNF223 C1orf159 SCNN1D TNFRSF4 UBE2J2 MIR6726 SDF4
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain MIR6727 ACAP3 TTLL10 TNFRSF18 CPSF3L MIR200B B3GALT6 MIR429 TTLL10-AS1 MIR200A FAM132A PUSL1 SCNN1D TNFRSF4 UBE2J2 MIR6726 SDF4

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline