This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00472618_cn
See other CPSF3L CNV Assays ›
Gene Symbol
CPSF3L
Assay Reference Genome
Location

Chr.1:1320472 on build GRCh38
Cytoband
1p36.33
Assay ID Hs00472618_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

54973

Gene Name:

cleavage and polyadenylation specific factor 3-like

Gene Aliases:

CPSF73L, INT11, INTS11, RC-68, RC68

Location:

Chr.1:1311585-1324687 on Build GRCh38

Assay Gene Location:

Overlaps Intron 5 - Exon 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CPSF3L NM_001256456.1 NP_001243385.1
NM_001256460.1 NP_001243389.1
NM_001256462.1 NP_001243391.1
NM_001256463.1 NP_001243392.1
NM_017871.5 NP_060341.2
XM_011541647.1 XP_011539949.1
XM_011541648.1 XP_011539950.1
XM_011541650.1 XP_011539952.1
XM_017001557.1 XP_016857046.1
XM_017001558.1 XP_016857047.1
AK021939.1 BAB13943.1
AK022675.1 2 653
AK023356.1 BAB14541.1
AK027767.1
AK056652.1
AK058018.1
AK092190.1
AK291387.1
AK297350.1
AL136813.1 CAB66747.1
BC007978.2 AAH07978.1
BC008041.2 AAH08041.1
BC020199.1 AAH20199.1
CR533557.1 CAG38588.1
DC295791.1
DC299237.1
DC299993.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss CCNL2 NADK DDX11L1 ISG15 LOC100130417 SLC35E2B FAM87B LOC148413 TNFRSF18 TTLL10 TMEM88B CALML6 KLHL17 FAM41C MIR6723 UBE2J2 PERM1 PLEKHN1 LINC00115 SDF4 FAM132A LOC102724312 LOC100288069 FAM138A OR4F16 MIR200B RNF223 TTLL10-AS1 LOC729737 C1orf159 AURKAIP1 LINC01128 MIR6859-2 MIB2 B3GALT6 CPSF3L MIR6727 MMP23B MORN1 SSU72 LOC107985728 HES4 LOC101928626 LOC100287934 SAMD11 PRKCZ FNDC10 OR4F5 ATAD3B MIR1302-2 GNB1 CPTP LOC105378948 MXRA8 LOC100506504 LOC284600 MTND1P23 CDK11A TAS1R3 PUSL1 TNFRSF4 TMEM52 MRPL20 MIR6808 CFAP74 DVL1 LOC105378589 LOC100129534 ATAD3A LOC102725121 MIR200A SKI LOC105378947 MMP23A LOC100132287 ACAP3 OR4F29 MIR429 SLC35E2 LOC105378592 FAAP20 AGRN LOC105378949 GABRD LOC105378591 ATAD3C VWA1 NOC2L MTND2P28 LOC100288175 LOC100134822 LOC100133331 ANKRD65 SCNN1D MIR6859-1 WASH7P LINC01342 LOC107984841 TMEM240 LOC107985729 MIR6726 CDK11B
nsv544971 Chr.1:1294063 - 1351426 on Build GRCh38 Loss MIR6726 CPSF3L MIR6727 CPTP TAS1R3 PUSL1 ACAP3 MIR6808 DVL1
nsv544969 Chr.1:1290683 - 1374597 on Build GRCh38 Loss MIR6726 CPSF3L MIR6727 CPTP SCNN1D TAS1R3 MXRA8 PUSL1 ACAP3 MIR6808 AURKAIP1 DVL1
nsv997291 Chr.1:1306835 - 1376336 on Build GRCh38 Gain CPSF3L MIR6727 CPTP TAS1R3 MXRA8 PUSL1 ACAP3 MIR6808 AURKAIP1 DVL1
nsv470680 Chr.1:1070426 - 1378635 on Build GRCh38 Loss MIR429 MIR200B RNF223 TTLL10-AS1 CPTP MXRA8 TAS1R3 C1orf159 PUSL1 TNFRSF18 TNFRSF4 MIR6808 TTLL10 AURKAIP1 DVL1 B3GALT6 CPSF3L MIR6727 SCNN1D MIR200A LINC01342 UBE2J2 MIR6726 ACAP3 SDF4 FAM132A
dgv6n100 Chr.1:1301750 - 1362354 on Build GRCh38 Gain+Loss CPSF3L MIR6727 CPTP TAS1R3 MXRA8 PUSL1 ACAP3 MIR6808 DVL1
esv3584992 Chr.1:1314419 - 1330342 on Build GRCh38 Gain CPSF3L CPTP
nsv1160788 Chr.1:1282706 - 1376336 on Build GRCh38 Deletion MIR6726 CPSF3L MIR6727 CPTP SCNN1D TAS1R3 MXRA8 PUSL1 ACAP3 MIR6808 AURKAIP1 DVL1
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication MIR429 MIR200B RNF223 TTLL10-AS1 CPTP LOC105378948 AGRN TAS1R3 LOC100288175 C1orf159 PUSL1 TNFRSF18 TNFRSF4 TTLL10 B3GALT6 CPSF3L MIR6727 SCNN1D MIR200A LINC01342 UBE2J2 MIR6726 ACAP3 SDF4 FAM132A
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss CCNL2 NADK CPTP LOC105378948 MXRA8 ISG15 LOC284600 LOC100130417 CDK11A SLC35E2B TAS1R3 FAM87B LOC148413 PUSL1 TNFRSF18 TNFRSF4 MRPL20 MIR6808 TTLL10 TMEM88B DVL1 ATAD3A KLHL17 MIR200A FAM41C UBE2J2 MMP23A PERM1 PLEKHN1 ACAP3 LINC00115 SDF4 FAM132A LOC102724312 LOC100288069 MIR429 MIR200B RNF223 TTLL10-AS1 SLC35E2 AGRN ATAD3C VWA1 NOC2L LOC100288175 C1orf159 AURKAIP1 LINC01128 MIB2 ANKRD65 B3GALT6 CPSF3L MIR6727 MMP23B SCNN1D SSU72 LOC107985728 LINC01342 HES4 TMEM240 LOC107985729 LOC100287934 SAMD11 MIR6726 FNDC10 ATAD3B CDK11B
esv3890636 Chr.1:1304555 - 1353203 on Build GRCh38 Loss CPSF3L MIR6727 CPTP TAS1R3 MXRA8 PUSL1 ACAP3 MIR6808 DVL1
nsv1000169 Chr.1:1281678 - 1362354 on Build GRCh38 Gain MIR6726 CPSF3L MIR6727 CPTP SCNN1D TAS1R3 MXRA8 PUSL1 ACAP3 MIR6808 DVL1
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion MIR429 CCNL2 MIR200B TTLL10-AS1 CPTP MXRA8 ATAD3C VWA1 TAS1R3 C1orf159 LOC148413 PUSL1 TNFRSF18 TNFRSF4 MRPL20 MIR6808 TTLL10 AURKAIP1 TMEM88B MIB2 ANKRD65 DVL1 B3GALT6 CPSF3L MIR6727 MMP23B SCNN1D ATAD3A MIR200A SSU72 LINC01342 TMEM240 LOC107985729 UBE2J2 MIR6726 FNDC10 ACAP3 SDF4 ATAD3B FAM132A LOC102724312 CDK11B
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss CPSF3L MIR6727 CPTP SCNN1D MXRA8 UBE2J2 MIR6726 TAS1R3 PUSL1 ACAP3 MIR6808 AURKAIP1 DVL1
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain MIR429 CCNL2 MIR200B RNF223 TTLL10-AS1 CPTP LOC105378948 MXRA8 AGRN ISG15 LOC284600 LOC100130417 NOC2L TAS1R3 LOC100288175 C1orf159 LOC148413 PUSL1 TNFRSF18 TNFRSF4 MRPL20 MIR6808 TTLL10 AURKAIP1 TMEM88B ANKRD65 DVL1 B3GALT6 CPSF3L MIR6727 SCNN1D KLHL17 MIR200A LOC107985728 LINC01342 HES4 SAMD11 UBE2J2 MIR6726 PERM1 PLEKHN1 ACAP3 SDF4 FAM132A
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain MIR429 B3GALT6 MIR200B CPSF3L MIR6727 TTLL10-AS1 SCNN1D MIR200A UBE2J2 MIR6726 PUSL1 ACAP3 TNFRSF18 TNFRSF4 SDF4 FAM132A TTLL10

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

snRNA processing
snRNA transcription from RNA polymerase II promoter
protein binding
hydrolase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0