Assay Details
Target Gene Details
Entrez Gene ID: | 221785 |
Gene Name: | zinc finger and SCAN domain containing 25 |
Gene Aliases: |
ZNF498 |
Location: |
Chr.7:99616600-99738689 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZSCAN25 | NM_145115.2 | 8 | 2307 | NP_660090.2 |
| XM_005250194.2 | 6 | 2079 | XP_005250251.1 | |
| XM_011515905.2 | 8 | 2618 | XP_011514207.1 | |
| XM_011515906.2 | 9 | 2725 | XP_011514208.1 | |
| XM_011515907.2 | 7 | 2562 | XP_011514209.1 | |
| XM_011515908.2 | 8 | 2632 | XP_011514210.1 | |
| XM_011515909.2 | XP_011514211.1 | |||
| XM_011515910.2 | XP_011514212.1 | |||
| XM_017011824.1 | 8 | 2354 | XP_016867313.1 | |
| XM_017011825.1 | 8 | 2659 | XP_016867314.1 | |
| XM_017011826.1 | 9 | 2336 | XP_016867315.1 | |
| XM_017011827.1 | XP_016867316.1 | |||
| AK057030.1 | 8 | 2284 | ||
| AK160369.1 | 3 | 2879 | BAD18712.1 | |
| BC089402.1 | 5 | 1849 | AAH89402.1 | |
| BX640720.1 | CAE45839.1 | |||
| BX648158.1 | 7 | 3690 |
More Information
Set Membership: |
|

Genomic Map