Assay Details
Target Gene Details
Entrez Gene ID: | 51313 |
Gene Name: | family with sequence similarity 198 member B |
Gene Aliases: |
AD021, AD036, C4orf18 |
Location: |
Chr.4:158124580-158173050 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM198B | NM_001031700.2 | 6 | 1932 | NP_001026870.2 |
| NM_001128424.1 | 5 | 1908 | NP_001121896.1 | |
| NM_016613.6 | 5 | 1953 | NP_057697.2 | |
| AF223468.1 | 4 | 895 | AAF64143.1 | |
| AK304112.1 | 4 | 833 | ||
| AK315768.1 | 4 | 1024 | ||
| AL832589.1 | 5 | 1779 | ||
| AL834177.1 | 5 | 1404 | CAD38874.1 | |
| AY358785.1 | 5 | 1870 | AAQ89145.1 | |
| BC017701.1 | 4 | 856 | AAH17701.1 | |
| BC043193.2 | 6 | 1932 | AAH43193.1 | |
| BC100016.1 | 4 | 792 | AAI00017.1 | |
| CR457196.1 | 4 | 682 | CAG33477.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1021067 | Chr.4:158051130 - 158254323 on Build GRCh38 | Gain |
|

Genomic Map