This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00517723_cn
See other RENBP CNV Assays ›
Gene Symbol
RENBP
Assay Reference Genome
Location

Chr.X:153944774 on build GRCh38
Cytoband
Xq28
Assay ID Hs00517723_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

5973

Gene Name:

renin binding protein

Gene Aliases:

RBP, RNBP

Location:

Chr.X:153935263-153944780 on Build GRCh38

Assay Gene Location:

Overlaps - Exon 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
RENBP NM_002910.5 NP_002901.2

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv33199 Chr.X:153065734 - 154591978 on Build GRCh38 Gain+Loss OPN1MW3 LOC105373383 DNASE1L1 MIR6858 PNCK NAA10 SRPK3 EMD DUSP9 ATP2B3 PNMA6A RPL10 ATP6AP1 MIR718 UBL4A OPN1MW2 FAM58A LOC105373378 TEX28 ARHGAP4 MAGEA1 SNORA70 TREX2 FAM223A PLXNA3 LAGE3 TKTL1 IRAK1 LOC107983954 FAM3A FAM50A ZFP92 IDH3G MIR3202-2 HAUS7 AVPR2 HCFC1-AS1 IKBKG BCAP31 OPN1LW PDZD4 TAZ HCFC1 LOC107987332 MECP2 MIR3202-1 FLNA ABCD1 L1CAM SLC10A3 OPN1MW ZNF275 PLXNB3 G6PD CTAG1A LOC105373386 LOC105373381 LOC105373377 BGN CH17-340M24.3 SLC6A8 TMEM187 GDI1 SSR4 LCA10 RENBP
nsv469867 Chr.X:153813281 - 154018926 on Build GRCh38 Loss MIR3202-2 NAA10 AVPR2 ARHGAP4 HCFC1-AS1 PDZD4 HCFC1 TMEM187 MIR3202-1 IRAK1 L1CAM LCA10 RENBP

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

epimerase/racemase isomerase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

N-acetylglucosamine metabolic process
UDP-N-acetylglucosamine biosynthetic process
N-acetylmannosamine metabolic process
regulation of blood pressure
negative regulation of endopeptidase activity
N-acetylneuraminate catabolic process
endopeptidase inhibitor activity
ATP binding
protein homodimerization activity
N-acylglucosamine 2-epimerase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0