This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00519354_cn
See other CD160 CNV Assays ›
Gene Symbol
CD160
Assay Reference Genome
Location

Chr.1:145728284 on build GRCh38
Cytoband
1q21.1
Assay ID Hs00519354_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

11126

Gene Name:

CD160 molecule

Gene Aliases:

BY55, NK1, NK28

Location:

Chr.1:145719312-145739288 on Build GRCh38

Assay Gene Location:

Overlaps Intron 2 - Exon 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CD160 NM_007053.3 NP_008984.1
NR_103845.1
XM_005272929.3 XP_005272986.1
XM_011509104.2 XP_011507406.1
AF060981.1 AAC72302.1
AK128370.1
BC014465.1 AAH14465.1
CD638675.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1015025 Chr.1:145642988 - 145851919 on Build GRCh38 Gain CD160 PDZK1 GPR89A MIR6736 PIAS3 POLR3C RNF115 NUDT17
esv3587422 Chr.1:145649153 - 145751059 on Build GRCh38 Gain CD160 PDZK1 GPR89A RNF115
esv3572342 Chr.1:145601946 - 145809476 on Build GRCh38 Gain CD160 PDZK1 GPR89A NBPF25P RNF115
nsv469752 Chr.1:145639438 - 145790042 on Build GRCh38 Loss CD160 PDZK1 GPR89A RNF115
nsv831392 Chr.1:145632374 - 145813165 on Build GRCh38 Gain CD160 PDZK1 GPR89A RNF115
esv3894089 Chr.1:145672100 - 145824903 on Build GRCh38 Loss CD160 PDZK1 POLR3C RNF115
dgv531n54 Chr.1:145689573 - 145824903 on Build GRCh38 Loss CD160 PDZK1 POLR3C RNF115
dgv532n54 Chr.1:145624561 - 145813526 on Build GRCh38 Gain CD160 PDZK1 GPR89A RNF115
dgv95e212 Chr.1:145631218 - 145812561 on Build GRCh38 Gain CD160 PDZK1 GPR89A RNF115
nsv470735 Chr.1:145687602 - 145809105 on Build GRCh38 Gain CD160 PDZK1 RNF115

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

cellular defense response
cell surface receptor signaling pathway
cell proliferation
regulation of immune response
defense response to Gram-negative bacterium
receptor activity
receptor binding
protein binding
MHC class I receptor activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0