This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00620345_cn
See other LIME1 CNV Assays ›
Gene Symbol
LIME1
Assay Reference Genome
Location

Chr.20:63739070 on build GRCh38
Cytoband
20q13.33
Assay ID Hs00620345_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

54923

Gene Name:

Lck interacting transmembrane adaptor 1

Gene Aliases:

LIME, dJ583P15.4

Location:

Chr.20:63735701-63739107 on Build GRCh38

Assay Gene Location:

Overlaps - Exon 7
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LIME1 NM_001305654.1 NP_001292583.1
NM_001305655.1 NP_001292584.1
NM_017806.3 NP_060276.2
AI763397.1
AK000413.1 BAA91148.1
AK054722.1
AY203957.1 AAP34480.1
BC017016.1 AAH17016.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2722959 Chr.20:63715664 - 63806918 on Build GRCh38 Deletion LIME1 SLC2A4RG ZGPAT ZBTB46
nsv953308 Chr.20:63650148 - 63831847 on Build GRCh38 Deletion LIME1 RTEL1 ZBTB46-AS1 ARFRP1 SLC2A4RG ZGPAT RTEL1-TNFRSF6B STMN3 ZBTB46 TNFRSF6B
nsv586615 Chr.20:63734204 - 63741603 on Build GRCh38 Gain LIME1 SLC2A4RG ZGPAT
dgv7684n54 Chr.20:63738644 - 63789659 on Build GRCh38 Loss LIME1 SLC2A4RG ZBTB46
nsv586643 Chr.20:63738872 - 63740448 on Build GRCh38 Gain LIME1 SLC2A4RG
nsv586614 Chr.20:63725275 - 63741460 on Build GRCh38 Gain LIME1 SLC2A4RG ZGPAT
nsv1160690 Chr.20:63639552 - 63744287 on Build GRCh38 Deletion LIME1 RTEL1 ARFRP1 SLC2A4RG ZGPAT RTEL1-TNFRSF6B STMN3 ZBTB46 TNFRSF6B
nsv586618 Chr.20:63736025 - 63740688 on Build GRCh38 Gain LIME1 SLC2A4RG ZGPAT
nsv428381 Chr.20:63583375 - 64334167 on Build GRCh38 Gain+Loss UCKL1-AS1 TCEA2 MIR941-4 ZBTB46-AS1 RGS19 SLC2A4RG ABHD16B MYT1 UCKL1 C20orf181 RTEL1 MIR6813 GMEB2 LINC00266-1 ARFRP1 MIR1914 TPD52L2 DNAJC5 MIR941-1 SAMD10 PRPF6 ZGPAT LOC100505771 STMN3 MIR647 NPBWR2 ZBTB46 LINC00176 LIME1 ZNF512B PCMTD2 MIR941-5 SOX18 MIR941-3 MIR941-2 RTEL1-TNFRSF6B OPRL1 LKAAEAR1 TNFRSF6B
esv7227 Chr.20:63715550 - 63807161 on Build GRCh38 Loss LIME1 SLC2A4RG ZGPAT ZBTB46
nsv1059142 Chr.20:63731978 - 63798885 on Build GRCh38 Gain LIME1 SLC2A4RG ZGPAT ZBTB46
nsv470563 Chr.20:63486715 - 63793308 on Build GRCh38 Loss EEF1A2 SLC2A4RG ZGPAT LOC100505771 STMN3 SRMS ZBTB46 PPDPF LIME1 RTEL1 GMEB2 ARFRP1 RTEL1-TNFRSF6B HELZ2 PTK6 TNFRSF6B FNDC11
esv29394 Chr.20:63737285 - 63740490 on Build GRCh38 Loss LIME1 SLC2A4RG
dgv7682n54 Chr.20:63737667 - 63740090 on Build GRCh38 Loss LIME1 SLC2A4RG
esv2758806 Chr.20:63470996 - 63745014 on Build GRCh38 Gain+Loss KCNQ2 EEF1A2 SLC2A4RG ZGPAT LOC100505771 STMN3 SRMS ZBTB46 PPDPF LIME1 RTEL1 GMEB2 ARFRP1 RTEL1-TNFRSF6B HELZ2 PTK6 TNFRSF6B FNDC11
dgv7679n54 Chr.20:63737451 - 63740688 on Build GRCh38 Gain LIME1 SLC2A4RG

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s)

adaptive immune response
T cell receptor signaling pathway
B cell receptor signaling pathway

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline