This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00634855_cn
See other NSUN4 CNV Assays ›
Gene Symbol
NSUN4
Assay Reference Genome
Location

Chr.1:46344814 on build GRCh38
Cytoband
1p33
Assay ID Hs00634855_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

387338

Gene Name:

NOP2/Sun RNA methyltransferase family member 4

Gene Aliases:

SHTAP

Location:

Chr.1:46340177-46365152 on Build GRCh38

Assay Gene Location:

Overlaps Intron 2 - Exon 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NSUN4 NM_001256127.1 NP_001243056.1
NM_001256128.1 NP_001243057.1
NM_199044.3 NP_950245.2
NR_045789.1
NR_045790.1
NR_045791.1
AK021577.1 BAB13847.1
AK057420.1
AK097524.1
AK128066.1 BAC87258.1
AK291741.1
AK292353.1
AK295003.1
AK311009.1
BC014441.1 AAH14441.1
BM153143.1
BX360729.2
BX648592.1
DA799314.1
DA905308.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3577837 Chr.1:46338361 - 46357126 on Build GRCh38 Loss NSUN4
nsv829748 Chr.1:46258836 - 46410450 on Build GRCh38 Loss LRRC41 UQCRH RAD54L FAAH NSUN4
nsv470711 Chr.1:46194623 - 46344858 on Build GRCh38 Gain POMGNT1 LRRC41 LURAP1 UQCRH RAD54L TSPAN1 NSUN4
esv2761743 Chr.1:45874186 - 46561071 on Build GRCh38 Gain LRRC41 LURAP1 UQCRH LOC101929626 KNCN FAAH MKNK1 NSUN4 FAAHP1 PIK3R3 POMGNT1 RAD54L MAST2 DMBX1 MKNK1-AS1 TSPAN1 LINC01398
nsv546162 Chr.1:46341925 - 46405089 on Build GRCh38 Loss FAAH NSUN4

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

RNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

rRNA methylation
mature ribosome assembly
positive regulation of mitochondrial translation
protein binding
methyltransferase activity
small ribosomal subunit rRNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline