Assay Details
Target Gene Details
Entrez Gene ID: | 479 |
Gene Name: | ATPase H+/K+ transporting non-gastric alpha2 subunit |
Gene Aliases: |
#945, ATP1AL1, HK |
Location: |
Chr.13:24680411-24711785 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP12A | NM_001185085.1 | 8 | 1287 | NP_001172014.1 |
| NM_001676.5 | 8 | 1269 | NP_001667.4 | |
| AK292968.1 | 8 | 1269 | ||
| AK303498.1 | ||||
| BC031609.1 | 8 | 1121 | AAH31609.1 | |
| U02076.1 | 8 | 1103 | AAB37755.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv561345 | Chr.13:24606013 - 24763162 on Build GRCh38 | Gain |
|
| nsv832568 | Chr.13:24522385 - 24743328 on Build GRCh38 | Gain |
|
| nsv561348 | Chr.13:24643343 - 24729117 on Build GRCh38 | Gain |
|
| esv2751140 | Chr.13:24611237 - 24717762 on Build GRCh38 | Gain |
|
| nsv528064 | Chr.13:24689899 - 24695269 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map