This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00655767_cn
See other JMJD4 CNV Assays ›
Gene Symbol
JMJD4
Assay Reference Genome
Location

Chr.1:227734818 on build GRCh38
Cytoband
1q42.13
Assay ID Hs00655767_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

65094

Gene Name:

jumonji domain containing 4

Gene Aliases:

-

Location:

Chr.1:227731189-227735423 on Build GRCh38

Assay Gene Location:

Overlaps Intron 1 - Exon 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
JMJD4 NM_001161465.1 NP_001154937.1
NM_023007.2 NP_075383.2
XM_011544262.2 XP_011542564.1
AK022579.1 BAB14109.1
AK023030.1 BAB14366.1

Target Gene Details

Entrez Gene ID:

116841

Gene Name:

synaptosome associated protein 47

Gene Aliases:

C1orf142, ESFI5812, HEL-S-290, HEL170, SNAP-47, SVAP1

Location:

Chr.1:227728168-227781231 on Build GRCh38

Assay Gene Location:

Within Exon 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SNAP47 NM_001323930.1 NP_001310859.1
NM_001323932.1 NP_001310861.1
NM_001323933.1 NP_001310862.1
NR_136654.1
XM_017000231.1 3 2862 XP_016855720.1
XM_017000232.1 2 2668 XP_016855721.1
BC032775.1 AAH32775.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1007479 Chr.1:227508167 - 227796765 on Build GRCh38 Gain LOC105373289 ZNF678 JMJD4 SNAP47
nsv826908 Chr.1:227707685 - 228503399 on Build GRCh38 Gain GJC2 PRSS38 MIR3620 HIST3H2A OBSCN ARF1 SNAP47 C1orf35 IBA57-AS1 GUK1 MIR6742 LOC100506571 MIR5008 HIST3H2BB LOC101060022 IBA57 RNF187 TRIM17 WNT3A HIST3H3 MRPL55 LOC107985355 MIR4666A LOC105373289 LOC101927401 C1orf145 TRIM11 JMJD4 WNT9A
esv32853 Chr.1:227692415 - 228670462 on Build GRCh38 Gain RNA5S13 MIR3620 RNA5S11 ARF1 RNA5S6 RNA5S15 C1orf35 IBA57-AS1 MIR6742 HIST3H2BB LOC101060022 RNF187 RNA5S4 TRIM17 WNT3A RNA5S16 RNA5S10 RNA5S17 RNA5S14 RNA5S2 TRIM11 WNT9A RNA5S9 GJC2 PRSS38 HIST3H2A OBSCN RNA5S7 SNAP47 RNA5S5 GUK1 LOC100506571 MIR5008 IBA57 HIST3H3 MRPL55 LOC107985355 RNA5S3 MIR4666A LOC105373289 LOC101927401 DUSP5P1 C1orf145 BTNL10 RNA5S12 RHOU LOC105373132 RNA5S1 RNA5S8 JMJD4
nsv515496 Chr.1:227724773 - 227780237 on Build GRCh38 Loss JMJD4 SNAP47

Back To Top

More Information


Set Membership:

Intragenic Exonic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

protein modifying enzyme SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

synaptic vesicle priming
synaptic vesicle fusion to presynaptic active zone membrane
long-term synaptic potentiation
protein binding
SNAP receptor activity
syntaxin binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline