This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00668905_cn
See other NUDT22 CNV Assays ›
Gene Symbol
NUDT22
Assay Reference Genome
Location

Chr.11:64227658 on build GRCh38
Cytoband
11q13.1
Assay ID Hs00668905_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

84304

Gene Name:

nudix hydrolase 22

Gene Aliases:

-

Location:

Chr.11:64226258-64230016 on Build GRCh38

Assay Gene Location:

Overlaps Exon 3 - Intron 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NUDT22 NM_001128612.2 NP_001122084.1
NM_001128613.2 NP_001122085.1
NM_001271831.1 NP_001258760.1
NM_032344.3 NP_115720.1
BC006129.1 AAH06129.1
BG705568.1
BM923060.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1159904 Chr.11:64150129 - 64279169 on Build GRCh38 Deletion GPR137 PLCB3 NUDT22 BAD STIP1 MACROD1 TRPT1 FERMT3 FKBP2 VEGFB PPP1R14B DNAJC4 LOC105369340
esv992196 Chr.11:64225062 - 64234883 on Build GRCh38 Deletion NUDT22 TRPT1 VEGFB DNAJC4
nsv951010 Chr.11:63942829 - 64352128 on Build GRCh38 Deletion GPR137 STIP1 KCNK4 TRPT1 MIR7155 ESRRA VEGFB LOC105369340 PLCB3 COX8A NUDT22 FLRT1 OTUB1 PRDX5 BAD MACROD1 KCNK4-TEX40 FERMT3 FKBP2 CCDC88B TEX40 PPP1R14B NAA40 DNAJC4 TRMT112
dgv1963n54 Chr.11:64174262 - 64329761 on Build GRCh38 Loss GPR137 PLCB3 NUDT22 PRDX5 BAD STIP1 KCNK4 TRPT1 KCNK4-TEX40 FERMT3 FKBP2 ESRRA VEGFB TEX40 PPP1R14B DNAJC4 TRMT112 LOC105369340
nsv555187 Chr.11:64185649 - 64260416 on Build GRCh38 Loss STIP1 PLCB3 NUDT22 TRPT1 FERMT3 FKBP2 VEGFB PPP1R14B DNAJC4 LOC105369340
nsv555184 Chr.11:64156974 - 64275525 on Build GRCh38 Loss GPR137 PLCB3 NUDT22 BAD STIP1 MACROD1 TRPT1 FERMT3 FKBP2 VEGFB PPP1R14B DNAJC4 LOC105369340
nsv832186 Chr.11:64204788 - 64402587 on Build GRCh38 Loss GPR137 PLCB3 NUDT22 MIR1237 PRDX5 BAD KCNK4 TRPT1 KCNK4-TEX40 FERMT3 FKBP2 CCDC88B MIR7155 RPS6KA4 ESRRA VEGFB TEX40 PPP1R14B DNAJC4 TRMT112 LOC105369340
nsv469961 Chr.11:64055468 - 64318495 on Build GRCh38 Loss GPR137 PLCB3 NUDT22 FLRT1 PRDX5 BAD STIP1 MACROD1 KCNK4 TRPT1 KCNK4-TEX40 FERMT3 FKBP2 ESRRA VEGFB TEX40 PPP1R14B DNAJC4 TRMT112 LOC105369340

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Process(es)

protein binding
hydrolase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-tfltf:80/100.66.78.247:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0