Assay Details
Target Gene Details
Entrez Gene ID: | 10650 |
Gene Name: | PRELI domain containing 3A |
Gene Aliases: |
C18orf43, HFL-EDDG1, SLMO1 |
Location: |
Chr.18:12407896-12432238 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRELID3A | NM_001142405.1 | NP_001135877.1 | ||
| NM_001142406.1 | NP_001135878.1 | |||
| NM_006553.3 | NP_006544.2 | |||
| AK056046.1 | BAB71083.1 | |||
| AK303325.1 | ||||
| AL359942.1 | CAB95773.1 | |||
| BC106750.2 | AAI06751.1 | |||
| CR627465.1 | CAH10669.1 | |||
| DB018070.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519557 | Chr.18:12417906 - 12431507 on Build GRCh38 | Loss |
|
| nsv1060808 | Chr.18:12302793 - 12514418 on Build GRCh38 | Gain |
|
| nsv576496 | Chr.18:12247873 - 12490503 on Build GRCh38 | Gain |
|
| nsv1063125 | Chr.18:12046893 - 12918252 on Build GRCh38 | Loss |
|
| esv2758714 | Chr.18:12240682 - 12499526 on Build GRCh38 | Loss |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map