Assay Details
Target Gene Details
Entrez Gene ID: | 343066 |
Gene Name: | arylacetamide deacetylase like 4 |
Gene Aliases: |
- |
Location: |
Chr.1:12644547-12667086 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AADACL4 | NM_001013630.1 | NP_001013652.1 | ||
| XM_017001152.1 | XP_016856641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515525 | Chr.1:12659200 - 12663173 on Build GRCh38 | Loss |
|
| nsv1011540 | Chr.1:12601623 - 12832443 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map