This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00718715_cn
See other SERPINB8 CNV Assays ›
Gene Symbol
SERPINB8
Assay Reference Genome
Location

Chr.18:63970054 on build GRCh38
Cytoband
18q22.1
Assay ID Hs00718715_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

5271

Gene Name:

serpin family B member 8

Gene Aliases:

CAP2, PI8

Location:

Chr.18:63970028-64019779 on Build GRCh38

Assay Gene Location:

Overlaps - Exon 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SERPINB8 NM_001031848.1 NP_001027018.1
NM_001276490.1 NP_001263419.1
NM_002640.3 NP_002631.3
NM_198833.1 NP_942130.1
XM_006722484.2 XP_006722547.1
XM_011526025.1 XP_011524327.1
XM_017025790.1 XP_016881279.1
XM_017025792.1 XP_016881281.1
AK300391.1
AL708543.1
BQ435307.1
BX280409.1
DQ656055.2
L40377.1 AAC41939.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1066552 Chr.18:63147299 - 64608176 on Build GRCh38 Gain SERPINB10 KDSR SERPINB8 VPS4B LINC00305 SERPINB2 SERPINB3 SERPINB4 HMSD LOC284294 BCL2 SERPINB11 SERPINB5 SERPINB7 SERPINB12 LINC01538 SERPINB13
nsv833673 Chr.18:63897072 - 64097021 on Build GRCh38 Gain SERPINB10 SERPINB8 LINC00305 SERPINB2 HMSD
esv21739 Chr.18:63715122 - 64278129 on Build GRCh38 Gain SERPINB11 SERPINB10 SERPINB7 SERPINB8 LINC01538 LINC00305 SERPINB2 HMSD LOC284294
esv3642785 Chr.18:63831255 - 63986077 on Build GRCh38 Gain SERPINB10 SERPINB8 SERPINB2 HMSD
nsv1067311 Chr.18:63818011 - 66637685 on Build GRCh38 Loss SERPINB10 CDH19 SERPINB8 LINC01538 LINC00305 SERPINB2 CDH7 HMSD LOC284294

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of endopeptidase activity
serine-type endopeptidase inhibitor activity
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline