This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00732588_cn
See other MUTYH CNV Assays ›
Gene Symbol
MUTYH
Assay Reference Genome
Location

Chr.1:45329330 on build GRCh38
Cytoband
1p34.1
Assay ID Hs00732588_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,505.00
Your Price
Online offer:
4505.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

4595

Gene Name:

mutY DNA glycosylase

Gene Aliases:

MYH

Location:

Chr.1:45329242-45340925 on Build GRCh38

Assay Gene Location:

Within Exon 22
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MUTYH NM_001048171.1 16 1800 NP_001041636.1
NM_001048172.1 16 1691 NP_001041637.1
NM_001048173.1 16 1688 NP_001041638.1
NM_001048174.1 16 1607 NP_001041639.1
NM_001128425.1 16 1842 NP_001121897.1
NM_001293190.1 16 1803 NP_001280119.1
NM_001293191.1 16 1640 NP_001280120.1
NM_001293192.1 16 1736 NP_001280121.1
NM_001293195.1 17 1757 NP_001280124.1
NM_001293196.1 16 1625 NP_001280125.1
NM_012222.2 16 1833 NP_036354.1
XM_011541497.2 16 1899 XP_011539799.1
XM_011541498.1 16 1677 XP_011539800.1
XM_011541499.1 16 1663 XP_011539801.1
XM_011541500.2 16 1858 XP_011539802.1
XM_011541501.2 17 1772 XP_011539803.1
XM_011541502.2 16 1677 XP_011539804.1
XM_011541503.2 15 1645 XP_011539805.1
XM_011541504.2 16 1726 XP_011539806.1
XM_011541505.2 12 1346 XP_011539807.1
XM_011541506.1 12 1330 XP_011539808.1
XM_011541507.2 11 1142 XP_011539809.2
XM_017001331.1 16 1676 XP_016856820.1
XM_017001332.1 16 1678 XP_016856821.1
XM_017001333.1 16 1731 XP_016856822.1
XM_017001334.1 16 1624 XP_016856823.1
XM_017001335.1 16 1570 XP_016856824.1
XM_017001336.1 15 1654 XP_016856825.1
XM_017001337.1 15 1594 XP_016856826.1
XM_017001338.1 15 2181 XP_016856827.1
XM_017001339.1 15 1632 XP_016856828.1
AB032920.1 16 1751 BAA89336.1
AB032921.1 16 1721 BAA89337.1
AB032922.1 16 1718 BAA89338.1
AB032923.1 16 1654 BAA89339.1
AB032924.1 16 1618 BAA89340.1
AB032925.1 16 1585 BAA89341.1
AB032926.1 17 1735 BAA89342.1
AB032927.1 16 1656 BAA89343.1
AB032928.1 16 1653 BAA89344.1
AB032929.1 16 1589 BAA89345.1
AK293830.1 7 862
BC003178.1 16 1738 AAH03178.1
KU178203.1
KU178204.1

Back To Top

More Information


Set Membership:

Intragenic Exonic non-DGV Variation

Panther Classification:

Molecular Function -

DNA glycosylase

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
mismatch repair
response to oxidative stress
depurination
DNA binding
protein binding
DNA N-glycosylase activity
MutLalpha complex binding
MutLbeta complex binding
MutSalpha complex binding
MutSbeta complex binding
metal ion binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0