Assay Details
Target Gene Details
Entrez Gene ID: | 51111 |
Gene Name: | lysine methyltransferase 5B |
Gene Aliases: |
CGI-85, CGI85, SUV420H1 |
Location: |
Chr.11:68154863-68213904 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KMT5B | NM_001300907.1 | NP_001287836.1 | ||
| NM_001300908.1 | NP_001287837.1 | |||
| NM_001300909.1 | 9 | 1435 | NP_001287838.1 | |
| NM_016028.4 | 10 | 1388 | NP_057112.3 | |
| NM_017635.4 | NP_060105.3 | |||
| XM_005274035.3 | XP_005274092.2 | |||
| XM_005274036.3 | XP_005274093.2 | |||
| XM_006718581.1 | XP_006718644.1 | |||
| XM_011545091.1 | XP_011543393.1 | |||
| XM_011545092.2 | XP_011543394.1 | |||
| XM_011545093.2 | XP_011543395.1 | |||
| XM_011545094.2 | 10 | 1636 | XP_011543396.1 | |
| XM_017017876.1 | XP_016873365.1 | |||
| XM_017017877.1 | XP_016873366.1 | |||
| XM_017017878.1 | XP_016873367.1 | |||
| XM_017017879.1 | XP_016873368.1 | |||
| XM_017017880.1 | XP_016873369.1 | |||
| AF151843.1 | 9 | 1130 | AAD34080.1 | |
| AK025183.1 | 9 | 1076 | ||
| AK025461.1 | ||||
| AK026428.1 | ||||
| AK299201.1 | 9 | 1264 | ||
| AK314336.1 | ||||
| BC002522.2 | 10 | 1335 | AAH02522.2 | |
| BC012933.1 | ||||
| BC065287.1 | 10 | 1340 | AAH65287.1 | |
| BC087834.1 | 10 | 1357 | AAH87834.1 | |
| BC098121.1 | AAH98121.1 | |||
| BC099714.1 | AAH99714.1 | |||
| BC103498.1 | AAI03499.1 | |||
| BC104483.1 | AAI04484.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825963 | Chr.11:67996205 - 68429586 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map