This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00822886_cn
See other CD300C CNV Assays ›
Gene Symbol
CD300C
Assay Reference Genome
Location

Chr.17:74545738 on build GRCh38
Cytoband
17q25.1
Assay ID Hs00822886_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

10871

Gene Name:

CD300c molecule

Gene Aliases:

CLM-6, CMRF-35, CMRF-35A, CMRF35, CMRF35-A1, CMRF35A, CMRF35A1, IGSF16, LIR

Location:

Chr.17:74534359-74546171 on Build GRCh38

Assay Gene Location:

Overlaps Intron 1 - Exon 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CD300C NM_006678.4 NP_006669.1
XM_017024033.1 XP_016879522.1
XM_017024034.1 XP_016879523.1
BC022279.1 AAH22279.1
BP298972.1
X66171.1 CAA46948.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv12e196 Chr.17:73675043 - 74565539 on Build GRCh38 Duplication LOC100130520 BTBD17 LOC100134391 GPR142 MGC16275 LINC00469 CD300A CD300LB RPL38 LOC400620 GPRC5C TTYH2 CD300C DNAI2 KIF19
nsv470600 Chr.17:74466882 - 74550979 on Build GRCh38 Loss CD300A CD300LB CD300C
esv2761978 Chr.17:73835799 - 74684915 on Build GRCh38 Gain RAB37 CD300E LOC100130520 BTBD17 GPR142 MGC16275 LOC101928343 CD300A CD300LB RPL38 GPRC5C TTYH2 C17orf77 CD300LD CD300C DNAI2 KIF19
esv3641230 Chr.17:73831787 - 74694861 on Build GRCh38 Gain RAB37 CD300E LOC100130520 BTBD17 GPR142 MGC16275 LOC101928343 CD300A CD300LB RPL38 GPRC5C TTYH2 C17orf77 CD300LF CD300LD CD300C DNAI2 KIF19
dgv3282n100 Chr.17:73814343 - 74719597 on Build GRCh38 Gain RAB37 CD300E LOC100130520 BTBD17 GPR142 MGC16275 LINC00469 LOC101928343 CD300A CD300LB RPL38 GPRC5C TTYH2 C17orf77 CD300LF CD300LD CD300C DNAI2 KIF19
esv33995 Chr.17:74261053 - 74732093 on Build GRCh38 Loss RAB37 CD300E LOC100130520 BTBD17 GPR142 LOC101928343 CD300A CD300LB GPRC5C TTYH2 C17orf77 CD300LF CD300LD CD300C DNAI2 KIF19
dgv372n27 Chr.17:73826016 - 74685802 on Build GRCh38 Gain RAB37 CD300E LOC100130520 BTBD17 GPR142 MGC16275 LINC00469 LOC101928343 CD300A CD300LB RPL38 GPRC5C TTYH2 C17orf77 CD300LD CD300C DNAI2 KIF19
esv2751770 Chr.17:73837862 - 74716223 on Build GRCh38 Gain RAB37 CD300E LOC100130520 BTBD17 GPR142 MGC16275 LOC101928343 CD300A CD300LB RPL38 GPRC5C TTYH2 C17orf77 CD300LF CD300LD CD300C DNAI2 KIF19
esv3573820 Chr.17:73817932 - 74694378 on Build GRCh38 Gain RAB37 CD300E LOC100130520 BTBD17 GPR142 MGC16275 LINC00469 LOC101928343 CD300A CD300LB RPL38 GPRC5C TTYH2 C17orf77 CD300LF CD300LD CD300C DNAI2 KIF19
dgv5774n54 Chr.17:74428381 - 74673021 on Build GRCh38 Gain LOC101928343 RAB37 CD300E CD300A CD300LB LOC100130520 GPRC5C C17orf77 CD300LD CD300C
esv3641244 Chr.17:74527395 - 74686883 on Build GRCh38 Gain LOC101928343 RAB37 CD300E CD300LB LOC100130520 C17orf77 CD300LD CD300C

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

immune system process
cellular defense response
signal transduction
regulation of immune response
transmembrane signaling receptor activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0