This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00841001_cn
See other MIB2 CNV Assays ›
Gene Symbol
MIB2
Assay Reference Genome
Location

Chr.1:1627123 on build GRCh38
Cytoband
1p36.33
Assay ID Hs00841001_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

142678

Gene Name:

mindbomb E3 ubiquitin protein ligase 2

Gene Aliases:

ZZANK1, ZZZ5

Location:

Chr.1:1615188-1630610 on Build GRCh38

Assay Gene Location:

Within Exon 12
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MIB2 NM_001170686.1 11 1668 NP_001164157.1
NM_001170687.1 11 1638 NP_001164158.1
NM_001170688.1 10 1485 NP_001164159.1
NM_001170689.1 10 1460 NP_001164160.1
NM_080875.2 11 1680 NP_543151.2
NR_033183.1 11 1463
XM_006710372.1 10 1544 XP_006710435.1
XM_011540731.2 12 2417 XP_011539033.1
XM_011540736.2 12 2417 XP_011539038.1
XM_011540737.2 11 2354 XP_011539039.1
XM_011540741.2 10 2249 XP_011539043.1
XM_011540742.2 10 2246 XP_011539044.1
XM_017000349.1 12 2417 XP_016855838.1
XM_017000350.1 12 2405 XP_016855839.1
XM_017000351.1 12 2399 XP_016855840.1
XM_017000352.1 12 2375 XP_016855841.1
XM_017000353.1 11 2354 XP_016855842.1
XM_017000354.1 12 2417 XP_016855843.1
XM_017000355.1 10 2249 XP_016855844.1
XM_017000356.1 10 2246 XP_016855845.1
XM_017000357.1 11 2222 XP_016855846.1
XM_017000358.1 12 1715 XP_016855847.1
XM_017000359.1 12 1703 XP_016855848.1
XM_017000360.1 12 1662 XP_016855849.1
XM_017000361.1 11 1652 XP_016855850.1
XM_017000362.1 12 1755 XP_016855851.1
XM_017000363.1 12 1718 XP_016855852.1
XM_017000364.1 10 1457 XP_016855853.1
XM_017000365.1 12 1445 XP_016855854.1
XM_017000366.1 11 1382 XP_016855855.1
AB074480.1 11 1591 BAB92950.1
AB076691.1 11 1549 BAC00992.1
AB076692.1 10 1396 BAC00993.1
AB076693.1 11 1579 BAC00994.1
AB097000.1 11 1514 BAC77353.1
AK091610.1 5 484 BAC03707.1
AK095914.1 5 486 BAC04646.1
AK096295.1 6 591 BAC04752.1
AK097106.1 7 910 BAC04952.1
AK098785.1 11 1549 BAC05413.1
AK122751.1 11 1382
AK122863.1 11 1581
AK128167.1 10 1420
AK128553.1 10 2594
AK293642.1 10 1660
AK302757.1 11 1632
AK309362.1 9 1550
AL834527.1 6 712 CAD39183.1
BC016490.1 11 1517 AAH16490.1
BC037542.1 11 1560 AAH37542.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion SLC35E2B MIB2 FNDC10 MMP23B NADK CDK11A SLC35E2 CDK11B MMP23A GNB1
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss SLC35E2B MIB2 FNDC10 MMP23B SSU72 CDK11A SLC35E2 CDK11B MMP23A
esv3575582 Chr.1:1624086 - 1721336 on Build GRCh38 Gain SLC35E2B MIB2 MMP23B CDK11A CDK11B MMP23A
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss LOC100288069 ATAD3C LOC100130417 LOC107985729 MIR200B LOC100287934 HES4 MMP23B SDF4 SLC35E2 SCNN1D CDK11A LINC01342 TTLL10 MMP23A TTLL10-AS1 CPTP TMEM88B SLC35E2B LOC102724312 TNFRSF4 NOC2L FNDC10 NADK LINC01128 RNF223 LOC284600 MRPL20 VWA1 KLHL17 ATAD3A SSU72 MIR429 CDK11B FAM132A SAMD11 LOC148413 FAM41C MIR6726 B3GALT6 CCNL2 MIB2 AURKAIP1 CPSF3L FAM87B PLEKHN1 LOC100288175 LOC105378948 LINC00115 TAS1R3 UBE2J2 MIR200A TMEM240 TNFRSF18 AGRN DVL1 PERM1 MIR6808 ISG15 PUSL1 ATAD3B MIR6727 ACAP3 LOC107985728 MXRA8 C1orf159 ANKRD65
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC100288069 ATAD3C LOC100130417 LOC107985729 MIR200B LOC100287934 CALML6 LOC100133331 HES4 FAAP20 SLC35E2 GNB1 LOC100129534 LOC729737 LINC01342 TTLL10 TTLL10-AS1 CPTP TMEM88B LOC102724312 TNFRSF4 NOC2L FNDC10 LOC102725121 DDX11L1 MRPL20 LOC105378949 LOC101928626 ATAD3A MIR429 CDK11B FAM132A LOC148413 FAM41C MIB2 MIR6723 MORN1 TAS1R3 LOC105378592 GABRD PERM1 LOC100134822 ACAP3 LOC107985728 MXRA8 LOC100506504 ANKRD65 LOC107984841 MTND1P23 MIR6859-1 MTND2P28 LOC105378589 MMP23B SDF4 SCNN1D CDK11A OR4F5 MMP23A SLC35E2B LOC105378947 NADK LOC105378591 PRKCZ LINC01128 RNF223 LOC284600 OR4F16 SKI VWA1 KLHL17 SSU72 MIR6859-2 SAMD11 MIR6726 FAM138A TMEM52 B3GALT6 OR4F29 CCNL2 LOC100132287 AURKAIP1 CPSF3L FAM87B PLEKHN1 LOC100288175 LOC105378948 LINC00115 UBE2J2 MIR200A WASH7P TMEM240 TNFRSF18 AGRN CFAP74 DVL1 MIR6808 ISG15 PUSL1 ATAD3B MIR6727 MIR1302-2 C1orf159
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication SLC35E2B MIB2 FNDC10 MMP23B SSU72 NADK CDK11A SLC35E2 CDK11B MMP23A GNB1
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion MRPL20 VWA1 ATAD3C ATAD3A SSU72 MIR429 CDK11B LOC107985729 FAM132A MIR200B LOC148413 MIR6726 B3GALT6 CCNL2 MIB2 MMP23B SDF4 AURKAIP1 CPSF3L SCNN1D TAS1R3 UBE2J2 MIR200A TMEM240 TNFRSF18 LINC01342 TTLL10 DVL1 TTLL10-AS1 MIR6808 CPTP TMEM88B LOC102724312 TNFRSF4 FNDC10 PUSL1 ATAD3B MIR6727 ACAP3 MXRA8 C1orf159 ANKRD65
esv2661270 Chr.1:1625365 - 1636030 on Build GRCh38 Deletion MIB2 MMP23B CDK11B
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss SLC35E2B MIB2 FNDC10 MMP23B SSU72 NADK CDK11A SLC35E2 CDK11B MMP23A
nsv545017 Chr.1:1615322 - 1663759 on Build GRCh38 Loss SLC35E2B MIB2 MMP23B CDK11B
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss MRPL20 VWA1 LOC105378949 ATAD3C ATAD3A SSU72 CDK11B LOC107985729 LOC148413 LOC105378589 TMEM52 CALML6 CCNL2 MIB2 MMP23B SLC35E2 GNB1 GABRD TMEM240 CFAP74 CDK11A MMP23A TMEM88B SLC35E2B LOC102724312 FNDC10 NADK ATAD3B LOC105378591 PRKCZ ANKRD65

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
Notch signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
ubiquitin-protein transferase activity
signal transducer activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0