This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00862059_cn
See other VSTM1 CNV Assays ›
Gene Symbol
VSTM1
Assay Reference Genome
Location

Chr.19:54041812 on build GRCh38
Cytoband
19q13.42
Assay ID Hs00862059_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

284415

Gene Name:

V-set and transmembrane domain containing 1

Gene Aliases:

SIRL-1, SIRL1, UNQ3033

Location:

Chr.19:54040825-54063966 on Build GRCh38

Assay Gene Location:

Overlaps Intron 11 - Intron 12
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
VSTM1 NM_001288791.1 NP_001275720.1
NM_001288792.1 NP_001275721.1
NM_001288793.1 NP_001275722.1
NM_198481.3 NP_940883.2
NR_110142.1
XM_011526845.1 XP_011525147.1
XM_011526846.1 XP_011525148.1
XM_011526847.1 XP_011525149.1
XM_011526848.1 XP_011525150.1
XM_011526849.1 XP_011525151.1
AY358542.1 AAQ88906.1
BC100943.2 AAI00944.1
BG189223.1
BQ446114.1
DQ479397.1
FJ584316.1
FJ584317.1
FJ584318.1
FJ584319.1
FJ584320.1
FJ882051.1
FN398145.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv458782 Chr.19:53961144 - 54093354 on Build GRCh38 Gain TARM1 VSTM1 CACNG8 CACNG6 MIR935
nsv510773 Chr.19:54017180 - 54132236 on Build GRCh38 Deletion TFPT TARM1 VSTM1 NDUFA3 PRPF31 OSCAR
nsv1063127 Chr.19:53926876 - 54068977 on Build GRCh38 Gain VSTM1 CACNG8 CACNG7 CACNG6 MIR935
nsv1057516 Chr.19:53986233 - 54046901 on Build GRCh38 Gain VSTM1 CACNG8 CACNG6
esv2718812 Chr.19:53184636 - 54322450 on Build GRCh38 Deletion RPS9 MIR525 MIR520G MIR518A1 MIR373 MIR519E NLRP12 MIR1283-2 MIR371B VN1R4 CACNG6 MIR526A1 ZNF813 MIR516B1 MIR516A1 MBOAT7 DPRX VSTM1 ZNF765 LILRA6 FAM90A27P MIR518A2 MIR517C PRKCG MIR516B2 MIR520F MIR524 CACNG7 LENG1 MIR518F MIR519A2 MIR521-1 MIR520C ZNF677 ZNF525 MIR518C MIR518B MIR522 MIR371A MIR520E MYADM CNOT3 MIR516A2 MIR515-1 MIR519A1 BIRC8 ZNF665 LOC284379 ZNF818P MIR523 CACNG8 MIR526A2 PRPF31 MIR520H MIR935 TPM3P9 ZNF845 MIR519B MIR512-2 MIR519D MIR512-1 ZNF761 LILRA5 MIR517B LILRB5 MIR519C MIR1323 MIR517A MIR527 TFPT TARM1 VN1R2 MIR520A MIR518D MIR372 MIR515-2 MIR520D LILRB3 OSCAR MIR520B MIR518E MIR1283-1 LOC105372457 TSEN34 MIR498 NDUFA3 MIR526B ZNF331 TMC4 MIR4752 LILRB2 MIR521-2
nsv833877 Chr.19:53951529 - 54117323 on Build GRCh38 Loss TFPT TARM1 VSTM1 NDUFA3 CACNG8 PRPF31 OSCAR CACNG6 MIR935

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

immune system process
cytokine activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline