This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00873764_cn
See other CPSF3L CNV Assays ›
Gene Symbol
CPSF3L
Assay Reference Genome
Location

Chr.1:1321048 on build GRCh38
Cytoband
1p36.33
Assay ID Hs00873764_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,505.00
Your Price
Online offer:
4505.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

54973

Gene Name:

cleavage and polyadenylation specific factor 3-like

Gene Aliases:

CPSF73L, INT11, INTS11, RC-68, RC68

Location:

Chr.1:1311585-1324687 on Build GRCh38

Assay Gene Location:

Overlaps Intron 3 - Exon 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CPSF3L NM_001256456.1 NP_001243385.1
NM_001256460.1 NP_001243389.1
NM_001256462.1 NP_001243391.1
NM_001256463.1 NP_001243392.1
NM_017871.5 NP_060341.2
XM_011541647.1 XP_011539949.1
XM_011541648.1 XP_011539950.1
XM_011541650.1 XP_011539952.1
XM_017001557.1 XP_016857046.1
XM_017001558.1 XP_016857047.1
AK021939.1 BAB13943.1
AK022675.1
AK023356.1 BAB14541.1
AK027767.1
AK056652.1
AK058018.1
AK092190.1
AK291387.1
AK297350.1
AL136813.1 CAB66747.1
BC007978.2 AAH07978.1
BC008041.2 AAH08041.1
BC020199.1 AAH20199.1
CR533557.1 CAG38588.1
DC295791.1
DC299237.1
DC299993.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC105378592 LOC100133331 CPSF3L LOC100132287 SLC35E2B KLHL17 ATAD3C MIR6726 LOC100287934 DVL1 MIR200A CDK11A UBE2J2 ATAD3A WASH7P FAM41C CCNL2 FAM87B SKI LOC100506504 LOC105378949 SDF4 ATAD3B C1orf159 ISG15 MIR6727 LOC729737 PLEKHN1 MTND2P28 LINC01342 MXRA8 FAAP20 ANKRD65 LOC107985729 LOC284600 PERM1 CPTP CFAP74 SCNN1D LINC00115 MIR6859-1 LOC105378589 LOC105378591 SAMD11 MIR200B B3GALT6 MIB2 OR4F16 GABRD MTND1P23 VWA1 LOC107985728 TTLL10-AS1 MIR6808 LOC100134822 AGRN OR4F5 NADK LOC105378948 FAM132A LOC105378947 LOC102725121 MRPL20 PRKCZ MMP23A SLC35E2 DDX11L1 LOC148413 TNFRSF4 MIR1302-2 TTLL10 HES4 SSU72 NOC2L PUSL1 RNF223 LOC102724312 CDK11B AURKAIP1 LINC01128 TNFRSF18 LOC100129534 LOC100130417 TAS1R3 GNB1 LOC100288069 MMP23B LOC101928626 LOC100288175 LOC107984841 FAM138A MORN1 ACAP3 MIR6723 CALML6 MIR6859-2 FNDC10 TMEM240 MIR429 TMEM88B OR4F29 TMEM52
nsv544971 Chr.1:1294063 - 1351426 on Build GRCh38 Loss ACAP3 DVL1 TAS1R3 PUSL1 CPSF3L MIR6726 MIR6808 MIR6727 CPTP
nsv544969 Chr.1:1290683 - 1374597 on Build GRCh38 Loss ACAP3 DVL1 TAS1R3 PUSL1 MXRA8 CPSF3L MIR6726 SCNN1D AURKAIP1 MIR6808 MIR6727 CPTP
nsv997291 Chr.1:1306835 - 1376336 on Build GRCh38 Gain ACAP3 DVL1 TAS1R3 PUSL1 MXRA8 CPSF3L AURKAIP1 MIR6808 MIR6727 CPTP
nsv470680 Chr.1:1070426 - 1378635 on Build GRCh38 Loss C1orf159 CPSF3L TNFRSF4 MIR6727 MIR200B B3GALT6 TTLL10 PUSL1 LINC01342 RNF223 MXRA8 TTLL10-AS1 MIR6726 AURKAIP1 MIR6808 TNFRSF18 DVL1 TAS1R3 MIR200A FAM132A UBE2J2 CPTP ACAP3 MIR429 SCNN1D SDF4
dgv6n100 Chr.1:1301750 - 1362354 on Build GRCh38 Gain+Loss ACAP3 DVL1 TAS1R3 PUSL1 MXRA8 CPSF3L MIR6808 MIR6727 CPTP
esv3584992 Chr.1:1314419 - 1330342 on Build GRCh38 Gain CPSF3L CPTP
nsv1160788 Chr.1:1282706 - 1376336 on Build GRCh38 Deletion ACAP3 DVL1 TAS1R3 PUSL1 MXRA8 CPSF3L MIR6726 SCNN1D AURKAIP1 MIR6808 MIR6727 CPTP
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication C1orf159 CPSF3L TNFRSF4 MIR6727 MIR200B B3GALT6 TTLL10 PUSL1 LINC01342 RNF223 TTLL10-AS1 MIR6726 AGRN TNFRSF18 LOC105378948 TAS1R3 MIR200A FAM132A LOC100288175 UBE2J2 CPTP ACAP3 MIR429 SCNN1D SDF4
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss CPSF3L SAMD11 MIR200B SLC35E2B B3GALT6 MIB2 KLHL17 ATAD3C VWA1 LOC107985728 TTLL10-AS1 MIR6726 MIR6808 LOC100287934 AGRN NADK DVL1 LOC105378948 MIR200A FAM132A CDK11A UBE2J2 ATAD3A MRPL20 MMP23A FAM41C CCNL2 FAM87B SLC35E2 SDF4 ATAD3B LOC148413 C1orf159 ISG15 TNFRSF4 MIR6727 TTLL10 PLEKHN1 HES4 SSU72 NOC2L PUSL1 LINC01342 RNF223 MXRA8 LOC102724312 CDK11B ANKRD65 AURKAIP1 LOC107985729 LINC01128 TNFRSF18 LOC100130417 TAS1R3 LOC284600 LOC100288069 MMP23B LOC100288175 PERM1 CPTP ACAP3 FNDC10 TMEM240 MIR429 TMEM88B SCNN1D LINC00115
esv3890636 Chr.1:1304555 - 1353203 on Build GRCh38 Loss ACAP3 DVL1 TAS1R3 PUSL1 MXRA8 CPSF3L MIR6808 MIR6727 CPTP
nsv1000169 Chr.1:1281678 - 1362354 on Build GRCh38 Gain ACAP3 DVL1 TAS1R3 PUSL1 MXRA8 CPSF3L MIR6726 SCNN1D MIR6808 MIR6727 CPTP
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion ATAD3B LOC148413 C1orf159 CPSF3L TNFRSF4 MIR6727 MIR200B B3GALT6 TTLL10 MIB2 SSU72 PUSL1 LINC01342 ATAD3C VWA1 MXRA8 LOC102724312 TTLL10-AS1 MIR6726 CDK11B ANKRD65 AURKAIP1 LOC107985729 MIR6808 TNFRSF18 DVL1 TAS1R3 MIR200A FAM132A MMP23B UBE2J2 ATAD3A CPTP ACAP3 MRPL20 FNDC10 TMEM240 MIR429 TMEM88B SCNN1D CCNL2 SDF4
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss DVL1 TAS1R3 CPSF3L MIR6727 UBE2J2 CPTP ACAP3 PUSL1 MXRA8 MIR6726 SCNN1D AURKAIP1 MIR6808
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain LOC148413 C1orf159 ISG15 CPSF3L TNFRSF4 SAMD11 MIR6727 MIR200B B3GALT6 TTLL10 PLEKHN1 HES4 KLHL17 NOC2L PUSL1 LINC01342 RNF223 LOC107985728 MXRA8 TTLL10-AS1 MIR6726 ANKRD65 AURKAIP1 MIR6808 AGRN TNFRSF18 DVL1 LOC105378948 LOC100130417 TAS1R3 MIR200A FAM132A LOC284600 LOC100288175 UBE2J2 PERM1 CPTP ACAP3 MRPL20 MIR429 TMEM88B SCNN1D CCNL2 SDF4
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain TNFRSF18 MIR200A FAM132A CPSF3L TNFRSF4 MIR6727 MIR200B UBE2J2 B3GALT6 ACAP3 TTLL10 PUSL1 TTLL10-AS1 MIR6726 MIR429 SCNN1D SDF4

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

snRNA processing
snRNA transcription from RNA polymerase II promoter
protein binding
hydrolase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0