This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00914162_cn
See other CHD1L CNV Assays ›
Gene Symbol
CHD1L
Assay Reference Genome
Location

Chr.1:147187212 on build GRCh38
Cytoband
1q21.1
Assay ID Hs00914162_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9557

Gene Name:

chromodomain helicase DNA binding protein 1 like

Gene Aliases:

ALC1, CHDL

Location:

Chr.1:147172804-147295766 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CHD1L XM_017002858.1 XP_016858347.1

Target Gene Details

Entrez Gene ID:

2330

Gene Name:

flavin containing monooxygenase 5

Gene Aliases:

-

Location:

Chr.1:147183963-147225798 on Build GRCh38

Assay Gene Location:

Overlaps Intron 9 - Exon 10
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FMO5 NM_001144829.2 NP_001138301.1
NM_001144830.2 NP_001138302.1
NM_001461.3 NP_001452.2
XM_005272946.4 XP_005273003.1
XM_005272947.4 XP_005273004.1
XM_005272948.4 XP_005273005.1
XM_006711245.3 XP_006711308.1
XM_011509350.2 XP_011507652.1
XM_011509351.2 XP_011507653.1
XM_017000801.1 XP_016856290.1
XM_017000802.1 XP_016856291.1
AK222728.1 BAD96448.1
AK225739.1
AK314647.1
BC035687.1 AAH35687.1
L37080.1 AAA67849.1
Z47553.1 CAA87633.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv327n100 Chr.1:146028214 - 148860637 on Build GRCh38 Gain NUDT4P2 GJA8 CH17-408M7.1 LINC00624 CHD1L LINC01138 PRKAB2 NBPF11 RNU1-13P RNVU1-8 GJA5 PPIAL4G LOC101927468 RNVU1-6 LOC105371242 PDIA3P1 HYDIN2 LOC100132057 RNVU1-1 LOC728989 NUDT4P1 RNVU1-7 LOC100996763 NBPF12 MIR6077 LOC100505824 PDE4DIP NBPF10 LOC105371226 LOC101928979 NOTCH2NL NBPF14 RNVU1-3 FMO5 GPR89B MIR5087 NBPF13P LOC100996517 ACP6 PDZK1P1 LOC284561 BCL9
nsv1005055 Chr.1:145642988 - 148991068 on Build GRCh38 Loss NUDT4P2 CHD1L PRKAB2 CD160 NBPF11 RNU1-13P GJA5 PPIAL4G LOC101927468 RNVU1-6 LOC105371242 PDIA3P1 HFE2 RNVU1-1 PDZK1 MIR6077 POLR3GL ANKRD34A PDE4DIP LOC101928979 LIX1L MIR5087 POLR3C ITGA10 PDZK1P1 LOC284561 GNRHR2 PIAS3 GJA8 CH17-408M7.1 LINC00624 TXNIP LINC01138 RNVU1-8 RBM8A HYDIN2 RNF115 LOC100132057 MIR6736 LOC728989 NUDT4P1 RNVU1-7 LOC100996763 NBPF12 LOC100505824 PEX11B NBPF10 LOC105371226 ANKRD35 NOTCH2NL NBPF14 RNVU1-3 NUDT17 FMO5 GPR89B NBPF13P LOC100996517 ACP6 GPR89A BCL9
nsv1121713 Chr.1:147010816 - 147207577 on Build GRCh38 Deletion NBPF13P CHD1L PDIA3P1 PRKAB2 RNVU1-8 LOC728989 FMO5
esv2761299 Chr.1:146068341 - 148926801 on Build GRCh38 Gain+Loss NUDT4P2 GJA8 CH17-408M7.1 LINC00624 CHD1L LINC01138 PRKAB2 NBPF11 RNU1-13P RNVU1-8 GJA5 PPIAL4G LOC101927468 LOC105371242 PDIA3P1 HYDIN2 LOC100132057 RNVU1-1 LOC728989 NUDT4P1 RNVU1-7 LOC100996763 NBPF12 MIR6077 LOC100505824 PDE4DIP NBPF10 LOC105371226 NOTCH2NL NBPF14 RNVU1-3 FMO5 GPR89B MIR5087 NBPF13P LOC100996517 ACP6 PDZK1P1 LOC284561 BCL9
nsv509457 Chr.1:145948024 - 149021821 on Build GRCh38 Insertion NUDT4P2 GJA8 CH17-408M7.1 LINC00624 CHD1L TXNIP LINC01138 PRKAB2 NBPF11 RNU1-13P RNVU1-8 GJA5 PPIAL4G LOC101927468 RNVU1-6 LOC105371242 PDIA3P1 HFE2 HYDIN2 LOC100132057 RNVU1-1 LOC728989 NUDT4P1 RNVU1-7 LOC100996763 NBPF12 MIR6077 LOC100505824 POLR3GL ANKRD34A PDE4DIP NBPF10 LOC105371226 LOC101928979 LIX1L NOTCH2NL NBPF14 RNVU1-3 FMO5 GPR89B MIR5087 NBPF13P LOC100996517 ACP6 PDZK1P1 LOC284561 BCL9
nsv1132304 Chr.1:146186094 - 148977674 on Build GRCh38 Duplication NUDT4P2 GJA8 CH17-408M7.1 LINC00624 CHD1L LINC01138 PRKAB2 NBPF11 RNU1-13P RNVU1-8 GJA5 PPIAL4G LOC101927468 LOC105371242 PDIA3P1 HYDIN2 LOC100132057 RNVU1-1 LOC728989 NUDT4P1 RNVU1-7 LOC100996763 NBPF12 MIR6077 LOC100505824 PDE4DIP LOC105371226 NOTCH2NL NBPF14 RNVU1-3 FMO5 GPR89B MIR5087 NBPF13P LOC100996517 ACP6 PDZK1P1 LOC284561 BCL9
nsv1003688 Chr.1:146036817 - 148934526 on Build GRCh38 Gain NUDT4P2 GJA8 CH17-408M7.1 LINC00624 CHD1L LINC01138 PRKAB2 NBPF11 RNU1-13P RNVU1-8 GJA5 PPIAL4G LOC101927468 RNVU1-6 LOC105371242 PDIA3P1 HYDIN2 LOC100132057 RNVU1-1 LOC728989 NUDT4P1 RNVU1-7 LOC100996763 NBPF12 MIR6077 LOC100505824 PDE4DIP NBPF10 LOC105371226 LOC101928979 NOTCH2NL NBPF14 RNVU1-3 FMO5 GPR89B MIR5087 NBPF13P LOC100996517 ACP6 PDZK1P1 LOC284561 BCL9
dgv66e199 Chr.1:146055970 - 149056514 on Build GRCh38 Deletion NUDT4P2 GJA8 CH17-408M7.1 LINC00624 CHD1L LINC01138 PRKAB2 NBPF11 RNU1-13P RNVU1-8 GJA5 PPIAL4G LOC101927468 LOC105371242 PDIA3P1 HYDIN2 LOC100132057 RNVU1-1 LOC728989 NUDT4P1 RNVU1-7 LOC100996763 NBPF12 MIR6077 LOC100505824 PDE4DIP NBPF10 LOC105371226 LOC101928979 NOTCH2NL NBPF14 RNVU1-3 FMO5 GPR89B MIR5087 NBPF13P LOC100996517 NBPF9 ACP6 PDZK1P1 LOC284561 BCL9
nsv437047 Chr.1:147171976 - 147369886 on Build GRCh38 Loss CHD1L PDIA3P1 PRKAB2 FMO5

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
DNA repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
chromatin remodeling
cellular response to DNA damage stimulus
nucleotide-excision repair, DNA incision
global genome nucleotide-excision repair
biological_process
drug metabolic process
oxidation-reduction process
nucleotide binding
nucleic acid binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
ATPase activity
N,N-dimethylaniline monooxygenase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline