This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00935868_cn
See other GNB1 CNV Assays ›
Gene Symbol
GNB1
Assay Reference Genome
Location

Chr.1:1790456 on build GRCh38
Cytoband
1p36.33
Assay ID Hs00935868_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2782

Gene Name:

G protein subunit beta 1

Gene Aliases:

MRD42

Location:

Chr.1:1785285-1891117 on Build GRCh38

Assay Gene Location:

Overlaps Intron 10 - Exon 10
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GNB1 NM_001282538.1 NP_001269467.1
NM_001282539.1 NP_001269468.1
NM_002074.4 NP_002065.1
XM_017001059.1 XP_016856548.1
XM_017001060.1 XP_016856549.1
XM_017001061.1 XP_016856550.1
AF070597.1 AAC28652.1
AF070603.1 AAC28655.1
AF501882.1 AAM15918.1
AK096614.1
AK122949.1
AK312614.1
BC004186.2 AAH04186.1
BC005888.2 AAH05888.1
BC008991.1 AAH08991.1
BC114618.1 AAI14619.1
BT007305.1 AAP35969.1
CR456784.1 CAG33065.1
M36430.1 AAA63265.1
X04526.1 CAA28207.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion SLC35E2 NADK MMP23B SLC35E2B FNDC10 GNB1 CDK11A MMP23A MIB2 CDK11B
nsv834091 Chr.1:1701359 - 1880638 on Build GRCh38 Loss SLC35E2 NADK GNB1 CDK11A MMP23A
nsv951380 Chr.1:1785162 - 1791261 on Build GRCh38 Deletion GNB1
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication SLC35E2 NADK MMP23B SLC35E2B FNDC10 GNB1 CDK11A SSU72 MMP23A MIB2 CDK11B
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC101928626 LOC102725121 SCNN1D LOC107985728 B3GALT6 LOC105378589 LOC107984841 LOC148413 LOC105378947 RNF223 GABRD PUSL1 CDK11A TNFRSF18 CDK11B LOC100134822 TTLL10 CCNL2 SLC35E2B LINC01128 FAAP20 MXRA8 FAM41C SSU72 WASH7P ANKRD65 LOC100129534 MMP23B DDX11L1 MIR6723 MIR1302-2 LOC100130417 NOC2L LOC105378948 MMP23A CPSF3L LOC100288175 LOC100133331 ACAP3 ATAD3B TTLL10-AS1 SKI UBE2J2 OR4F29 TMEM52 FNDC10 OR4F16 MORN1 C1orf159 MRPL20 FAM87B LOC107985729 FAM138A KLHL17 MIR200B CALML6 AGRN MIR6726 LOC284600 MIR6859-2 LINC01342 TNFRSF4 MIR429 LOC729737 LOC100506504 MIB2 LOC100132287 LOC105378591 MIR200A MIR6727 SLC35E2 MIR6859-1 ATAD3C VWA1 GNB1 PERM1 SAMD11 HES4 MTND2P28 LOC105378949 LOC102724312 FAM132A SDF4 TMEM240 LOC100287934 LOC100288069 OR4F5 MTND1P23 LOC105378592 CPTP TMEM88B ATAD3A DVL1 PRKCZ AURKAIP1 NADK TAS1R3 LINC00115 PLEKHN1 ISG15 MIR6808 CFAP74
nsv1160926 Chr.1:1772699 - 1905488 on Build GRCh38 Duplication LOC105378949 NADK GNB1
dgv10n100 Chr.1:1779975 - 1884542 on Build GRCh38 Gain NADK GNB1
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss ANKRD65 SLC35E2 MMP23B ATAD3C TMEM52 FNDC10 VWA1 GNB1 MRPL20 LOC105378589 LOC107985729 LOC105378949 LOC102724312 LOC148413 TMEM240 GABRD CALML6 CDK11A CDK11B TMEM88B CCNL2 SLC35E2B ATAD3A MMP23A MIB2 PRKCZ ATAD3B NADK LOC105378591 SSU72 CFAP74

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

protein folding
signal transduction
adenylate cyclase-activating dopamine receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
G-protein coupled acetylcholine receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
Ras protein signal transduction
cell proliferation
cardiac muscle cell apoptotic process
rhodopsin mediated signaling pathway
platelet activation
sensory perception of taste
retina development in camera-type eye
cellular response to glucagon stimulus
cellular response to prostaglandin E stimulus
cellular response to hypoxia
cellular response to catecholamine stimulus
GTPase activity
signal transducer activity
protein binding
spectrin binding
type 1 angiotensin receptor binding
protein complex binding
GTPase binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline