This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00960573_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1267845 on build GRCh38
Cytoband
1p36.33
Assay ID Hs00960573_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,505.00
Your Price
Online offer:
4505.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Exon 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 2 337 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 2 326 XP_011538918.1
XM_017000238.1 XP_016855727.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1 2 334

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss ACAP3 SCNN1D MIR6726 UBE2J2
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication MIR6727 AGRN TTLL10-AS1 LOC100288175 CPSF3L MIR429 MIR200B PUSL1 TNFRSF18 SDF4 TNFRSF4 C1orf159 B3GALT6 SCNN1D MIR6726 UBE2J2 ACAP3 TTLL10 LOC105378948 RNF223 FAM132A LINC01342 TAS1R3 MIR200A CPTP
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss SAMD11 MIB2 MIR6727 MXRA8 LOC100288175 MIR200B CDK11B PUSL1 TNFRSF18 SDF4 C1orf159 ISG15 B3GALT6 UBE2J2 DVL1 NADK ANKRD65 PLEKHN1 ACAP3 FAM87B SLC35E2 VWA1 PERM1 LINC00115 HES4 SSU72 CDK11A KLHL17 LOC284600 ATAD3B MMP23B LOC102724312 CCNL2 RNF223 LINC01342 MRPL20 ATAD3A MMP23A MIR6808 CPTP AGRN TTLL10-AS1 TMEM88B CPSF3L MIR429 NOC2L FAM41C TNFRSF4 LOC100288069 LINC01128 LOC100287934 SCNN1D LOC107985729 MIR6726 FNDC10 TMEM240 TTLL10 LOC107985728 ATAD3C LOC100130417 SLC35E2B LOC105378948 LOC148413 FAM132A AURKAIP1 TAS1R3 MIR200A
nsv455661 Chr.1:1217251 - 1275912 on Build GRCh38 Loss B3GALT6 FAM132A UBE2J2 SDF4
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss SAMD11 MIB2 MIR6727 LOC100129534 MIR6859-2 MXRA8 LOC105378589 LOC100288175 LOC105378592 LOC105378949 TNFRSF18 OR4F16 OR4F29 ISG15 B3GALT6 FAM138A LOC102725121 UBE2J2 ANKRD65 MIR6859-1 ACAP3 FAM87B SLC35E2 VWA1 PERM1 LOC729737 SSU72 SKI ATAD3B LOC102724312 LINC01342 MRPL20 LOC101928626 CPTP AGRN MIR429 NOC2L FAM41C TNFRSF4 LOC100287934 LOC107985729 MORN1 TMEM240 LOC100133331 MTND2P28 MIR6723 LOC107985728 LOC100130417 LOC148413 FAM132A AURKAIP1 MIR200A OR4F5 LOC100132287 GABRD MIR200B CDK11B PUSL1 SDF4 C1orf159 DVL1 NADK PLEKHN1 LOC100134822 LINC00115 HES4 CDK11A KLHL17 LOC284600 MMP23B CCNL2 RNF223 LOC105378947 ATAD3A MMP23A MIR6808 WASH7P TTLL10-AS1 TMEM88B CPSF3L DDX11L1 GNB1 LOC100288069 LINC01128 MTND1P23 SCNN1D MIR6726 CALML6 FNDC10 TMEM52 CFAP74 FAAP20 MIR1302-2 LOC107984841 PRKCZ TTLL10 ATAD3C SLC35E2B LOC105378948 LOC105378591 TAS1R3 LOC100506504
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion MIB2 MIR6727 MXRA8 TTLL10-AS1 TMEM88B CPSF3L MIR429 MIR200B CDK11B PUSL1 TNFRSF18 SDF4 TNFRSF4 C1orf159 B3GALT6 SCNN1D LOC107985729 MIR6726 UBE2J2 DVL1 FNDC10 ANKRD65 ACAP3 TMEM240 VWA1 TTLL10 SSU72 ATAD3C ATAD3B MMP23B LOC102724312 CCNL2 LOC148413 FAM132A AURKAIP1 LINC01342 TAS1R3 MRPL20 ATAD3A MIR6808 MIR200A CPTP
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss SAMD11 AGRN TTLL10-AS1 LOC100288175 MIR429 MIR200B NOC2L FAM41C TNFRSF18 SDF4 TNFRSF4 LINC01128 C1orf159 ISG15 B3GALT6 UBE2J2 PLEKHN1 FAM87B PERM1 LINC00115 HES4 TTLL10 KLHL17 LOC107985728 LOC284600 LOC100130417 LOC105378948 RNF223 FAM132A LINC01342 MIR200A
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss MIR6727 ACAP3 MXRA8 CPSF3L PUSL1 SCNN1D MIR6726 AURKAIP1 UBE2J2 TAS1R3 DVL1 MIR6808 CPTP
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain SAMD11 MIR6727 AGRN MXRA8 TTLL10-AS1 LOC100288175 TMEM88B CPSF3L MIR429 MIR200B NOC2L PUSL1 TNFRSF18 SDF4 TNFRSF4 C1orf159 ISG15 B3GALT6 SCNN1D MIR6726 UBE2J2 DVL1 ANKRD65 PLEKHN1 ACAP3 PERM1 HES4 TTLL10 KLHL17 LOC107985728 LOC284600 LOC100130417 LOC105378948 CCNL2 LOC148413 RNF223 FAM132A AURKAIP1 LINC01342 TAS1R3 MRPL20 MIR6808 MIR200A CPTP
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain TTLL10-AS1 MIR429 TTLL10 MIR200B TNFRSF18 SDF4 TNFRSF4 LOC105378948 C1orf159 B3GALT6 RNF223 FAM132A SCNN1D UBE2J2 LINC01342 MIR200A
nsv470680 Chr.1:1070426 - 1378635 on Build GRCh38 Loss MIR6727 MXRA8 TTLL10-AS1 CPSF3L MIR429 MIR200B PUSL1 TNFRSF18 SDF4 TNFRSF4 C1orf159 B3GALT6 SCNN1D MIR6726 UBE2J2 DVL1 ACAP3 TTLL10 RNF223 FAM132A AURKAIP1 LINC01342 TAS1R3 MIR6808 MIR200A CPTP
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain MIR6727 ACAP3 TTLL10-AS1 CPSF3L MIR429 TTLL10 MIR200B PUSL1 TNFRSF18 SDF4 TNFRSF4 B3GALT6 FAM132A SCNN1D MIR6726 UBE2J2 MIR200A

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline