This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00979998_cn
See other MUTYH CNV Assays ›
Gene Symbol
MUTYH
Assay Reference Genome
Location

Chr.1:45331504 on build GRCh38
Cytoband
1p34.1
Assay ID Hs00979998_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

4595

Gene Name:

mutY DNA glycosylase

Gene Aliases:

MYH

Location:

Chr.1:45329242-45340925 on Build GRCh38

Assay Gene Location:

Within Exon 19
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MUTYH NM_001048171.1 13 1413 NP_001041636.1
NM_001048172.1 13 1304 NP_001041637.1
NM_001048173.1 13 1301 NP_001041638.1
NM_001048174.1 13 1220 NP_001041639.1
NM_001128425.1 13 1455 NP_001121897.1
NM_001293190.1 13 1416 NP_001280119.1
NM_001293191.1 13 1253 NP_001280120.1
NM_001293192.1 13 1349 NP_001280121.1
NM_001293195.1 14 1370 NP_001280124.1
NM_001293196.1 13 1238 NP_001280125.1
NM_012222.2 13 1446 NP_036354.1
XM_011541497.2 13 1512 XP_011539799.1
XM_011541498.1 13 1290 XP_011539800.1
XM_011541499.1 13 1276 XP_011539801.1
XM_011541500.2 13 1471 XP_011539802.1
XM_011541501.2 14 1385 XP_011539803.1
XM_011541502.2 13 1290 XP_011539804.1
XM_011541503.2 12 1258 XP_011539805.1
XM_011541504.2 13 1339 XP_011539806.1
XM_011541505.2 9 959 XP_011539807.1
XM_011541506.1 9 943 XP_011539808.1
XM_011541507.2 8 755 XP_011539809.2
XM_017001331.1 13 1289 XP_016856820.1
XM_017001332.1 13 1291 XP_016856821.1
XM_017001333.1 13 1344 XP_016856822.1
XM_017001334.1 13 1237 XP_016856823.1
XM_017001335.1 13 1183 XP_016856824.1
XM_017001336.1 12 1267 XP_016856825.1
XM_017001337.1 12 1207 XP_016856826.1
XM_017001338.1 12 1794 XP_016856827.1
XM_017001339.1 12 1245 XP_016856828.1
AB032920.1 13 1364 BAA89336.1
AB032921.1 13 1334 BAA89337.1
AB032922.1 13 1331 BAA89338.1
AB032923.1 13 1267 BAA89339.1
AB032924.1 13 1231 BAA89340.1
AB032925.1 13 1198 BAA89341.1
AB032926.1 14 1348 BAA89342.1
AB032927.1 13 1269 BAA89343.1
AB032928.1 13 1266 BAA89344.1
AB032929.1 13 1202 BAA89345.1
AK293830.1 4 475
BC003178.1 13 1351 AAH03178.1
KU178203.1 13 1197
KU178204.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2750837 Chr.1:45330443 - 45442591 on Build GRCh38 Loss TOE1 TESK2 MUTYH

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

DNA glycosylase

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
mismatch repair
response to oxidative stress
depurination
DNA binding
protein binding
DNA N-glycosylase activity
MutLalpha complex binding
MutLbeta complex binding
MutSalpha complex binding
MutSbeta complex binding
metal ion binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0