This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00994207_cn
See other HES4 CNV Assays ›
Gene Symbol
HES4
Assay Reference Genome
Location

Chr.1:999369 on build GRCh38
Cytoband
1p36.33
Assay ID Hs00994207_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

57801

Gene Name:

hes family bHLH transcription factor 4

Gene Aliases:

bHLHb42

Location:

Chr.1:998962-1001052 on Build GRCh38

Assay Gene Location:

Within Exon 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
HES4 NM_001142467.1 3 633 NP_001135939.1
NM_021170.3 4 555 NP_066993.1
XM_005244771.4 3 1339 XP_005244828.1
AB048791.1 4 356 BAB13510.1
BC012351.1 4 473 AAH12351.1
BI757407.1 3 633

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2762302 Chr.1:914637 - 1106320 on Build GRCh38 Gain C1orf159 LOC100130417 LOC284600 LOC100288175 LOC105378948 NOC2L HES4 SAMD11 AGRN RNF223 LOC107985728 KLHL17 PERM1 PLEKHN1 ISG15
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC105378947 PUSL1 FAM138A CDK11A LOC100288069 MIR6859-2 B3GALT6 LOC102725121 ACAP3 SKI MORN1 CCNL2 PLEKHN1 CALML6 PRKCZ ATAD3A DDX11L1 MIR200A LOC101928626 OR4F29 ATAD3C TNFRSF18 NOC2L TMEM88B MIR1302-2 TTLL10-AS1 TMEM240 GNB1 LOC107984841 ANKRD65 LOC105378592 MMP23B CPSF3L FAAP20 SDF4 ISG15 LINC01342 C1orf159 TAS1R3 LOC107985729 FAM87B LINC00115 OR4F16 LOC100132287 LOC105378949 MXRA8 MIR6808 UBE2J2 LOC729737 LOC100134822 DVL1 SAMD11 GABRD TNFRSF4 LOC105378591 TMEM52 NADK FNDC10 LINC01128 PERM1 LOC105378948 TTLL10 HES4 MIR6726 CFAP74 MTND1P23 OR4F5 SLC35E2B ATAD3B LOC102724312 SCNN1D MIR6727 MIR6859-1 MIB2 RNF223 MMP23A LOC100133331 LOC100506504 MIR6723 MIR429 LOC100130417 MRPL20 FAM132A SSU72 AGRN MIR200B CDK11B LOC148413 LOC107985728 VWA1 CPTP LOC284600 LOC100288175 FAM41C LOC100287934 AURKAIP1 SLC35E2 MTND2P28 LOC105378589 WASH7P LOC100129534 KLHL17
nsv950451 Chr.1:899421 - 1094520 on Build GRCh38 Deletion C1orf159 LOC100130417 LOC284600 LOC100288175 LOC105378948 NOC2L HES4 SAMD11 AGRN RNF223 LOC107985728 KLHL17 PERM1 PLEKHN1 ISG15
nsv428334 Chr.1:874371 - 1220569 on Build GRCh38 Gain MIR429 C1orf159 LOC100130417 LOC105378948 TTLL10 HES4 AGRN MIR200B LOC107985728 PLEKHN1 MIR200A LOC284600 LOC100288175 TNFRSF18 NOC2L FAM41C TTLL10-AS1 SAMD11 TNFRSF4 RNF223 KLHL17 PERM1 SDF4 ISG15 LINC01342
dgv5n100 Chr.1:585989 - 1114424 on Build GRCh38 Gain MIR6723 C1orf159 LOC100130417 LOC105378948 FAM87B LOC105378947 LINC00115 OR4F16 HES4 LOC100288069 AGRN MTND1P23 LOC107985728 PLEKHN1 LOC101928626 LOC284600 LOC100288175 NOC2L FAM41C SAMD11 LOC100287934 RNF223 MTND2P28 LOC107984841 LINC01128 KLHL17 LOC100133331 PERM1 ISG15
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss LOC105378948 TTLL10 PUSL1 HES4 CDK11A LOC100288069 MIR6726 B3GALT6 ACAP3 CCNL2 SLC35E2B PLEKHN1 ATAD3A ATAD3B MIR200A LOC102724312 SCNN1D ATAD3C TNFRSF18 MIR6727 NOC2L TMEM88B TTLL10-AS1 MIB2 TMEM240 RNF223 MMP23A ANKRD65 MMP23B CPSF3L SDF4 ISG15 LINC01342 MIR429 C1orf159 TAS1R3 LOC100130417 LOC107985729 MRPL20 FAM87B LINC00115 FAM132A SSU72 AGRN MIR200B CDK11B LOC148413 LOC107985728 VWA1 CPTP MXRA8 MIR6808 UBE2J2 LOC284600 LOC100288175 DVL1 FAM41C SAMD11 LOC100287934 AURKAIP1 TNFRSF4 SLC35E2 NADK FNDC10 LINC01128 KLHL17 PERM1
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss MIR429 C1orf159 LOC100130417 LOC105378948 FAM87B TTLL10 LINC00115 HES4 FAM132A AGRN B3GALT6 MIR200B LOC107985728 PLEKHN1 MIR200A UBE2J2 LOC284600 LOC100288175 TNFRSF18 NOC2L FAM41C TTLL10-AS1 SAMD11 TNFRSF4 RNF223 LINC01128 KLHL17 PERM1 SDF4 ISG15 LINC01342
nsv827252 Chr.1:998725 - 1001127 on Build GRCh38 Gain HES4
nsv1013524 Chr.1:939510 - 1161955 on Build GRCh38 Gain C1orf159 LOC100288175 LOC105378948 NOC2L HES4 SAMD11 AGRN RNF223 KLHL17 PERM1 PLEKHN1 ISG15 LINC01342
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain MIR429 C1orf159 TAS1R3 LOC100130417 MRPL20 LOC105378948 TTLL10 PUSL1 HES4 MIR6726 FAM132A AGRN B3GALT6 MIR200B ACAP3 LOC148413 LOC107985728 CCNL2 CPTP PLEKHN1 MXRA8 MIR6808 MIR200A UBE2J2 SCNN1D LOC284600 LOC100288175 TNFRSF18 MIR6727 NOC2L TMEM88B DVL1 TTLL10-AS1 SAMD11 AURKAIP1 TNFRSF4 RNF223 ANKRD65 CPSF3L KLHL17 PERM1 SDF4 ISG15 LINC01342
nsv544895 Chr.1:917392 - 1054900 on Build GRCh38 Loss LOC107985728 LOC100130417 LOC284600 NOC2L KLHL17 HES4 PERM1 SAMD11 PLEKHN1 AGRN ISG15
nsv1160644 Chr.1:939652 - 1220663 on Build GRCh38 Deletion MIR200A MIR429 C1orf159 LOC100288175 LOC105378948 TNFRSF18 NOC2L TTLL10 HES4 TTLL10-AS1 SAMD11 TNFRSF4 AGRN RNF223 MIR200B KLHL17 PERM1 PLEKHN1 SDF4 ISG15 LINC01342
nsv832980 Chr.1:848279 - 1007037 on Build GRCh38 Gain LOC107985728 LOC100130417 LOC284600 NOC2L LINC01128 FAM41C KLHL17 HES4 PERM1 SAMD11 PLEKHN1

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
nervous system development
cell differentiation
DNA binding
transcription factor binding
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline