This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01001558_cn
See other FGGY CNV Assays ›
Gene Symbol
FGGY
Assay Reference Genome
Location

Chr.1:59321591 on build GRCh38
Cytoband
1p32.1
Assay ID Hs01001558_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

55277

Gene Name:

FGGY carbohydrate kinase domain containing

Gene Aliases:

-

Location:

Chr.1:59296405-59762730 on Build GRCh38

Assay Gene Location:

Within Exon 7
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FGGY NM_001113411.1 2 254 NP_001106882.1
NM_001244714.1 2 155 NP_001231643.1
NM_018291.3 2 254 NP_060761.3
NR_103473.1 3 212
XM_011541730.1 2 178 XP_011540032.1
XM_011541731.1 2 112 XP_011540033.1
XM_011541732.1 2 155 XP_011540034.1
XM_011541733.1 2 73 XP_011540035.1
XM_011541735.1 2 73 XP_011540037.1
XM_011541736.2 2 283 XP_011540038.1
XM_017001643.1 2 284 XP_016857132.1
XM_017001644.1 2 77 XP_016857133.1
XM_017001645.1 2 178 XP_016857134.1
XM_017001646.1 2 155 XP_016857135.1
XM_017001647.1 2 283 XP_016857136.1
XM_017001649.1 2 223 XP_016857138.1
XM_017001650.1 2 279 XP_016857139.1
XM_017001651.1 2 135 XP_016857140.1
XM_017001652.1 2 223 XP_016857141.1
XM_017001653.1 2 155 XP_016857142.1
XM_017001654.1 2 238 XP_016857143.1
XM_017001655.1 2 147 XP_016857144.1
XM_017001656.1 2 100 XP_016857145.1
XM_017001659.1 XP_016857148.1
XM_017001660.1 XP_016857149.1
XM_017001661.1 2 283 XP_016857150.1
XM_017001663.1 2 143 XP_016857152.1
XM_017001664.1 XP_016857153.1
XM_017001665.1 XP_016857154.1
XM_017001666.1 2 233 XP_016857155.1
XM_017001667.1 2 287 XP_016857156.1
XM_017001669.1 XP_016857158.1
XM_017001671.1 XP_016857160.1
XM_017001672.1 XP_016857161.1
XM_017001673.1 2 280 XP_016857162.1
XM_017001674.1 2 179 XP_016857163.1
XM_017001675.1 XP_016857164.1
XM_017001676.1 2 283 XP_016857165.1
XM_017001679.1 2 223 XP_016857168.1
AK022237.1 2 223 BAB13993.1
AK054842.1 2 146
AK090568.1 3 212
AK315649.1 2 139
BC014947.1 2 139 AAH14947.1
BX647626.1 2 226
DA072156.1 2 157
DA969344.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3586180 Chr.1:59319533 - 59367279 on Build GRCh38 Loss FGGY
nsv829982 Chr.1:59247176 - 59367120 on Build GRCh38 Loss FGGY

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

carbohydrate kinase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
phosphorylation
neuron cellular homeostasis
molecular_function
kinase activity
phosphotransferase activity, alcohol group as acceptor

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bwr8z:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0