Assay Details
Target Gene Details
Entrez Gene ID: | 10207 |
Gene Name: | PATJ, crumbs cell polarity complex component |
Gene Aliases: |
Cipp, INADL, InaD-like, hINADL |
Location: |
Chr.1:61742477-62163920 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
More Information
Set Membership: |
|

Genomic Map