Assay Details
Target Gene Details
Entrez Gene ID: | 83594 |
Gene Name: | nudix hydrolase 12 |
Gene Aliases: |
- |
Location: |
Chr.5:103548855-103562805 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT12 | NM_001300741.1 | NP_001287670.1 | ||
| NM_031438.3 | NP_113626.1 | |||
| XM_005272095.1 | XP_005272152.1 | |||
| XM_005272097.3 | XP_005272154.1 | |||
| AK098066.1 | ||||
| AK304010.1 | ||||
| AL136592.1 | CAB66527.1 | |||
| BC041099.1 | AAH41099.1 | |||
| BU621954.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3606077 | Chr.5:103549971 - 103712728 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map