This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01032829_cn
See other VSTM1 CNV Assays ›
Gene Symbol
VSTM1
Assay Reference Genome
Location

Chr.19:54051463 on build GRCh38
Cytoband
19q13.42
Assay ID Hs01032829_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

284415

Gene Name:

V-set and transmembrane domain containing 1

Gene Aliases:

SIRL-1, SIRL1, UNQ3033

Location:

Chr.19:54040825-54063966 on Build GRCh38

Assay Gene Location:

Overlaps Intron 5 - Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
VSTM1 NM_001288791.1 NP_001275720.1
NM_001288792.1 NP_001275721.1
NM_001288793.1 NP_001275722.1
NM_198481.3 NP_940883.2
NR_110142.1
XM_011526845.1 XP_011525147.1
XM_011526846.1 XP_011525148.1
XM_011526847.1 XP_011525149.1
XM_011526848.1 XP_011525150.1
XM_011526849.1 XP_011525151.1
XM_017026666.1 XP_016882155.1
AY358542.1 AAQ88906.1
BC100943.2 AAI00944.1
BQ446114.1
DQ479397.1
FJ584316.1
FJ584317.1
FJ584318.1
FJ584319.1
FJ584320.1
FJ882051.1
FN398145.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv458782 Chr.19:53961144 - 54093354 on Build GRCh38 Gain VSTM1 CACNG8 CACNG6 TARM1 MIR935
nsv510773 Chr.19:54017180 - 54132236 on Build GRCh38 Deletion VSTM1 OSCAR PRPF31 NDUFA3 TFPT TARM1
nsv182 Chr.19:54050737 - 54097924 on Build GRCh38 Deletion VSTM1 OSCAR TARM1
nsv435707 Chr.19:54050726 - 54058212 on Build GRCh38 Deletion VSTM1
nsv1071550 Chr.19:54050645 - 54057446 on Build GRCh38 Deletion VSTM1
nsv1063127 Chr.19:53926876 - 54068977 on Build GRCh38 Gain VSTM1 CACNG8 CACNG7 CACNG6 MIR935
nsv9750 Chr.19:54049927 - 54058628 on Build GRCh38 Gain VSTM1
esv2718812 Chr.19:53184636 - 54322450 on Build GRCh38 Deletion MIR518D MIR371A DPRX LILRA6 MIR371B CACNG6 BIRC8 MIR1323 MIR1283-2 MIR373 MIR517A MIR520G ZNF331 LOC284379 VSTM1 LENG1 MIR512-1 MIR518F MIR521-1 PRKCG NDUFA3 MIR526A2 LILRB3 MIR517B MIR525 MIR519A2 TARM1 TSEN34 MIR520C MYADM LILRB2 ZNF818P CNOT3 MIR527 MIR520F MIR526A1 ZNF813 MIR524 MIR372 MIR516A2 PRPF31 MIR519D TFPT MIR520A VN1R4 MIR521-2 TMC4 NLRP12 OSCAR MIR1283-1 MIR512-2 MIR523 ZNF665 ZNF761 MIR516B2 RPS9 MIR515-1 MIR515-2 MBOAT7 MIR520H ZNF845 CACNG8 MIR4752 LILRA5 MIR518A1 MIR498 LILRB5 MIR518E MIR519B MIR520E MIR518C FAM90A27P MIR522 ZNF525 ZNF677 MIR516B1 MIR517C ZNF765 VN1R2 CACNG7 MIR519C MIR518B TPM3P9 LOC105372457 MIR519E MIR520D MIR516A1 MIR520B MIR526B MIR518A2 MIR519A1 MIR935
dgv6541n54 Chr.19:54049610 - 54057172 on Build GRCh38 Loss VSTM1
esv2552009 Chr.19:54051426 - 54057890 on Build GRCh38 Deletion VSTM1
nsv833877 Chr.19:53951529 - 54117323 on Build GRCh38 Loss VSTM1 OSCAR CACNG8 PRPF31 NDUFA3 TFPT CACNG6 TARM1 MIR935

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

immune system process
cytokine activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline