This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01039918_cn
See other NOC2L CNV Assays ›
Gene Symbol
NOC2L
Assay Reference Genome
Location

Chr.1:957011 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01039918_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

26155

Gene Name:

NOC2 like nucleolar associated transcriptional repressor

Gene Aliases:

NET15, NET7, NIR, PPP1R112

Location:

Chr.1:944203-959299 on Build GRCh38

Assay Gene Location:

Overlaps Intron 3 - Exon 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NOC2L NM_015658.3 NP_056473.2
AK024284.1
AK225239.1
AK315080.1
AL050019.1 CAB43240.2
BC003555.1 AAH03555.1
DA002961.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2762302 Chr.1:914637 - 1106320 on Build GRCh38 Gain SAMD11 HES4 LOC100288175 ISG15 KLHL17 PERM1 LOC100130417 NOC2L AGRN LOC107985728 PLEKHN1 LOC105378948 LOC284600 C1orf159 RNF223
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss MTND1P23 CFAP74 LOC105378949 LOC107985729 ISG15 MIR6726 TTLL10-AS1 NOC2L MIR6859-1 MTND2P28 MIR1302-2 ATAD3A MXRA8 GABRD FNDC10 MMP23B LOC101928626 CPSF3L PERM1 LOC729737 LOC100506504 TMEM88B MRPL20 FAM132A LOC100133331 MORN1 LOC100132287 MIR6859-2 OR4F29 KLHL17 LOC100130417 CCNL2 LOC102725121 LOC100129534 MIB2 LOC107984841 PRKCZ DDX11L1 VWA1 MIR200B TMEM52 MIR429 OR4F16 SKI LOC100287934 TNFRSF18 LOC105378948 MMP23A ATAD3B SLC35E2B C1orf159 NADK FAM87B AURKAIP1 LINC01342 FAM41C SDF4 TAS1R3 CDK11A LOC107985728 PLEKHN1 OR4F5 MIR200A LOC148413 FAAP20 LOC100134822 ANKRD65 PUSL1 MIR6727 AGRN ATAD3C CDK11B GNB1 LINC01128 CPTP FAM138A LINC00115 LOC105378592 DVL1 SAMD11 MIR6723 MIR6808 ACAP3 SSU72 LOC105378947 SCNN1D TNFRSF4 SLC35E2 LOC284600 LOC102724312 LOC100288069 HES4 LOC100288175 CALML6 WASH7P LOC105378589 LOC105378591 TTLL10 TMEM240 RNF223 B3GALT6 UBE2J2
nsv950451 Chr.1:899421 - 1094520 on Build GRCh38 Deletion SAMD11 HES4 LOC100288175 ISG15 KLHL17 PERM1 LOC100130417 NOC2L AGRN LOC107985728 PLEKHN1 LOC105378948 LOC284600 C1orf159 RNF223
nsv509035 Chr.1:891406 - 985724 on Build GRCh38 Insertion LOC107985728 PLEKHN1 SAMD11 LOC284600 KLHL17 PERM1 LOC100130417 NOC2L
nsv428334 Chr.1:874371 - 1220569 on Build GRCh38 Gain LINC01342 FAM41C SAMD11 SDF4 ISG15 KLHL17 LOC100130417 TTLL10-AS1 NOC2L LOC107985728 PLEKHN1 MIR200A TNFRSF4 LOC284600 MIR200B MIR429 HES4 LOC100288175 PERM1 AGRN TNFRSF18 LOC105378948 C1orf159 TTLL10 RNF223
dgv5n100 Chr.1:585989 - 1114424 on Build GRCh38 Gain LOC100133331 LINC00115 MTND1P23 FAM41C SAMD11 MIR6723 ISG15 KLHL17 LOC100130417 NOC2L LOC107985728 PLEKHN1 MTND2P28 LOC105378947 LOC107984841 LOC284600 LOC101928626 LOC100288069 HES4 LOC100288175 PERM1 AGRN OR4F16 LOC100287934 LOC105378948 LINC01128 C1orf159 FAM87B RNF223
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss LINC01342 FAM41C LOC107985729 SDF4 TAS1R3 ISG15 MIR6726 CDK11A TTLL10-AS1 NOC2L LOC107985728 PLEKHN1 MIR200A LOC148413 ATAD3A MXRA8 ANKRD65 FNDC10 PUSL1 MMP23B MIR6727 CPSF3L PERM1 AGRN ATAD3C CDK11B LINC01128 CPTP TMEM88B MRPL20 FAM132A LINC00115 DVL1 SAMD11 MIR6808 KLHL17 LOC100130417 CCNL2 ACAP3 SSU72 SCNN1D MIB2 TNFRSF4 SLC35E2 LOC284600 LOC102724312 VWA1 MIR200B MIR429 LOC100288069 HES4 LOC100288175 LOC100287934 TNFRSF18 LOC105378948 MMP23A ATAD3B SLC35E2B C1orf159 TTLL10 TMEM240 NADK FAM87B RNF223 B3GALT6 AURKAIP1 UBE2J2
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss FAM132A LINC00115 LINC01342 FAM41C SAMD11 SDF4 ISG15 KLHL17 LOC100130417 TTLL10-AS1 NOC2L LOC107985728 PLEKHN1 MIR200A TNFRSF4 LOC284600 MIR200B MIR429 HES4 LOC100288175 PERM1 AGRN TNFRSF18 LOC105378948 LINC01128 C1orf159 TTLL10 FAM87B RNF223 B3GALT6 UBE2J2
nsv1013524 Chr.1:939510 - 1161955 on Build GRCh38 Gain LINC01342 SAMD11 HES4 LOC100288175 ISG15 KLHL17 PERM1 NOC2L AGRN PLEKHN1 LOC105378948 C1orf159 RNF223
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain FAM132A LINC01342 DVL1 SAMD11 MIR6808 SDF4 TAS1R3 ISG15 KLHL17 LOC100130417 MIR6726 CCNL2 TTLL10-AS1 NOC2L ACAP3 LOC107985728 PLEKHN1 MIR200A SCNN1D LOC148413 TNFRSF4 MXRA8 LOC284600 ANKRD65 MIR200B PUSL1 MIR429 MIR6727 HES4 LOC100288175 CPSF3L PERM1 AGRN TNFRSF18 LOC105378948 C1orf159 CPTP TTLL10 RNF223 TMEM88B B3GALT6 AURKAIP1 MRPL20 UBE2J2
nsv544895 Chr.1:917392 - 1054900 on Build GRCh38 Loss LOC107985728 PLEKHN1 SAMD11 LOC284600 HES4 ISG15 KLHL17 PERM1 LOC100130417 NOC2L AGRN
nsv1160644 Chr.1:939652 - 1220663 on Build GRCh38 Deletion LINC01342 MIR200B SAMD11 MIR429 SDF4 HES4 LOC100288175 ISG15 KLHL17 PERM1 TTLL10-AS1 NOC2L AGRN PLEKHN1 MIR200A TNFRSF18 LOC105378948 TNFRSF4 C1orf159 TTLL10 RNF223
nsv832980 Chr.1:848279 - 1007037 on Build GRCh38 Gain LOC107985728 PLEKHN1 FAM41C SAMD11 LINC01128 LOC284600 HES4 KLHL17 PERM1 LOC100130417 NOC2L

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
negative regulation of B cell apoptotic process
transcription, DNA-templated
apoptotic process
chromatin assembly
nucleolus to nucleoplasm transport
cellular response to UV
negative regulation of histone acetylation
ribosomal large subunit biogenesis
regulation of signal transduction by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway
chromatin binding
transcription corepressor activity
protein binding
nucleosome binding
histone binding
poly(A) RNA binding
repressing transcription factor binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline