Assay Details
Target Gene Details
Entrez Gene ID: | 7075 |
Gene Name: | tyrosine kinase with immunoglobulin like and EGF like domains 1 |
Gene Aliases: |
JTK14, TIE |
Location: |
Chr.1:43300895-43323110 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIE1 | NM_001253357.1 | 4 | 665 | NP_001240286.1 |
| NM_005424.4 | 4 | 748 | NP_005415.1 | |
| XM_005271163.1 | 4 | 691 | XP_005271220.1 | |
| XM_006710869.1 | 4 | 882 | XP_006710932.1 | |
| XM_017002207.1 | 4 | 691 | XP_016857696.1 | |
| AF217976.1 | AAG17219.1 | |||
| AK225560.1 | 4 | 626 | ||
| AK300399.1 | 4 | 555 | ||
| AK300623.1 | 5 | 577 | ||
| AK307639.1 | ||||
| BC038239.1 | 4 | 650 | AAH38239.1 | |
| EU826586.1 | 4 | 571 | ||
| EU826587.1 | 4 | 571 | ||
| EU826588.1 | 4 | 571 | ||
| EU826590.1 | 4 | 571 | ||
| X60957.1 | 4 | 607 | CAA43290.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546128 | Chr.1:43301467 - 43451738 on Build GRCh38 | Loss |
|
| nsv829693 | Chr.1:43129457 - 43312977 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map