This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01070795_cn
See other MIB2 CNV Assays ›
Gene Symbol
MIB2
Assay Reference Genome
Location

Chr.1:1626851 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01070795_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

142678

Gene Name:

mindbomb E3 ubiquitin protein ligase 2

Gene Aliases:

ZZANK1, ZZZ5

Location:

Chr.1:1615188-1630610 on Build GRCh38

Assay Gene Location:

Overlaps Intron 10 - Exon 11
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MIB2 NM_001170686.1 NP_001164157.1
NM_001170687.1 NP_001164158.1
NM_001170688.1 NP_001164159.1
NM_001170689.1 NP_001164160.1
NM_080875.2 NP_543151.2
NR_033183.1
XM_006710372.1 XP_006710435.1
XM_011540731.2 XP_011539033.1
XM_011540736.2 XP_011539038.1
XM_011540737.2 XP_011539039.1
XM_011540741.2 XP_011539043.1
XM_011540742.2 XP_011539044.1
XM_017000349.1 XP_016855838.1
XM_017000350.1 XP_016855839.1
XM_017000351.1 XP_016855840.1
XM_017000352.1 XP_016855841.1
XM_017000353.1 XP_016855842.1
XM_017000354.1 XP_016855843.1
XM_017000355.1 XP_016855844.1
XM_017000356.1 XP_016855845.1
XM_017000357.1 XP_016855846.1
XM_017000358.1 XP_016855847.1
XM_017000359.1 XP_016855848.1
XM_017000360.1 XP_016855849.1
XM_017000361.1 XP_016855850.1
XM_017000362.1 XP_016855851.1
XM_017000363.1 XP_016855852.1
XM_017000364.1 XP_016855853.1
XM_017000365.1 XP_016855854.1
XM_017000366.1 XP_016855855.1
AB074480.1 BAB92950.1
AB076691.1 BAC00992.1
AB076692.1 BAC00993.1
AB076693.1 BAC00994.1
AB097000.1 BAC77353.1
AK091610.1 BAC03707.1
AK095914.1 BAC04646.1
AK096295.1 BAC04752.1
AK097106.1 BAC04952.1
AK098785.1 BAC05413.1
AK122751.1
AK122863.1
AK128167.1
AK128553.1
AK293642.1
AK302757.1
AK309362.1
AL834527.1 CAD39183.1
BC016490.1 AAH16490.1
BC037542.1 AAH37542.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion CDK11B MMP23B MMP23A CDK11A SLC35E2B NADK MIB2 SLC35E2 GNB1 FNDC10
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss CDK11B MMP23B SSU72 MMP23A CDK11A SLC35E2B MIB2 SLC35E2 FNDC10
esv3575582 Chr.1:1624086 - 1721336 on Build GRCh38 Gain CDK11B MMP23B MMP23A CDK11A SLC35E2B MIB2
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss MMP23B MIR6727 MIR429 PUSL1 ATAD3C LOC100288069 HES4 LINC01342 NOC2L MMP23A NADK LOC100287934 RNF223 TTLL10-AS1 SSU72 LOC105378948 FAM132A MIB2 LINC00115 MIR200A ACAP3 VWA1 SAMD11 MIR6726 KLHL17 CDK11A MRPL20 LOC107985729 MIR6808 TNFRSF4 ANKRD65 ATAD3A SCNN1D SLC35E2 AGRN TMEM88B LOC100288175 LOC102724312 LOC284600 ATAD3B CPTP UBE2J2 SLC35E2B ISG15 LOC107985728 FNDC10 FAM41C PLEKHN1 LINC01128 LOC148413 MIR200B TAS1R3 CPSF3L AURKAIP1 LOC100130417 SDF4 CDK11B TNFRSF18 CCNL2 B3GALT6 TTLL10 PERM1 MXRA8 DVL1 FAM87B TMEM240 C1orf159
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss TMEM52 LOC100133331 MIR429 ATAD3C LOC107984841 DDX11L1 LOC100288069 HES4 LINC01342 NOC2L OR4F5 TTLL10-AS1 GABRD LOC102725121 SSU72 LOC105378948 FAM132A CALML6 LINC00115 WASH7P FAAP20 SAMD11 MIR6726 CDK11A MTND1P23 MRPL20 LOC107985729 ANKRD65 LOC100134822 MIR1302-2 SCNN1D TMEM88B LOC102724312 LOC100132287 UBE2J2 SLC35E2B ISG15 LOC107985728 LOC148413 TAS1R3 SDF4 LOC100506504 LOC105378589 PERM1 MXRA8 DVL1 FAM87B GNB1 TMEM240 C1orf159 MMP23B MIR6727 LOC100129534 PUSL1 MIR6859-2 MIR6723 CFAP74 MMP23A NADK LOC100287934 RNF223 MORN1 LOC105378591 OR4F16 MIB2 MIR200A ACAP3 VWA1 SKI LOC105378592 KLHL17 MIR6808 TNFRSF4 ATAD3A SLC35E2 AGRN LOC100288175 LOC284600 LOC101928626 ATAD3B MTND2P28 PRKCZ CPTP FNDC10 FAM41C PLEKHN1 LINC01128 MIR200B CPSF3L AURKAIP1 LOC100130417 LOC729737 CDK11B TNFRSF18 CCNL2 B3GALT6 TTLL10 OR4F29 LOC105378949 MIR6859-1 FAM138A LOC105378947
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication CDK11B MMP23B SSU72 MMP23A CDK11A SLC35E2B NADK MIB2 SLC35E2 GNB1 FNDC10
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion MIR6808 MMP23B MIR6727 TNFRSF4 ANKRD65 ATAD3A MIR429 SCNN1D PUSL1 ATAD3C TMEM88B LOC102724312 ATAD3B LINC01342 CPTP UBE2J2 TTLL10-AS1 FNDC10 LOC148413 SSU72 FAM132A MIR200B TAS1R3 MIB2 CPSF3L AURKAIP1 MIR200A ACAP3 SDF4 VWA1 CDK11B TNFRSF18 CCNL2 B3GALT6 TTLL10 MIR6726 MXRA8 DVL1 MRPL20 TMEM240 C1orf159 LOC107985729
esv2661270 Chr.1:1625365 - 1636030 on Build GRCh38 Deletion CDK11B MMP23B MIB2
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss CDK11B MMP23B SSU72 MMP23A CDK11A SLC35E2B NADK MIB2 SLC35E2 FNDC10
nsv545017 Chr.1:1615322 - 1663759 on Build GRCh38 Loss CDK11B MMP23B SLC35E2B MIB2
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss MMP23B TMEM52 ANKRD65 ATAD3A SLC35E2 ATAD3C TMEM88B LOC102724312 ATAD3B PRKCZ CFAP74 MMP23A SLC35E2B NADK FNDC10 LOC148413 GABRD SSU72 LOC105378591 CALML6 MIB2 VWA1 CDK11B CCNL2 LOC105378589 LOC105378949 CDK11A GNB1 MRPL20 TMEM240 LOC107985729

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
Notch signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
ubiquitin-protein transferase activity
signal transducer activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline