Assay Details
Target Gene Details
Entrez Gene ID: | 931 |
Gene Name: | membrane spanning 4-domains A1 |
Gene Aliases: |
B1, Bp35, CD20, CVID5, LEU-16, MS4A2, S7 |
Location: |
Chr.11:60455809-60470752 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MS4A1 | NM_021950.3 | 3 | 394 | NP_068769.2 |
| NM_152866.2 | 4 | 657 | NP_690605.1 | |
| AK225630.1 | 3 | 356 | ||
| AK292168.1 | 3 | 530 | ||
| AK300025.1 | ||||
| AK310118.1 | 3 | 344 | ||
| AW504475.1 | ||||
| BC002807.2 | 3 | 344 | AAH02807.1 | |
| CD691924.1 | 4 | 417 | ||
| X07203.1 | 3 | 331 | CAA30179.1 | |
| X12530.1 | 3 | 334 | CAA31046.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2744569 | Chr.11:60310239 - 60485227 on Build GRCh38 | Deletion |
|
| dgv51n111 | Chr.11:60447593 - 60463593 on Build GRCh38 | Deletion |
|
| nsv508635 | Chr.11:60422385 - 60487633 on Build GRCh38 | Deletion |
|
| nsv555151 | Chr.11:60448972 - 60472250 on Build GRCh38 | Loss |
|
| dgv391e212 | Chr.11:60455519 - 60463836 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map