This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01093740_cn
See other NOC2L CNV Assays ›
Gene Symbol
NOC2L
Assay Reference Genome
Location

Chr.1:956239 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01093740_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

26155

Gene Name:

NOC2 like nucleolar associated transcriptional repressor

Gene Aliases:

NET15, NET7, NIR, PPP1R112

Location:

Chr.1:944203-959299 on Build GRCh38

Assay Gene Location:

Overlaps Intron 4 - Exon 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NOC2L NM_015658.3 NP_056473.2
AK024284.1
AK225239.1
AK315080.1
AL050019.1 CAB43240.2
BC003555.1 AAH03555.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2762302 Chr.1:914637 - 1106320 on Build GRCh38 Gain LOC284600 ISG15 C1orf159 LOC107985728 AGRN HES4 LOC105378948 NOC2L PERM1 LOC100288175 RNF223 SAMD11 LOC100130417 KLHL17 PLEKHN1
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC101928626 C1orf159 CDK11A SLC35E2B LOC102725121 SAMD11 LOC105378947 TAS1R3 OR4F5 LOC100506504 KLHL17 MTND2P28 OR4F29 LINC01128 HES4 MMP23B MIR6723 ANKRD65 MIR200A SCNN1D RNF223 SSU72 TNFRSF18 LOC105378949 DVL1 SKI TMEM88B MRPL20 TMEM52 MIR6727 FNDC10 MMP23A AGRN MIR6859-2 LOC107984841 WASH7P LOC105378592 ACAP3 LOC105378591 MIB2 LOC100130417 LOC107985729 FAM41C PLEKHN1 PUSL1 ISG15 ATAD3A CDK11B CPTP SLC35E2 LOC100129534 FAAP20 PERM1 LOC100288175 LOC100132287 B3GALT6 LOC729737 LOC100288069 CCNL2 ATAD3C AURKAIP1 MXRA8 GNB1 MIR6808 LOC100134822 TMEM240 CFAP74 ATAD3B MIR1302-2 LOC148413 MORN1 NADK GABRD OR4F16 LOC107985728 FAM132A MTND1P23 MIR6859-1 LOC105378948 SDF4 LOC100287934 NOC2L LOC100133331 LINC00115 MIR200B LINC01342 PRKCZ TNFRSF4 FAM138A LOC102724312 MIR6726 TTLL10 UBE2J2 MIR429 VWA1 FAM87B CPSF3L LOC284600 TTLL10-AS1 CALML6 DDX11L1 LOC105378589
nsv950451 Chr.1:899421 - 1094520 on Build GRCh38 Deletion LOC284600 ISG15 C1orf159 LOC107985728 AGRN HES4 LOC105378948 NOC2L PERM1 LOC100288175 RNF223 SAMD11 LOC100130417 KLHL17 PLEKHN1
nsv509035 Chr.1:891406 - 985724 on Build GRCh38 Insertion NOC2L LOC284600 PERM1 LOC107985728 SAMD11 LOC100130417 KLHL17 PLEKHN1
nsv428334 Chr.1:874371 - 1220569 on Build GRCh38 Gain C1orf159 AGRN TTLL10 MIR429 SAMD11 LOC100130417 KLHL17 FAM41C PLEKHN1 LOC284600 ISG15 LOC107985728 TTLL10-AS1 HES4 LOC105378948 SDF4 NOC2L MIR200B PERM1 LINC01342 MIR200A LOC100288175 RNF223 TNFRSF18 TNFRSF4
dgv5n100 Chr.1:585989 - 1114424 on Build GRCh38 Gain LOC101928626 C1orf159 AGRN LOC107984841 SAMD11 LOC100130417 LOC105378947 KLHL17 MTND2P28 FAM41C FAM87B PLEKHN1 LOC284600 OR4F16 ISG15 LINC01128 LOC107985728 MTND1P23 HES4 LOC105378948 MIR6723 LOC100287934 NOC2L LOC100133331 LINC00115 PERM1 LOC100288175 RNF223 LOC100288069
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss ATAD3C AURKAIP1 C1orf159 MXRA8 MIR6808 CDK11A TMEM240 SLC35E2B ATAD3B SAMD11 TAS1R3 LOC148413 KLHL17 NADK LINC01128 LOC107985728 FAM132A HES4 LOC105378948 SDF4 MMP23B LOC100287934 NOC2L ANKRD65 LINC00115 MIR200B LINC01342 MIR200A SCNN1D RNF223 SSU72 TNFRSF18 DVL1 TNFRSF4 TMEM88B MRPL20 MIR6727 FNDC10 LOC102724312 MMP23A MIR6726 AGRN ACAP3 TTLL10 UBE2J2 MIB2 MIR429 LOC100130417 LOC107985729 VWA1 FAM41C FAM87B PLEKHN1 CPSF3L LOC284600 PUSL1 ISG15 ATAD3A TTLL10-AS1 CDK11B CPTP SLC35E2 PERM1 LOC100288175 B3GALT6 LOC100288069 CCNL2
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss C1orf159 AGRN TTLL10 UBE2J2 MIR429 SAMD11 LOC100130417 KLHL17 FAM41C FAM87B PLEKHN1 LOC284600 ISG15 LINC01128 LOC107985728 FAM132A TTLL10-AS1 HES4 LOC105378948 SDF4 NOC2L LINC00115 MIR200B PERM1 LINC01342 MIR200A LOC100288175 RNF223 TNFRSF18 B3GALT6 TNFRSF4
nsv1013524 Chr.1:939510 - 1161955 on Build GRCh38 Gain ISG15 C1orf159 AGRN HES4 LOC105378948 NOC2L PERM1 LINC01342 LOC100288175 RNF223 SAMD11 KLHL17 PLEKHN1
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain AURKAIP1 C1orf159 MXRA8 MIR6726 AGRN MIR6808 ACAP3 TTLL10 UBE2J2 MIR429 SAMD11 LOC100130417 TAS1R3 LOC148413 KLHL17 PLEKHN1 CPSF3L LOC284600 PUSL1 ISG15 LOC107985728 FAM132A TTLL10-AS1 HES4 CPTP LOC105378948 SDF4 NOC2L ANKRD65 MIR200B PERM1 LINC01342 MIR200A LOC100288175 SCNN1D RNF223 TNFRSF18 B3GALT6 DVL1 TNFRSF4 CCNL2 TMEM88B MRPL20 MIR6727
nsv544895 Chr.1:917392 - 1054900 on Build GRCh38 Loss NOC2L LOC284600 ISG15 PERM1 LOC107985728 AGRN SAMD11 LOC100130417 HES4 KLHL17 PLEKHN1
nsv1160644 Chr.1:939652 - 1220663 on Build GRCh38 Deletion ISG15 C1orf159 TTLL10-AS1 AGRN HES4 LOC105378948 SDF4 TTLL10 MIR429 NOC2L MIR200B PERM1 LINC01342 MIR200A LOC100288175 RNF223 TNFRSF18 SAMD11 TNFRSF4 KLHL17 PLEKHN1
nsv832980 Chr.1:848279 - 1007037 on Build GRCh38 Gain NOC2L LOC284600 PERM1 LINC01128 LOC107985728 SAMD11 LOC100130417 HES4 KLHL17 FAM41C PLEKHN1

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
negative regulation of B cell apoptotic process
transcription, DNA-templated
apoptotic process
chromatin assembly
nucleolus to nucleoplasm transport
cellular response to UV
negative regulation of histone acetylation
ribosomal large subunit biogenesis
regulation of signal transduction by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway
chromatin binding
transcription corepressor activity
protein binding
nucleosome binding
histone binding
poly(A) RNA binding
repressing transcription factor binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-65nqn:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline