This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01124975_cn
See other NDRG2 CNV Assays ›
Gene Symbol
NDRG2
Assay Reference Genome
Location

Chr.14:21017589 on build GRCh38
Cytoband
14q11.2
Assay ID Hs01124975_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,496.00
Your Price
Online offer:
4496.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

57447

Gene Name:

NDRG family member 2

Gene Aliases:

SYLD

Location:

Chr.14:21016763-21070872 on Build GRCh38

Assay Gene Location:

Within Exon 21
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NDRG2 NM_001282211.1 15 1132 NP_001269140.1
NM_001282212.1 15 1358 NP_001269141.1
NM_001282213.1 16 1411 NP_001269142.1
NM_001282214.1 16 1399 NP_001269143.1
NM_001282215.1 16 1196 NP_001269144.1
NM_001282216.1 15 1147 NP_001269145.1
NM_001320329.1 16 2064 NP_001307258.1
NM_016250.2 16 1254 NP_057334.1
NM_201535.1 17 1296 NP_963293.1
NM_201536.1 15 1187 NP_963294.1
NM_201537.1 16 1229 NP_963831.1
NM_201538.1 15 1164 NP_963832.1
NM_201539.1 16 1206 NP_963833.1
NM_201540.1 17 1273 NP_963834.1
NM_201541.1 16 1231 NP_963835.1
XM_011536996.2 17 1283 XP_011535298.1
XM_011536997.1 17 1284 XP_011535299.1
XM_011536998.1 17 1274 XP_011535300.1
XM_011536999.1 15 1679 XP_011535301.1
XM_011537001.1 16 1237 XP_011535303.1
XM_011537002.1 16 1383 XP_011535304.1
XM_017021480.1 17 1385 XP_016876969.1
XM_017021481.1 17 1424 XP_016876970.1
XM_017021482.1 17 1433 XP_016876971.1
XM_017021483.1 16 1317 XP_016876972.1
XM_017021484.1 16 1359 XP_016876973.1
XM_017021485.1 17 1424 XP_016876974.1
XM_017021486.1 17 1434 XP_016876975.1
XM_017021487.1 16 1343 XP_016876976.1
XM_017021488.1 16 1384 XP_016876977.1
XM_017021489.1 16 1489 XP_016876978.1
XM_017021490.1 15 1281 XP_016876979.1
XM_017021491.1 15 1316 XP_016876980.1
XM_017021492.1 16 1387 XP_016876981.1
XM_017021493.1 16 1533 XP_016876982.1
XM_017021494.1 18 1289 XP_016876983.1
XM_017021495.1 17 1221 XP_016876984.1
XM_017021496.1 16 1220 XP_016876985.1
XM_017021497.1 11 914 XP_016876986.1
AB033074.1 15 1116
AF087872.1 15 1170 AAM10500.1
AF159092.3 16 1177 AAD43131.2
AF304051.1 17 1296 AAL08624.1
AK024521.1 17 1229
AK057843.1 16 1200
AK057847.1 15 1164
AK090657.1 15 1103
AK096999.1 16 2064
AK130029.1 15 1102
AK293514.1 15 1132
AL136574.1 15 1141 CAB66509.1
BC010458.1 15 1135 AAH10458.1
BC011240.1 16 1156 AAH11240.1
BC093038.1 16 1230 AAH93038.1
BX247987.1 15 1217 CAD62321.1
BX248031.1 13 1006 CAD62350.1
BX647091.1 13 1329
BX647748.1 16 1346
CR749252.1 16 1399 CAH18108.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv428293 Chr.14:20747949 - 21063624 on Build GRCh38 Gain+Loss ECRP SLC39A2 RNASE13 EDDM3A RNASE1 MIR6717 EDDM3B LOC101929718 RNASE3 RNASE8 RNASE2 TPPP2 ARHGEF40 RNASE7 NDRG2 RNASE6 METTL17
nsv832740 Chr.14:20863901 - 21076893 on Build GRCh38 Gain ECRP SLC39A2 RNASE13 MIR6717 LOC101929718 RNASE3 RNASE8 RNASE2 TPPP2 ARHGEF40 RNASE7 NDRG2 METTL17

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cytokine production
signal transduction
regulation of vascular endothelial growth factor production
Wnt signaling pathway
substantia nigra development
cell differentiation
negative regulation of smooth muscle cell proliferation
negative regulation of ERK1 and ERK2 cascade
regulation of platelet-derived growth factor production
molecular_function
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-v4fgr:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline