This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01126099_cn
See other CPSF3L CNV Assays ›
Gene Symbol
CPSF3L
Assay Reference Genome
Location

Chr.1:1313521 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01126099_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,505.00
Your Price
Online offer:
4505.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

54973

Gene Name:

cleavage and polyadenylation specific factor 3-like

Gene Aliases:

CPSF73L, INT11, INTS11, RC-68, RC68

Location:

Chr.1:1311585-1324687 on Build GRCh38

Assay Gene Location:

Overlaps Intron 12 - Exon 12
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CPSF3L NM_001256456.1 NP_001243385.1
NM_001256460.1 NP_001243389.1
NM_001256462.1 NP_001243391.1
NM_001256463.1 NP_001243392.1
NM_017871.5 NP_060341.2
XM_011541647.1 XP_011539949.1
XM_011541648.1 XP_011539950.1
XM_011541650.1 XP_011539952.1
XM_017001557.1 XP_016857046.1
XM_017001558.1 XP_016857047.1
AK000549.1 BAA91246.1
AK021939.1 BAB13943.1
AK023356.1 BAB14541.1
AK056652.1
AK058018.1
AK092190.1
AK123641.1 2 1511
AK291387.1
AK297350.1
AL136813.1 CAB66747.1
BC000675.2 AAH00675.2
BC007978.2 AAH07978.1
BC008041.2 AAH08041.1
BC013904.2 AAH13904.2
BC020199.1 AAH20199.1
CR533557.1 CAG38588.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC102725121 OR4F5 AGRN KLHL17 MTND1P23 FAAP20 ACAP3 LOC284600 MIR429 CCNL2 RNF223 FAM132A TTLL10-AS1 MTND2P28 TNFRSF18 SLC35E2B LOC100129534 LOC100134822 ATAD3C LOC101928626 SCNN1D PERM1 LINC01342 SSU72 WASH7P DVL1 CDK11A FAM41C MIR6859-1 PUSL1 MIB2 MRPL20 MORN1 GNB1 TMEM52 LOC107984841 TMEM88B TAS1R3 LINC00115 OR4F16 FAM87B PRKCZ SLC35E2 MIR200B FAM138A CALML6 UBE2J2 TTLL10 ISG15 SKI LOC105378949 CPTP LOC105378589 HES4 LOC100133331 MXRA8 NADK LOC100506504 TNFRSF4 LINC01128 NOC2L MIR6808 FNDC10 LOC105378592 MIR6859-2 MIR6723 MIR1302-2 ANKRD65 DDX11L1 LOC729737 LOC102724312 TMEM240 LOC107985728 LOC100288069 SAMD11 VWA1 MMP23B LOC100287934 AURKAIP1 OR4F29 B3GALT6 MIR6726 LOC105378948 LOC107985729 GABRD LOC148413 MIR6727 SDF4 ATAD3A LOC100132287 LOC100130417 PLEKHN1 LOC100288175 C1orf159 CFAP74 CPSF3L LOC105378591 ATAD3B LOC105378947 CDK11B MMP23A MIR200A
nsv544971 Chr.1:1294063 - 1351426 on Build GRCh38 Loss PUSL1 ACAP3 MIR6727 DVL1 MIR6726 CPTP CPSF3L MIR6808 TAS1R3
nsv544969 Chr.1:1290683 - 1374597 on Build GRCh38 Loss PUSL1 AURKAIP1 ACAP3 MIR6727 DVL1 MIR6726 CPTP SCNN1D CPSF3L MIR6808 MXRA8 TAS1R3
nsv997291 Chr.1:1306835 - 1376336 on Build GRCh38 Gain PUSL1 AURKAIP1 ACAP3 MIR6727 DVL1 CPTP CPSF3L MIR6808 MXRA8 TAS1R3
nsv470680 Chr.1:1070426 - 1378635 on Build GRCh38 Loss LINC01342 AURKAIP1 DVL1 B3GALT6 MIR6726 CPTP MXRA8 PUSL1 ACAP3 MIR6727 MIR429 TNFRSF4 RNF223 FAM132A MIR6808 SDF4 TAS1R3 TTLL10-AS1 TNFRSF18 C1orf159 CPSF3L MIR200B SCNN1D UBE2J2 TTLL10 MIR200A
dgv6n100 Chr.1:1301750 - 1362354 on Build GRCh38 Gain+Loss PUSL1 ACAP3 MIR6727 DVL1 CPTP CPSF3L MIR6808 MXRA8 TAS1R3
nsv1160788 Chr.1:1282706 - 1376336 on Build GRCh38 Deletion PUSL1 AURKAIP1 ACAP3 MIR6727 DVL1 MIR6726 CPTP SCNN1D CPSF3L MIR6808 MXRA8 TAS1R3
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication LINC01342 B3GALT6 MIR6726 CPTP LOC105378948 AGRN PUSL1 ACAP3 MIR6727 MIR429 TNFRSF4 RNF223 FAM132A SDF4 TAS1R3 TTLL10-AS1 TNFRSF18 LOC100288175 C1orf159 CPSF3L MIR200B SCNN1D UBE2J2 TTLL10 MIR200A
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss CPTP AGRN KLHL17 HES4 MXRA8 NADK ACAP3 LOC284600 MIR429 TNFRSF4 LINC01128 CCNL2 RNF223 NOC2L FAM132A MIR6808 FNDC10 TTLL10-AS1 TNFRSF18 SLC35E2B ANKRD65 LOC102724312 TMEM240 LOC107985728 LOC100288069 ATAD3C SAMD11 SCNN1D VWA1 PERM1 MMP23B LOC100287934 LINC01342 AURKAIP1 SSU72 DVL1 B3GALT6 MIR6726 LOC105378948 CDK11A FAM41C LOC107985729 PUSL1 LOC148413 MIB2 MIR6727 MRPL20 SDF4 TMEM88B TAS1R3 ATAD3A LINC00115 LOC100130417 PLEKHN1 LOC100288175 C1orf159 FAM87B CPSF3L SLC35E2 ATAD3B MIR200B CDK11B UBE2J2 MMP23A TTLL10 MIR200A ISG15
esv3890636 Chr.1:1304555 - 1353203 on Build GRCh38 Loss PUSL1 ACAP3 MIR6727 DVL1 CPTP CPSF3L MIR6808 MXRA8 TAS1R3
nsv1000169 Chr.1:1281678 - 1362354 on Build GRCh38 Gain PUSL1 ACAP3 MIR6727 DVL1 MIR6726 CPTP SCNN1D CPSF3L MIR6808 MXRA8 TAS1R3
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion LINC01342 AURKAIP1 SSU72 DVL1 B3GALT6 MIR6726 CPTP MXRA8 LOC107985729 PUSL1 LOC148413 MIB2 ACAP3 MIR6727 MIR429 TNFRSF4 CCNL2 MRPL20 FAM132A MIR6808 SDF4 FNDC10 TMEM88B TAS1R3 TTLL10-AS1 ATAD3A TNFRSF18 ANKRD65 C1orf159 CPSF3L LOC102724312 TMEM240 ATAD3C ATAD3B MIR200B CDK11B SCNN1D UBE2J2 VWA1 TTLL10 MMP23B MIR200A
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss AURKAIP1 DVL1 MIR6726 CPTP CPSF3L MXRA8 PUSL1 ACAP3 MIR6727 SCNN1D UBE2J2 MIR6808 TAS1R3
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain LINC01342 AURKAIP1 DVL1 B3GALT6 MIR6726 CPTP LOC105378948 AGRN KLHL17 HES4 MXRA8 PUSL1 LOC148413 ACAP3 MIR6727 LOC284600 MIR429 TNFRSF4 CCNL2 RNF223 MRPL20 NOC2L FAM132A MIR6808 SDF4 TMEM88B TAS1R3 TTLL10-AS1 TNFRSF18 LOC100130417 PLEKHN1 LOC100288175 ANKRD65 C1orf159 CPSF3L LOC107985728 SAMD11 MIR200B SCNN1D UBE2J2 PERM1 TTLL10 MIR200A ISG15
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain TTLL10-AS1 TNFRSF18 B3GALT6 MIR6726 CPSF3L PUSL1 MIR200B ACAP3 MIR6727 MIR429 TNFRSF4 SCNN1D UBE2J2 FAM132A TTLL10 MIR200A SDF4

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

snRNA processing
snRNA transcription from RNA polymerase II promoter
protein binding
hydrolase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0