This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01138536_cn
See other NOC2L CNV Assays ›
Gene Symbol
NOC2L
Assay Reference Genome
Location

Chr.1:955944 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01138536_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

26155

Gene Name:

NOC2 like nucleolar associated transcriptional repressor

Gene Aliases:

NET15, NET7, NIR, PPP1R112

Location:

Chr.1:944203-959299 on Build GRCh38

Assay Gene Location:

Overlaps Intron 6 - Exon 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NOC2L NM_015658.3 NP_056473.2
AK024284.1
AK225239.1
AK315080.1
AL050019.1 CAB43240.2
BC003555.1 AAH03555.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2762302 Chr.1:914637 - 1106320 on Build GRCh38 Gain ISG15 LOC107985728 NOC2L LOC105378948 LOC100288175 PLEKHN1 LOC284600 AGRN PERM1 RNF223 LOC100130417 C1orf159 KLHL17 HES4 SAMD11
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss MIR6859-2 CDK11B MIB2 MIR6859-1 AGRN LOC105378947 TMEM88B MIR200A LOC100288175 MIR6723 TMEM240 DVL1 DDX11L1 LOC101928626 MIR200B OR4F29 CCNL2 LOC100134822 HES4 WASH7P UBE2J2 LOC102725121 LOC729737 LINC00115 SSU72 VWA1 SDF4 MIR6727 TNFRSF4 PUSL1 CDK11A MIR6808 LOC107985729 LOC284600 TMEM52 CPTP FAM138A LOC107984841 MXRA8 LOC100288069 MORN1 ISG15 FAM87B CALML6 PLEKHN1 LOC105378949 LINC01342 LOC100133331 LOC100287934 LOC100132287 LOC100506504 LOC102724312 SKI OR4F16 LOC100129534 FNDC10 TTLL10-AS1 OR4F5 LOC105378589 LOC107985728 TAS1R3 SCNN1D PRKCZ MIR1302-2 PERM1 FAM41C LINC01128 GABRD KLHL17 SLC35E2 TTLL10 MMP23B ACAP3 B3GALT6 TNFRSF18 MMP23A CPSF3L FAAP20 NADK MIR6726 AURKAIP1 C1orf159 MTND1P23 SLC35E2B GNB1 MIR429 LOC105378591 NOC2L LOC105378948 MTND2P28 ANKRD65 ATAD3B ATAD3A RNF223 ATAD3C LOC100130417 LOC105378592 MRPL20 LOC148413 FAM132A SAMD11 CFAP74
nsv950451 Chr.1:899421 - 1094520 on Build GRCh38 Deletion ISG15 LOC107985728 NOC2L LOC105378948 LOC100288175 PLEKHN1 LOC284600 AGRN PERM1 RNF223 LOC100130417 C1orf159 KLHL17 HES4 SAMD11
nsv509035 Chr.1:891406 - 985724 on Build GRCh38 Insertion LOC100130417 LOC107985728 NOC2L KLHL17 PLEKHN1 LOC284600 PERM1 SAMD11
nsv428334 Chr.1:874371 - 1220569 on Build GRCh38 Gain ISG15 PLEKHN1 LINC01342 AGRN C1orf159 MIR200A SDF4 TNFRSF4 MIR429 TTLL10-AS1 LOC107985728 NOC2L LOC105378948 LOC100288175 LOC284600 PERM1 RNF223 FAM41C MIR200B LOC100130417 KLHL17 TTLL10 HES4 TNFRSF18 SAMD11
dgv5n100 Chr.1:585989 - 1114424 on Build GRCh38 Gain ISG15 FAM87B PLEKHN1 AGRN LOC105378947 LOC100133331 LOC100287934 C1orf159 MTND1P23 LINC00115 OR4F16 LOC107985728 NOC2L LOC105378948 LOC100288175 MTND2P28 MIR6723 LOC284600 PERM1 LOC101928626 RNF223 FAM41C LOC100130417 LINC01128 KLHL17 LOC107984841 HES4 LOC100288069 SAMD11
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss ISG15 CDK11B MIB2 FAM87B PLEKHN1 LINC01342 AGRN LOC100287934 TMEM88B LOC102724312 MIR200A FNDC10 TTLL10-AS1 LOC107985728 LOC100288175 TMEM240 TAS1R3 DVL1 SCNN1D PERM1 FAM41C MIR200B LINC01128 KLHL17 SLC35E2 TTLL10 CCNL2 MMP23B HES4 ACAP3 B3GALT6 TNFRSF18 MMP23A CPSF3L UBE2J2 NADK MIR6726 AURKAIP1 C1orf159 SLC35E2B LINC00115 SSU72 VWA1 SDF4 MIR6727 TNFRSF4 MIR429 PUSL1 CDK11A MIR6808 NOC2L LOC105378948 ANKRD65 LOC107985729 LOC284600 ATAD3B ATAD3A RNF223 CPTP ATAD3C LOC100130417 MRPL20 MXRA8 LOC148413 FAM132A LOC100288069 SAMD11
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss ISG15 UBE2J2 FAM87B PLEKHN1 LINC01342 AGRN C1orf159 MIR200A LINC00115 SDF4 TNFRSF4 MIR429 TTLL10-AS1 LOC107985728 NOC2L LOC105378948 LOC100288175 LOC284600 PERM1 RNF223 FAM41C MIR200B LOC100130417 LINC01128 KLHL17 TTLL10 HES4 B3GALT6 TNFRSF18 FAM132A SAMD11
nsv1013524 Chr.1:939510 - 1161955 on Build GRCh38 Gain ISG15 NOC2L LOC105378948 LOC100288175 PLEKHN1 LINC01342 AGRN PERM1 RNF223 C1orf159 KLHL17 HES4 SAMD11
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain ISG15 CPSF3L UBE2J2 MIR6726 PLEKHN1 LINC01342 AGRN AURKAIP1 C1orf159 TMEM88B MIR200A SDF4 MIR6727 TNFRSF4 MIR429 PUSL1 TTLL10-AS1 MIR6808 LOC107985728 NOC2L LOC105378948 LOC100288175 TAS1R3 ANKRD65 DVL1 SCNN1D LOC284600 PERM1 RNF223 CPTP MIR200B LOC100130417 MRPL20 KLHL17 TTLL10 CCNL2 MXRA8 HES4 ACAP3 B3GALT6 LOC148413 TNFRSF18 FAM132A SAMD11
nsv544895 Chr.1:917392 - 1054900 on Build GRCh38 Loss LOC100130417 ISG15 LOC107985728 NOC2L KLHL17 HES4 PLEKHN1 LOC284600 AGRN PERM1 SAMD11
nsv1160644 Chr.1:939652 - 1220663 on Build GRCh38 Deletion ISG15 NOC2L LOC105378948 LOC100288175 PLEKHN1 LINC01342 AGRN PERM1 RNF223 MIR200B C1orf159 KLHL17 TTLL10 HES4 MIR200A SDF4 TNFRSF18 TNFRSF4 SAMD11 MIR429 TTLL10-AS1
nsv832980 Chr.1:848279 - 1007037 on Build GRCh38 Gain LOC100130417 LINC01128 LOC107985728 NOC2L KLHL17 HES4 PLEKHN1 LOC284600 PERM1 SAMD11 FAM41C

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
negative regulation of B cell apoptotic process
transcription, DNA-templated
apoptotic process
chromatin assembly
nucleolus to nucleoplasm transport
cellular response to UV
negative regulation of histone acetylation
ribosomal large subunit biogenesis
regulation of signal transduction by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway
chromatin binding
transcription corepressor activity
protein binding
nucleosome binding
histone binding
poly(A) RNA binding
repressing transcription factor binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline