This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01147611_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1254078 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01147611_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Exon 10
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 7 2094 NP_477515.2
NM_194315.1 8 2142 NP_919296.1
NM_194457.1 6 1963 NP_919439.1
NM_194458.1 7 2223 NP_919440.1
XM_005244718.3 7 2372 XP_005244775.1
XM_005244719.4 7 2260 XP_005244776.1
XM_006710333.3 7 2539 XP_006710396.1
XM_011540613.2 6 1967 XP_011538915.1
XM_011540614.2 7 2325 XP_011538916.1
XM_011540616.2 8 2334 XP_011538918.1
XM_011540617.2 7 2101 XP_011538919.1
XM_017000238.1 8 2205 XP_016855727.1
XM_017000239.1 6 2423 XP_016855728.1
AF116687.2 1 458 AAF71107.1
AK056065.1 7 2004 BAB71086.1
AK098803.1 2 1460
BC094777.1 7 2093 AAH94777.1
BE515187.1 2 182

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication MIR6726 SDF4 ACAP3 TNFRSF18 FAM132A TTLL10 LOC100288175 MIR200A LOC105378948 PUSL1 AGRN CPTP MIR6727 UBE2J2 LINC01342 CPSF3L TAS1R3 TTLL10-AS1 SCNN1D C1orf159 MIR429 B3GALT6 RNF223 TNFRSF4 MIR200B
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss CDK11B SAMD11 SDF4 LOC148413 TNFRSF18 TMEM88B TTLL10 MIR200A PUSL1 AGRN LINC01128 DVL1 UBE2J2 LOC107985728 FAM87B SCNN1D MRPL20 NADK PLEKHN1 AURKAIP1 TMEM240 LOC100130417 MIR200B MIR6726 ATAD3C LOC284600 NOC2L ACAP3 FAM132A PERM1 SLC35E2 LOC100287934 ANKRD65 LOC102724312 ISG15 LOC100288175 LOC105378948 FNDC10 CPTP MIR6727 SLC35E2B LINC01342 MMP23B CPSF3L FAM41C LOC100288069 KLHL17 TAS1R3 TTLL10-AS1 MIR6808 C1orf159 ATAD3B CDK11A ATAD3A LINC00115 MXRA8 MMP23A MIR429 B3GALT6 RNF223 MIB2 VWA1 TNFRSF4 LOC107985729 HES4 CCNL2 SSU72
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss UBE2J2 B3GALT6 MIR6726 SCNN1D SDF4 ACAP3 FAM132A
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss SAMD11 LOC100129534 SDF4 LOC105378591 LOC148413 TNFRSF18 SKI LOC107984841 TTLL10 OR4F16 LINC01128 UBE2J2 SCNN1D NADK FAAP20 PLEKHN1 FAM138A AURKAIP1 MIR200B MIR6726 ATAD3C LOC284600 ACAP3 PERM1 SLC35E2 MTND2P28 LOC102724312 LOC105378947 LOC100288175 FNDC10 MIR6727 SLC35E2B LINC01342 CPSF3L FAM41C LOC100288069 KLHL17 C1orf159 CDK11A MXRA8 CALML6 LOC105378589 B3GALT6 RNF223 TNFRSF4 HES4 CCNL2 SSU72 CDK11B LOC729737 TMEM88B MIR200A PUSL1 AGRN LOC105378592 DVL1 OR4F5 MIR6859-1 LOC105378949 LOC107985728 MTND1P23 TMEM52 FAM87B MRPL20 LOC100506504 LOC102725121 CFAP74 LOC100133331 TMEM240 LOC100130417 MIR6723 LOC100134822 MIR6859-2 NOC2L FAM132A LOC101928626 LOC100287934 ANKRD65 LOC100132287 ISG15 MORN1 OR4F29 MIR1302-2 LOC105378948 WASH7P CPTP GNB1 MMP23B TAS1R3 TTLL10-AS1 MIR6808 GABRD ATAD3B ATAD3A LINC00115 MMP23A MIR429 MIB2 VWA1 LOC107985729 PRKCZ DDX11L1
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss SAMD11 SDF4 LOC284600 NOC2L TNFRSF18 FAM132A PERM1 TTLL10 ISG15 LOC100288175 MIR200A LOC105378948 AGRN LINC01128 UBE2J2 LINC01342 LOC107985728 FAM41C KLHL17 FAM87B TTLL10-AS1 C1orf159 LINC00115 MIR429 B3GALT6 RNF223 PLEKHN1 TNFRSF4 LOC100130417 HES4 MIR200B
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion CDK11B MIR6726 ATAD3C SDF4 ACAP3 LOC148413 TNFRSF18 FAM132A TMEM88B ANKRD65 LOC102724312 TTLL10 MIR200A PUSL1 FNDC10 CPTP DVL1 MIR6727 UBE2J2 LINC01342 MMP23B CPSF3L TAS1R3 TTLL10-AS1 MIR6808 SCNN1D MRPL20 C1orf159 ATAD3B ATAD3A MXRA8 MIR429 B3GALT6 MIB2 VWA1 TNFRSF4 AURKAIP1 TMEM240 LOC107985729 CCNL2 SSU72 MIR200B
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain SAMD11 MIR6726 SDF4 LOC284600 NOC2L ACAP3 LOC148413 TNFRSF18 FAM132A TMEM88B PERM1 ANKRD65 TTLL10 ISG15 LOC100288175 MIR200A LOC105378948 PUSL1 AGRN CPTP DVL1 MIR6727 UBE2J2 LINC01342 LOC107985728 CPSF3L KLHL17 TAS1R3 TTLL10-AS1 MIR6808 SCNN1D MRPL20 C1orf159 MXRA8 MIR429 B3GALT6 RNF223 PLEKHN1 TNFRSF4 AURKAIP1 LOC100130417 HES4 CCNL2 MIR200B
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain UBE2J2 LINC01342 TTLL10-AS1 SCNN1D SDF4 TNFRSF18 C1orf159 FAM132A MIR429 TTLL10 B3GALT6 RNF223 MIR200A LOC105378948 TNFRSF4 MIR200B
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss UBE2J2 LINC01342 MIR6726 TTLL10-AS1 SCNN1D SDF4 ACAP3 TNFRSF18 C1orf159 FAM132A MIR429 TTLL10 B3GALT6 RNF223 MIR200A TNFRSF4 MIR200B
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain UBE2J2 CPSF3L MIR6726 TTLL10-AS1 SCNN1D SDF4 ACAP3 TNFRSF18 FAM132A MIR429 TTLL10 B3GALT6 MIR200A PUSL1 TNFRSF4 MIR6727 MIR200B

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-65nqn:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline