This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01212556_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1255215 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01212556_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Exon 10
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 7 957 NP_477515.2
NM_194315.1 8 1005 NP_919296.1
NM_194457.1 6 826 NP_919439.1
NM_194458.1 7 1086 NP_919440.1
XM_005244718.3 7 1235 XP_005244775.1
XM_005244719.4 7 1123 XP_005244776.1
XM_006710333.3 7 1402 XP_006710396.1
XM_011540613.2 6 830 XP_011538915.1
XM_011540614.2 7 1188 XP_011538916.1
XM_011540616.2 8 1197 XP_011538918.1
XM_011540617.2 7 964 XP_011538919.1
XM_017000238.1 8 1068 XP_016855727.1
XM_017000239.1 6 1286 XP_016855728.1
AF296658.1 7 910 AAK52609.1
AK056065.1 7 867 BAB71086.1
AK075017.1 7 936 BAC11355.1
AK098803.1 2 323
AK291935.1 7 936
AW411508.1 3 485
BC094777.1 7 956 AAH94777.1
BC104890.1 6 781 AAI04891.1
BC113430.1 6 781
BC143657.1 6 781
BM765278.1
BQ881730.1 5 646

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication LINC01342 MIR200A AGRN UBE2J2 PUSL1 FAM132A RNF223 C1orf159 TAS1R3 CPTP MIR6726 CPSF3L MIR429 MIR6727 SCNN1D B3GALT6 MIR200B TTLL10 TNFRSF4 TNFRSF18 TTLL10-AS1 LOC100288175 SDF4 LOC105378948 ACAP3
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss CDK11B SSU72 LOC100130417 UBE2J2 LOC107985729 MMP23A LOC102724312 MMP23B FAM132A C1orf159 FAM87B DVL1 NADK CCNL2 FNDC10 CPSF3L LOC148413 MIR429 ATAD3B LOC100287934 SCNN1D B3GALT6 MIR200B TTLL10 TNFRSF4 KLHL17 PLEKHN1 SAMD11 LINC01128 MIR6808 LOC284600 HES4 SLC35E2B LOC105378948 NOC2L MXRA8 LINC01342 ATAD3A TMEM240 MIR200A AGRN LOC107985728 PUSL1 TMEM88B MRPL20 RNF223 SLC35E2 AURKAIP1 MIB2 TAS1R3 CPTP MIR6726 LINC00115 FAM41C LOC100288069 PERM1 MIR6727 TNFRSF18 TTLL10-AS1 VWA1 ATAD3C ISG15 CDK11A LOC100288175 SDF4 ANKRD65 ACAP3
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss CDK11B SSU72 UBE2J2 LOC102724312 MMP23B C1orf159 LOC105378947 SKI FAM87B DVL1 NADK PRKCZ CPSF3L MIR1302-2 LOC100133331 MIR6859-2 LOC100287934 SCNN1D B3GALT6 LOC729737 MIR200B TMEM52 PLEKHN1 SAMD11 MIR6723 LINC01128 MIR6808 NOC2L MXRA8 ATAD3A MTND2P28 TMEM240 MIR200A OR4F5 LOC107985728 TMEM88B MRPL20 RNF223 AURKAIP1 MIR6859-1 LOC100129534 LOC100506504 TAS1R3 DDX11L1 MIR6726 LINC00115 FAM41C LOC102725121 LOC100288069 LOC105378591 MIR6727 CFAP74 ATAD3C CDK11A LOC100288175 ANKRD65 LOC100130417 LOC107985729 LOC101928626 MMP23A FAM132A FAM138A CCNL2 LOC105378592 GABRD FNDC10 LOC148413 MIR429 GNB1 ATAD3B FAAP20 MTND1P23 TTLL10 TNFRSF4 KLHL17 LOC284600 WASH7P HES4 LOC105378589 SLC35E2B LOC105378948 LINC01342 LOC105378949 AGRN MORN1 PUSL1 LOC100132287 SLC35E2 OR4F29 MIB2 CPTP CALML6 PERM1 LOC100134822 TNFRSF18 TTLL10-AS1 VWA1 ISG15 LOC107984841 SDF4 OR4F16 ACAP3
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss UBE2J2 SCNN1D B3GALT6 SDF4 FAM132A ACAP3 MIR6726
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss LINC01342 MIR200A LOC100130417 AGRN UBE2J2 LOC107985728 FAM132A RNF223 C1orf159 FAM87B MIR429 LINC00115 FAM41C PERM1 B3GALT6 MIR200B TTLL10 TNFRSF4 KLHL17 TNFRSF18 TTLL10-AS1 PLEKHN1 SAMD11 LINC01128 LOC284600 ISG15 HES4 LOC100288175 SDF4 LOC105378948 NOC2L
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion CDK11B MXRA8 LINC01342 ATAD3A TMEM240 SSU72 MIR200A UBE2J2 LOC107985729 PUSL1 TMEM88B LOC102724312 MMP23B FAM132A MRPL20 C1orf159 AURKAIP1 DVL1 MIB2 TAS1R3 CCNL2 CPTP MIR6726 FNDC10 CPSF3L LOC148413 MIR429 ATAD3B MIR6727 SCNN1D B3GALT6 MIR200B TTLL10 TNFRSF4 TNFRSF18 TTLL10-AS1 VWA1 MIR6808 ATAD3C SDF4 ANKRD65 ACAP3
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain MXRA8 LINC01342 MIR200A LOC100130417 AGRN UBE2J2 LOC107985728 PUSL1 TMEM88B FAM132A MRPL20 RNF223 C1orf159 AURKAIP1 DVL1 TAS1R3 CCNL2 CPTP MIR6726 CPSF3L LOC148413 MIR429 PERM1 MIR6727 SCNN1D B3GALT6 MIR200B TTLL10 TNFRSF4 KLHL17 TNFRSF18 TTLL10-AS1 PLEKHN1 SAMD11 MIR6808 LOC284600 ISG15 HES4 LOC100288175 SDF4 LOC105378948 ANKRD65 ACAP3 NOC2L
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain LINC01342 MIR429 MIR200A UBE2J2 SCNN1D B3GALT6 MIR200B TTLL10 TNFRSF4 TNFRSF18 TTLL10-AS1 FAM132A RNF223 C1orf159 SDF4 LOC105378948
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss LINC01342 MIR429 MIR200A UBE2J2 SCNN1D B3GALT6 MIR200B TTLL10 TNFRSF4 TNFRSF18 TTLL10-AS1 FAM132A RNF223 C1orf159 SDF4 ACAP3 MIR6726
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain CPSF3L MIR429 MIR200A MIR6727 UBE2J2 SCNN1D B3GALT6 MIR200B TTLL10 TNFRSF4 PUSL1 TNFRSF18 TTLL10-AS1 FAM132A SDF4 ACAP3 MIR6726

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-65nqn:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline