This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01230592_cn
See other CPSF3L CNV Assays ›
Gene Symbol
CPSF3L
Assay Reference Genome
Location

Chr.1:1311824 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01230592_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

54973

Gene Name:

cleavage and polyadenylation specific factor 3-like

Gene Aliases:

CPSF73L, INT11, INTS11, RC-68, RC68

Location:

Chr.1:1311585-1324687 on Build GRCh38

Assay Gene Location:

Within Exon 19
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CPSF3L NM_001256456.1 19 2390 NP_001243385.1
NM_001256460.1 18 2024 NP_001243389.1
NM_001256462.1 15 1623 NP_001243391.1
NM_001256463.1 15 1614 NP_001243392.1
NM_017871.5 17 1917 NP_060341.2
XM_011541647.1 19 2068 XP_011539949.1
XM_011541648.1 19 2072 XP_011539950.1
XM_011541650.1 17 1797 XP_011539952.1
XM_017001557.1 18 1873 XP_016857046.1
XM_017001558.1 19 1927 XP_016857047.1
AA687684.1 1 252
AK000549.1 9 1061 BAA91246.1
AK021939.1 15 1580 BAB13943.1
AK023356.1 BAB14541.1
AK056652.1 19 2312
AK058018.1 15 1589
AK092190.1 14 1650
AK123641.1 6 2861
AK291387.1 19 2353
AK297350.1
AL136813.1 17 1896 CAB66747.1
BC000675.2 14 1457 AAH00675.2
BC007978.2 17 1879 AAH07978.1
BC008041.2 12 1241 AAH08041.1
BC013904.2 13 1326 AAH13904.2
BC020199.1 12 1241 AAH20199.1
CR533557.1 CAG38588.1
KF274554.1 3 314

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss RNF223 LOC100506504 MIR6727 LOC100133331 KLHL17 LOC100287934 PUSL1 MORN1 LOC100134822 MIR429 ATAD3B LOC102724312 TMEM240 AURKAIP1 MIR200B SAMD11 ACAP3 CFAP74 PERM1 LOC100130417 LOC100288069 SLC35E2 LOC100132287 MIR200A MIR6723 LINC01342 MIR6859-2 LOC107985729 GNB1 CCNL2 ATAD3C MTND1P23 UBE2J2 CDK11B SLC35E2B FAM138A LOC101928626 PRKCZ LOC107984841 FAM132A LINC00115 PLEKHN1 TMEM88B LOC105378591 SCNN1D C1orf159 LOC102725121 TTLL10-AS1 LOC105378948 TMEM52 LOC105378949 CALML6 CPTP FAM41C MMP23A LOC105378592 LINC01128 OR4F29 GABRD NOC2L OR4F5 TTLL10 LOC105378947 MIR6808 DVL1 SDF4 LOC100288175 ANKRD65 TNFRSF4 AGRN MIR1302-2 SSU72 LOC729737 FNDC10 ISG15 DDX11L1 MIB2 TAS1R3 TNFRSF18 ATAD3A CDK11A MRPL20 NADK HES4 SKI WASH7P LOC284600 CPSF3L MTND2P28 FAAP20 FAM87B OR4F16 LOC100129534 LOC148413 VWA1 MIR6859-1 MXRA8 B3GALT6 MIR6726 LOC107985728 LOC105378589 MMP23B
nsv544971 Chr.1:1294063 - 1351426 on Build GRCh38 Loss CPTP DVL1 CPSF3L PUSL1 MIR6727 MIR6808 TAS1R3 MIR6726 ACAP3
nsv544969 Chr.1:1290683 - 1374597 on Build GRCh38 Loss CPTP DVL1 AURKAIP1 CPSF3L PUSL1 MXRA8 MIR6727 MIR6808 TAS1R3 SCNN1D MIR6726 ACAP3
nsv997291 Chr.1:1306835 - 1376336 on Build GRCh38 Gain CPTP DVL1 AURKAIP1 CPSF3L PUSL1 MXRA8 MIR6727 MIR6808 TAS1R3 ACAP3
nsv470680 Chr.1:1070426 - 1378635 on Build GRCh38 Loss TTLL10-AS1 RNF223 UBE2J2 MIR6727 TAS1R3 TNFRSF18 CPTP PUSL1 MIR429 AURKAIP1 CPSF3L FAM132A MIR200B TTLL10 MIR6808 ACAP3 DVL1 SDF4 MIR200A MXRA8 TNFRSF4 B3GALT6 LINC01342 SCNN1D MIR6726 C1orf159
dgv6n100 Chr.1:1301750 - 1362354 on Build GRCh38 Gain+Loss CPTP DVL1 CPSF3L PUSL1 MXRA8 MIR6727 MIR6808 TAS1R3 ACAP3
nsv1160788 Chr.1:1282706 - 1376336 on Build GRCh38 Deletion CPTP DVL1 AURKAIP1 CPSF3L PUSL1 MXRA8 MIR6727 MIR6808 TAS1R3 SCNN1D MIR6726 ACAP3
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication TTLL10-AS1 LOC105378948 RNF223 UBE2J2 MIR6727 TAS1R3 TNFRSF18 CPTP PUSL1 MIR429 CPSF3L FAM132A MIR200B TTLL10 ACAP3 SDF4 LOC100288175 MIR200A TNFRSF4 B3GALT6 LINC01342 SCNN1D MIR6726 C1orf159 AGRN
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss TTLL10-AS1 LOC105378948 RNF223 MIR6727 KLHL17 LOC100287934 CPTP FAM41C PUSL1 MMP23A MIR429 LINC01128 ATAD3B LOC102724312 TMEM240 AURKAIP1 NOC2L MIR200B TTLL10 MIR6808 SAMD11 ACAP3 PERM1 LOC100130417 DVL1 SDF4 LOC100288175 ANKRD65 LOC100288069 SLC35E2 MIR200A TNFRSF4 LINC01342 LOC107985729 CCNL2 AGRN ATAD3C SSU72 UBE2J2 FNDC10 ISG15 CDK11B MIB2 TAS1R3 TNFRSF18 SLC35E2B ATAD3A CDK11A MRPL20 NADK HES4 LOC284600 CPSF3L FAM132A FAM87B LINC00115 PLEKHN1 LOC148413 TMEM88B VWA1 MXRA8 B3GALT6 SCNN1D MIR6726 C1orf159 LOC107985728 MMP23B
esv3890636 Chr.1:1304555 - 1353203 on Build GRCh38 Loss CPTP DVL1 CPSF3L PUSL1 MXRA8 MIR6727 MIR6808 TAS1R3 ACAP3
nsv1000169 Chr.1:1281678 - 1362354 on Build GRCh38 Gain CPTP DVL1 CPSF3L PUSL1 MXRA8 MIR6727 MIR6808 TAS1R3 SCNN1D MIR6726 ACAP3
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion ATAD3C TTLL10-AS1 SSU72 UBE2J2 FNDC10 MIR6727 CDK11B MIB2 TAS1R3 TNFRSF18 ATAD3A CPTP PUSL1 MRPL20 MIR429 ATAD3B LOC102724312 TMEM240 AURKAIP1 CPSF3L FAM132A MIR200B TTLL10 MIR6808 ACAP3 DVL1 SDF4 LOC148413 ANKRD65 TMEM88B VWA1 MIR200A MXRA8 TNFRSF4 B3GALT6 LINC01342 SCNN1D LOC107985729 MIR6726 C1orf159 CCNL2 MMP23B
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss AURKAIP1 CPSF3L UBE2J2 MIR6727 MIR6808 TAS1R3 ACAP3 CPTP DVL1 PUSL1 MXRA8 SCNN1D MIR6726
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain TTLL10-AS1 LOC105378948 RNF223 UBE2J2 ISG15 MIR6727 TAS1R3 KLHL17 TNFRSF18 CPTP PUSL1 MRPL20 MIR429 HES4 LOC284600 AURKAIP1 CPSF3L NOC2L FAM132A MIR200B TTLL10 MIR6808 SAMD11 ACAP3 PERM1 LOC100130417 DVL1 PLEKHN1 SDF4 LOC100288175 LOC148413 ANKRD65 TMEM88B MIR200A MXRA8 TNFRSF4 B3GALT6 LINC01342 SCNN1D MIR6726 C1orf159 LOC107985728 CCNL2 AGRN
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain CPSF3L TTLL10-AS1 UBE2J2 FAM132A MIR200B MIR6727 TTLL10 TNFRSF18 ACAP3 SDF4 PUSL1 MIR200A MIR429 TNFRSF4 B3GALT6 SCNN1D MIR6726

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

snRNA processing
snRNA transcription from RNA polymerase II promoter
protein binding
hydrolase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0