This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01295886_cn
See other CPSF3L CNV Assays ›
Gene Symbol
CPSF3L
Assay Reference Genome
Location

Chr.1:1312270 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01295886_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

54973

Gene Name:

cleavage and polyadenylation specific factor 3-like

Gene Aliases:

CPSF73L, INT11, INTS11, RC-68, RC68

Location:

Chr.1:1311585-1324687 on Build GRCh38

Assay Gene Location:

Within Exon 17
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CPSF3L NM_001256456.1 17 2115 NP_001243385.1
NM_001256460.1 16 1749 NP_001243389.1
NM_001256462.1 13 1348 NP_001243391.1
NM_001256463.1 13 1339 NP_001243392.1
NM_017871.5 15 1642 NP_060341.2
XM_011541647.1 17 1793 XP_011539949.1
XM_011541648.1 17 1797 XP_011539950.1
XM_011541650.1 15 1522 XP_011539952.1
XM_017001557.1 16 1598 XP_016857046.1
XM_017001558.1 17 1652 XP_016857047.1
AK000549.1 7 786 BAA91246.1
AK021939.1 13 1305 BAB13943.1
AK023356.1 15 1560 BAB14541.1
AK056652.1 17 2037
AK058018.1 13 1314
AK092190.1 12 1375
AK123641.1 4 2586
AK291387.1 17 2078
AK297350.1 16 1699
AL136813.1 15 1621 CAB66747.1
BC000675.2 12 1182 AAH00675.2
BC007978.2 15 1604 AAH07978.1
BC008041.2 10 966 AAH08041.1
BC013904.2 11 1051 AAH13904.2
BC020199.1 10 966 AAH20199.1
CR533557.1 15 1563 CAG38588.1
KF274554.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC148413 MIR429 LOC107985729 ANKRD65 LOC105378589 LOC100288175 LOC100287934 MIR6726 TNFRSF4 MIR6808 CALML6 TMEM52 NOC2L LOC729737 TNFRSF18 LINC01128 MIR200B CFAP74 ATAD3C FAM41C SDF4 MIR6859-2 LOC100132287 MIB2 LOC100133331 DDX11L1 MMP23B LOC105378947 PERM1 SSU72 FAM132A MIR6723 KLHL17 LOC105378948 LOC105378591 MIR200A RNF223 CDK11B CPSF3L OR4F29 GNB1 LOC100129534 CDK11A MRPL20 SAMD11 TTLL10 LOC100130417 CPTP ACAP3 LOC102725121 MTND2P28 SLC35E2B MIR1302-2 TAS1R3 LOC107985728 MIR6859-1 FNDC10 B3GALT6 TTLL10-AS1 LOC105378949 LOC102724312 ATAD3B LOC100134822 TMEM88B NADK ATAD3A PRKCZ UBE2J2 FAM87B LOC105378592 HES4 CCNL2 VWA1 MXRA8 AGRN SLC35E2 OR4F16 GABRD MMP23A OR4F5 WASH7P TMEM240 AURKAIP1 PUSL1 LINC01342 SKI LOC284600 MTND1P23 PLEKHN1 SCNN1D MORN1 C1orf159 MIR6727 LOC100288069 FAAP20 LINC00115 ISG15 DVL1 LOC101928626 LOC107984841 FAM138A LOC100506504
nsv544971 Chr.1:1294063 - 1351426 on Build GRCh38 Loss MIR6808 MIR6727 CPTP ACAP3 PUSL1 TAS1R3 CPSF3L MIR6726 DVL1
nsv544969 Chr.1:1290683 - 1374597 on Build GRCh38 Loss MIR6808 MXRA8 MIR6727 CPTP ACAP3 AURKAIP1 SCNN1D PUSL1 TAS1R3 CPSF3L MIR6726 DVL1
nsv997291 Chr.1:1306835 - 1376336 on Build GRCh38 Gain MIR6808 MXRA8 MIR6727 CPTP ACAP3 AURKAIP1 PUSL1 TAS1R3 CPSF3L DVL1
nsv470680 Chr.1:1070426 - 1378635 on Build GRCh38 Loss MIR429 FAM132A AURKAIP1 PUSL1 LINC01342 TAS1R3 B3GALT6 TTLL10-AS1 MIR200A RNF223 SCNN1D CPSF3L MIR6726 TNFRSF4 C1orf159 MIR6808 UBE2J2 MIR6727 TNFRSF18 TTLL10 DVL1 MIR200B MXRA8 CPTP ACAP3 SDF4
dgv6n100 Chr.1:1301750 - 1362354 on Build GRCh38 Gain+Loss MIR6808 MXRA8 MIR6727 CPTP ACAP3 PUSL1 TAS1R3 CPSF3L DVL1
nsv1160788 Chr.1:1282706 - 1376336 on Build GRCh38 Deletion MIR6808 MXRA8 MIR6727 CPTP ACAP3 AURKAIP1 SCNN1D PUSL1 TAS1R3 CPSF3L MIR6726 DVL1
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication MIR429 FAM132A PUSL1 LINC01342 TAS1R3 B3GALT6 TTLL10-AS1 LOC100288175 LOC105378948 MIR200A RNF223 SCNN1D CPSF3L MIR6726 TNFRSF4 C1orf159 UBE2J2 MIR6727 TNFRSF18 TTLL10 MIR200B AGRN CPTP ACAP3 SDF4
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss LOC148413 SLC35E2B MIR429 LOC107985729 TAS1R3 LOC107985728 FNDC10 B3GALT6 ANKRD65 TTLL10-AS1 LOC100288175 LOC102724312 ATAD3B TMEM88B NADK ATAD3A LOC100287934 MIR6726 TNFRSF4 MIR6808 UBE2J2 FAM87B HES4 NOC2L CCNL2 TNFRSF18 LINC01128 MIR200B VWA1 MXRA8 AGRN ATAD3C SLC35E2 FAM41C MMP23A SDF4 MIB2 MMP23B PERM1 SSU72 FAM132A TMEM240 KLHL17 AURKAIP1 PUSL1 LINC01342 LOC284600 LOC105378948 PLEKHN1 MIR200A RNF223 SCNN1D CDK11B CPSF3L C1orf159 CDK11A MIR6727 LOC100288069 MRPL20 SAMD11 LINC00115 TTLL10 ISG15 DVL1 LOC100130417 CPTP ACAP3
esv3890636 Chr.1:1304555 - 1353203 on Build GRCh38 Loss MIR6808 MXRA8 MIR6727 CPTP ACAP3 PUSL1 TAS1R3 CPSF3L DVL1
nsv1000169 Chr.1:1281678 - 1362354 on Build GRCh38 Gain MIR6808 MXRA8 MIR6727 CPTP ACAP3 SCNN1D PUSL1 TAS1R3 CPSF3L MIR6726 DVL1
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion LOC148413 MIR429 SSU72 LOC107985729 FAM132A TMEM240 AURKAIP1 PUSL1 LINC01342 TAS1R3 FNDC10 B3GALT6 ANKRD65 TTLL10-AS1 LOC102724312 ATAD3B MIR200A TMEM88B SCNN1D CDK11B ATAD3A CPSF3L MIR6726 TNFRSF4 C1orf159 MIR6808 UBE2J2 MIR6727 CCNL2 MRPL20 TNFRSF18 TTLL10 DVL1 MIR200B VWA1 MXRA8 ATAD3C CPTP ACAP3 SDF4 MIB2 MMP23B
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss MIR6808 UBE2J2 MIR6727 AURKAIP1 PUSL1 TAS1R3 DVL1 MXRA8 CPTP ACAP3 SCNN1D CPSF3L MIR6726
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain PERM1 LOC148413 MIR429 FAM132A KLHL17 AURKAIP1 PUSL1 LINC01342 TAS1R3 LOC107985728 B3GALT6 ANKRD65 TTLL10-AS1 LOC100288175 LOC284600 LOC105378948 PLEKHN1 MIR200A RNF223 TMEM88B SCNN1D CPSF3L MIR6726 TNFRSF4 C1orf159 MIR6808 UBE2J2 MIR6727 HES4 NOC2L CCNL2 MRPL20 TNFRSF18 SAMD11 TTLL10 ISG15 DVL1 MIR200B MXRA8 AGRN LOC100130417 CPTP ACAP3 SDF4
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain MIR429 UBE2J2 MIR6727 FAM132A PUSL1 TNFRSF18 TTLL10 B3GALT6 TTLL10-AS1 MIR200B MIR200A ACAP3 SCNN1D CPSF3L SDF4 MIR6726 TNFRSF4

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

snRNA processing
snRNA transcription from RNA polymerase II promoter
protein binding
hydrolase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2d9fq:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0