This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01322595_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1257075 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01322595_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Overlaps Intron 7 - Exon 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
XM_017000239.1 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
AW411508.1
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1
BQ881730.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication AGRN LOC100288175 TNFRSF4 FAM132A MIR429 TTLL10 MIR6727 C1orf159 TNFRSF18 CPTP LINC01342 TTLL10-AS1 SDF4 TAS1R3 RNF223 ACAP3 LOC105378948 SCNN1D B3GALT6 MIR6726 UBE2J2 PUSL1 CPSF3L MIR200B MIR200A
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss MMP23B LINC00115 TNFRSF4 CDK11B FNDC10 MIR429 LINC01128 SAMD11 MMP23A C1orf159 TMEM240 MIB2 LINC01342 SDF4 TAS1R3 LOC107985729 LOC148413 LOC284600 FAM41C ANKRD65 B3GALT6 UBE2J2 PUSL1 CDK11A CPSF3L LOC100287934 MRPL20 PERM1 MXRA8 MIR200A AURKAIP1 AGRN LOC100288175 VWA1 LOC107985728 SLC35E2 ATAD3C DVL1 TMEM88B HES4 ATAD3B ATAD3A CCNL2 FAM132A TTLL10 MIR6727 LOC100130417 TNFRSF18 CPTP PLEKHN1 TTLL10-AS1 FAM87B RNF223 ACAP3 MIR6808 LOC105378948 ISG15 SCNN1D NADK NOC2L MIR6726 SLC35E2B MIR200B SSU72 KLHL17 LOC100288069 LOC102724312
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss MMP23B LINC00115 TNFRSF4 OR4F16 CDK11B FNDC10 MIR429 SAMD11 LOC105378592 MMP23A C1orf159 FAM138A OR4F5 TMEM240 TMEM52 MIB2 MIR1302-2 LINC01342 TAS1R3 LOC102725121 CALML6 ANKRD65 UBE2J2 PUSL1 CPSF3L MXRA8 AURKAIP1 AGRN VWA1 MIR6859-1 LOC107985728 DVL1 TMEM88B LOC729737 ATAD3B ATAD3A CCNL2 FAAP20 TTLL10 LOC100130417 TNFRSF18 TTLL10-AS1 ISG15 LOC100506504 LOC100133331 MORN1 SCNN1D NADK LOC105378949 MIR6726 SLC35E2B SKI KLHL17 LOC100132287 MIR6723 LINC01128 LOC107984841 GABRD LOC101928626 SDF4 LOC107985729 LOC148413 LOC284600 OR4F29 FAM41C B3GALT6 CFAP74 CDK11A LOC100287934 LOC105378591 MRPL20 PERM1 MIR200A LOC100288175 MIR6859-2 SLC35E2 ATAD3C LOC105378589 HES4 DDX11L1 FAM132A MIR6727 MTND1P23 CPTP PLEKHN1 PRKCZ FAM87B LOC100134822 RNF223 WASH7P MTND2P28 ACAP3 MIR6808 LOC105378948 GNB1 NOC2L LOC100129534 LOC105378947 MIR200B SSU72 LOC100288069 LOC102724312
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss FAM132A B3GALT6 ACAP3 MIR6726 UBE2J2 SDF4 SCNN1D
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss AGRN LOC100288175 LINC00115 TNFRSF4 LOC107985728 HES4 FAM132A MIR429 TTLL10 LINC01128 LOC100130417 SAMD11 C1orf159 TNFRSF18 PLEKHN1 LINC01342 TTLL10-AS1 SDF4 FAM87B RNF223 LOC284600 LOC105378948 ISG15 FAM41C NOC2L B3GALT6 UBE2J2 MIR200B PERM1 MIR200A KLHL17
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion MMP23B VWA1 TNFRSF4 ATAD3C DVL1 TMEM88B CDK11B FNDC10 ATAD3B ATAD3A CCNL2 FAM132A MIR429 TTLL10 MIR6727 C1orf159 TMEM240 TNFRSF18 CPTP MIB2 LINC01342 TTLL10-AS1 SDF4 TAS1R3 LOC107985729 LOC148413 ACAP3 MIR6808 ANKRD65 SCNN1D B3GALT6 MIR6726 UBE2J2 PUSL1 CPSF3L MIR200B SSU72 MRPL20 MXRA8 MIR200A AURKAIP1 LOC102724312
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain AGRN LOC100288175 TNFRSF4 LOC107985728 DVL1 TMEM88B HES4 CCNL2 FAM132A MIR429 TTLL10 MIR6727 LOC100130417 SAMD11 C1orf159 TNFRSF18 CPTP PLEKHN1 LINC01342 TTLL10-AS1 SDF4 TAS1R3 RNF223 LOC148413 ACAP3 MIR6808 LOC284600 LOC105378948 ISG15 ANKRD65 SCNN1D NOC2L B3GALT6 MIR6726 UBE2J2 PUSL1 CPSF3L MIR200B MRPL20 PERM1 MXRA8 MIR200A KLHL17 AURKAIP1
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain LOC105378948 TNFRSF4 SCNN1D FAM132A MIR429 TTLL10 B3GALT6 C1orf159 UBE2J2 MIR200B TNFRSF18 MIR200A LINC01342 TTLL10-AS1 SDF4 RNF223
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss ACAP3 TNFRSF4 SCNN1D FAM132A MIR429 TTLL10 B3GALT6 MIR6726 C1orf159 UBE2J2 MIR200B TNFRSF18 MIR200A LINC01342 TTLL10-AS1 SDF4 RNF223
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain ACAP3 TNFRSF4 SCNN1D FAM132A MIR429 TTLL10 B3GALT6 MIR6727 MIR6726 UBE2J2 PUSL1 CPSF3L MIR200B TNFRSF18 MIR200A TTLL10-AS1 SDF4

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline