This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01327911_cn
See other MUTYH CNV Assays ›
Gene Symbol
MUTYH
Assay Reference Genome
Location

Chr.1:45333140 on build GRCh38
Cytoband
1p34.1
Assay ID Hs01327911_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

4595

Gene Name:

mutY DNA glycosylase

Gene Aliases:

MYH

Location:

Chr.1:45329242-45340925 on Build GRCh38

Assay Gene Location:

Overlaps Intron 10 - Intron 11
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MUTYH NM_001048171.1 NP_001041636.1
NM_001048172.1 NP_001041637.1
NM_001048173.1 NP_001041638.1
NM_001048174.1 NP_001041639.1
NM_001128425.1 NP_001121897.1
NM_001293190.1 NP_001280119.1
NM_001293191.1 NP_001280120.1
NM_001293192.1 NP_001280121.1
NM_001293195.1 NP_001280124.1
NM_001293196.1 NP_001280125.1
NM_012222.2 NP_036354.1
XM_011541497.2 XP_011539799.1
XM_011541498.1 XP_011539800.1
XM_011541499.1 XP_011539801.1
XM_011541500.2 XP_011539802.1
XM_011541501.2 XP_011539803.1
XM_011541502.2 XP_011539804.1
XM_011541503.2 XP_011539805.1
XM_011541504.2 XP_011539806.1
XM_011541505.2 XP_011539807.1
XM_011541506.1 XP_011539808.1
XM_011541507.2 XP_011539809.2
XM_017001331.1 XP_016856820.1
XM_017001332.1 XP_016856821.1
XM_017001333.1 XP_016856822.1
XM_017001334.1 XP_016856823.1
XM_017001335.1 XP_016856824.1
XM_017001336.1 XP_016856825.1
XM_017001337.1 XP_016856826.1
XM_017001338.1 XP_016856827.1
XM_017001339.1 XP_016856828.1
AB032920.1 BAA89336.1
AB032921.1 BAA89337.1
AB032922.1 BAA89338.1
AB032923.1 BAA89339.1
AB032924.1 BAA89340.1
AB032925.1 BAA89341.1
AB032926.1 BAA89342.1
AB032927.1 BAA89343.1
AB032928.1 BAA89344.1
AB032929.1 BAA89345.1
AK293830.1
AL560568.3
BC003178.1 AAH03178.1
BI818560.1
BM926983.1
HY094590.1
KU178203.1
KU178204.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2750837 Chr.1:45330443 - 45442591 on Build GRCh38 Loss TOE1 TESK2 MUTYH

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

DNA glycosylase

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
mismatch repair
response to oxidative stress
depurination
DNA binding
protein binding
DNA N-glycosylase activity
MutLalpha complex binding
MutLbeta complex binding
MutSalpha complex binding
MutSbeta complex binding
metal ion binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0