This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01382574_cn
See other JMJD4 CNV Assays ›
Gene Symbol
JMJD4
Assay Reference Genome
Location

Chr.1:227735306 on build GRCh38
Cytoband
1q42.13
Assay ID Hs01382574_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

65094

Gene Name:

jumonji domain containing 4

Gene Aliases:

-

Location:

Chr.1:227731189-227735423 on Build GRCh38

Assay Gene Location:

Within Exon 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
JMJD4 NM_001161465.1 1 106 NP_001154937.1
NM_023007.2 1 106 NP_075383.2
XM_011544262.2 1 118 XP_011542564.1
AK022579.1 1 106 BAB14109.1
AK023030.1 1 84 BAB14366.1

Target Gene Details

Entrez Gene ID:

116841

Gene Name:

synaptosome associated protein 47

Gene Aliases:

C1orf142, ESFI5812, HEL-S-290, HEL170, SNAP-47, SVAP1

Location:

Chr.1:227728168-227781231 on Build GRCh38

Assay Gene Location:

Within Exon 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SNAP47 NM_001323930.1 NP_001310859.1
NM_001323931.1 1 311 NP_001310860.1
NM_001323932.1 NP_001310861.1
NM_001323933.1 NP_001310862.1
NM_001323935.1 1 311 NP_001310864.1
NM_053052.3 1 311 NP_444280.2
NR_136654.1
NR_136655.1 1 311
NR_136656.1 1 311
NR_136657.1 1 311
XM_017000231.1 3 3350 XP_016855720.1
XM_017000232.1 2 3156 XP_016855721.1
AY090635.2 1 311 AAM09082.1
BC018760.1 1 150 AAH18760.2
BC032775.1 AAH32775.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1007479 Chr.1:227508167 - 227796765 on Build GRCh38 Gain LOC105373289 JMJD4 ZNF678 SNAP47
nsv826908 Chr.1:227707685 - 228503399 on Build GRCh38 Gain LOC101060022 PRSS38 LOC105373289 GJC2 LOC107985355 C1orf35 HIST3H3 HIST3H2BB MIR3620 C1orf145 MIR5008 LOC100506571 HIST3H2A TRIM11 LOC101927401 ARF1 TRIM17 MIR6742 IBA57 MRPL55 WNT3A SNAP47 RNF187 GUK1 OBSCN WNT9A IBA57-AS1 JMJD4 MIR4666A
esv32853 Chr.1:227692415 - 228670462 on Build GRCh38 Gain RNA5S3 PRSS38 LOC107985355 HIST3H3 RNA5S15 HIST3H2BB MIR3620 C1orf145 MIR5008 LOC100506571 TRIM11 LOC105373132 RNA5S16 TRIM17 RNA5S5 RNA5S6 IBA57 MRPL55 WNT3A RNA5S4 RNA5S10 RNA5S14 BTNL10 IBA57-AS1 RNA5S17 LOC101060022 LOC105373289 RNA5S13 GJC2 C1orf35 HIST3H2A LOC101927401 ARF1 RNA5S2 MIR6742 SNAP47 RNF187 GUK1 RNA5S9 OBSCN RNA5S1 WNT9A RNA5S12 RNA5S7 JMJD4 DUSP5P1 RNA5S11 MIR4666A RNA5S8 RHOU
nsv515496 Chr.1:227724773 - 227780237 on Build GRCh38 Loss JMJD4 SNAP47

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

protein modifying enzyme SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

synaptic vesicle priming
synaptic vesicle fusion to presynaptic active zone membrane
long-term synaptic potentiation
protein binding
SNAP receptor activity
syntaxin binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0